MilliporeSigma
HPLC Analysis of Organic Acids Commonly Found in Wine and Beer on TSKgel® OApak-P and OApak-A
HPLC Analysis of Organic Acids Commonly Found in Wine and Beer on TSKgel® OApak-P and OApak-A application for HPLC
Materials
analytical column
Product No.
Description
Pricing
TSKgel® OApak-A HPLC Column
phase carboxylic acid, L × I.D. 30 cm × 7.8 mm, 7 μm particle size
guard column
Product No.
Description
Pricing
TSKgel® OApak-P Guard Column
phase sulphonic acid, L × I.D. 4 cm × 6 mm, 10 μm particle size
mobile phase component
Product No.
Description
Pricing
Sulfuric acid concentrate
0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
standard
Product No.
Description
Pricing
L-Pyroglutamic acid
≥99.0% (T)
Citric acid
United States Pharmacopeia (USP) Reference Standard
Lactic acid
United States Pharmacopeia (USP) Reference Standard
Malic acid
United States Pharmacopeia (USP) Reference Standard
Succinic acid
United States Pharmacopeia (USP) Reference Standard
Tartaric acid
United States Pharmacopeia (USP) Reference Standard
Pyruvic acid
United States Pharmacopeia (USP) Reference Standard
CONDITIONS
column
TSKgel OApak-A, 30 cm x 7.8 mm I.D., 5 μm particles (816653)
column
TSKgel OApak-P Guard Column, 4 cm x 6 mm I.D., 10 μm particles (816654)
mobile phase
0.75 mM sulfuric acid
flow rate
0.8 mL/min
column temp.
60 °C
detector
conductivity
injection
20 μL
sample
organic acid mixture, see figure for concentrations
Description
Other Notes
Data provided by Tosoh Bioscience LLC.
Legal Information
TSKgel is a registered trademark of Tosoh Corporation