MilliporeSigma
HPLC Analysis of Triacylglycerols in Sacha Inchi Oil on Ascentis® Express C18
HPLC Analysis of Triacylglycerols in Sacha Inchi Oil on Ascentis® Express C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 μm particles (53829-U)
mobile phase
[A] acetonitrile; [B] 2-propanol
gradient
0 min 0%; 50 min 70% B; 55 min 70% B; 56 min 0% B.
flow rate
1 mL/min
detector
APCI/MS (positive), TIC
injection
2 μL
Description
Analysis Note
An Ascentis Express C18 column packed with 2.7 μm Fused-Core particles was used to to perform this analysis.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany