MilliporeSigma
HPLC Analysis of Tricyclic Antidepressants on Ascentis® Express 2.7 μm RP-Amide
HPLC Analysis of Tricyclic Antidepressants on Ascentis® Express 2.7 μm RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
mobile phase component
Product No.
Description
Pricing
Phosphoric acid
85 wt. % in H2O, 99.99% trace metals basis
CONDITIONS
column
Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 μm particles (53913-U)
mobile phase
[A] 25 mM ammonium phophate pH 7.0 with 30% Phosporic Acid; [B] methanol; (30:70, A:B)
flow rate
0.3 mL/min
pressure
3162 psi (218 bar)
column temp.
35 °C
detector
UV, 254 nm
injection
0.5 μL
sample
Amitriptyline, Clomipramine, Desipramine, Doxepin, 
  Imipramine, Norclomipramine, Nortriptyline, 50        
  μg/mL in 60:40 mobile phase A:mobile phase B
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express 2.7 μm RP-Amide for the separation of tricylcic antidepressants. The phase chemistry is well suited to this class of hydrophobic bases and offers improved resolution and peak shape compared to traditional reversed phase C18 analysis. The core shell particle shows an increased speed of analysis compared to the standard Ascentis application.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany