LC/MS Analysis of Sodium, Potassium and Chloride on SeQuant® ZIC-HILIC
CONDITIONS
column
SeQuant ZIC-cHILIC, PEEK, 150 x 4.6 mm, 5 μm, 100 Å (1.50461.0001)
mobile phase
[A] acetonitrile; [B] ammonium acetate 62.5mM pH 5; (80:20, A:B)
flow rate
1.0 mL/min
pressure
1088 psi 75 Bar
column temp.
30 °C
detector
Sedere Sedex 85 ELSD (cell: standard nebulizer)
injection
20 μL
sample
100 μg/mL KCl and 100 μg/mL NaCl
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany