MilliporeSigma
Retention Time Stability after Repeated Injections on SeQuant® ZIC-HILIC
Retention Time Stability after Repeated Injections on SeQuant® ZIC-HILIC application for HPLC
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 50 x 4.6 mm, 5 μm, 200 Å (1.50451.0001)
mobile phase
[A] acetonitrile; [B] 5 mM ammonium acetate; (80:20, A,B)
flow rate
0.5 mL/min
pressure
158 psi (1.1 MPa)
column temp.
24 °C
detector
254 nm
injection
5 μL
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany