MilliporeSigma
Retention vs Particle Porosity (100 and 200 Å) on SeQuant® ZIC-HILIC
Retention vs Particle Porosity (100 and 200 Å) on SeQuant® ZIC-HILIC application for HPLC
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 150 x 2.1 mm, 3.5 μm, 200 Å (1.50448.0001)
mobile phase
[A] acetonitrile; [B] ammonium acetate; (80:20, A,B)
flow rate
0.1 mL/min
column temp.
30 °C
detector
254 nm
injection
5 μL
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany