MilliporeSigma
LC/MS Analysis of Citicoline on SeQuant® ZIC-HILIC
LC/MS Analysis of Citicoline on SeQuant® ZIC-HILIC application for LC-MS
CONDITIONS
column
SeQuant® ZIC-cHILIC, PEEK, 150 x 4.6 mm, 3 μm, 100 Å (1.50661.0001)
mobile phase
[A] acetonitrile; [B] 7.7 g ammonium acetate and dissolve in in 1L MilliQ water; (70:30, A:B)
flow rate
0.75 mL/min
pressure
856 psi (59 bar)
column temp.
30 °C
detector
276 nm
injection
20 μL
sample
Weigh 25 mg citicoline in 50 ml volumetric flask. Dissolve in diluent.
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany