MilliporeSigma
LC/MS Analysis of Equine Performance-Enhancing Dopants GABA and AICAR on Ascentis® Express HILIC
LC/MS Analysis of Equine Performance-Enhancing Dopants GABA and AICAR on Ascentis® Express HILIC application for LC-MS
Materials
Analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
mobile phase component
Product No.
Description
Pricing
Ammonium formate
eluent additive for LC-MS, LiChropur, ≥99.0%
standard
Product No.
Description
Pricing
γ-Aminobutyric acid
≥99%
5-Hydroxy-L-tryptophan
powder
AICAR
≥98% (HPLC), powder
Ethyl-β-D-glucuronide
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
column
Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase
[A] 5 mM ammonium formate in acetonitrile:water (95:5); [B] 5 mM ammonium formate in acetonitrile:water (70:30)
gradient
0 to 70% B in 3 min, held for 1 min
flow rate
0.4 mL/min
column temp.
35 °C
detector
MS, ESI+/ESI-
injection
2 μL
sample
1 μg/mL in acetonitrile
Description
Analysis Note
This study focuses on the analysis of possible dopants used in equine racing, polar compounds requiring HILIC chromatographic separation. The rapid separation of four such compounds is shown here on an Ascentis Express HILIC column. The highest grade solvents provided clean, robust operation. Cerilliant ethyl-β-D-glucuronide CRM was used for reliable quantification.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany