MilliporeSigma
UHPLC Analysis of Deuterated Isomers on Titan C18
UHPLC Analysis of Deuterated Isomers on Titan C18 application for UHPLC
Materials
related product
Product No.
Description
Pricing
Diazepam
p-Xylene
analytical standard
Naphthalene
analytical standard
Toluene
anhydrous, 99.8%
Biphenyl
≥99%
N,N-Dimethylaniline
99%
p-Xylene-d10
99 atom % D
Toluene-d8
anhydrous, 99.6 atom % D
CONDITIONS
column
Titan C18, 10 cm x 2.1 mm I.D., 1.9 μm particles (577124-U)
mobile phase
[A] water; [B] acetonitrile; (50:50, A:B)
flow rate
0.4 mL/min
pressure
5650 psi (390 bar)
column temp.
35 °C
detector
UV, 254 nm
injection
1 μL
Description
Legal Information
null