EN 14105: GC Analysis of Glycerin Impurity in Biodiesel on MET-Biodiesel
Conditions | |
|---|---|
| column | MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µm with integrated 2 m x 0.53 mm I.D. guard (28668-U) |
| oven | 50 °C (1 min), 15 °C/min to 180 °C, 7 °C/min to 230 °C, 30 °C/min to 380 °C (10 min) |
| carrier gas | helium, 3.0 mL/min |
| sample | EN 14105:2003 Monoglyceride Stock Solution (49446-U) plus Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA then diluted in heptane |
| injection | 1 µL, cold on-column |
| detector | FID, 380 °C |
Description | |
|---|---|
| Categories | Analytical Chromatography, GC Applications, Glycerides |
| Featured Industry | Agriculture Biofuels |
| Other Notes | Complementary products: Syringes Vials Gas Purification and Gas Management Inlet Liners, Glass Wool, and Specialized Hand Tools Septa and Specialized Hand Tools Column Ferrules, Nuts, and Specialized Hand Tools |
| suitability | application for GC |
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500