MilliporeSigma
HomeGas Chromatography (GC)Residual Solvents Testing By Gas Chromatography
Residual Solvents Testing By Gas Chromatography
Introduction
Residual solvent testing is a critical component of pharmaceutical quality control, helping ensure that drug substances, excipients, and finished drug products meet established safety and quality requirements. Organic solvents used during synthesis, purification, extraction, and crystallization may remain as trace impurities in the final product if not adequately removed. Because these residual solvents can pose toxicological risks, guidelines such as ICH Q3C(R9) and USP <467> establish acceptable limits for their presence. Gas chromatography (GC), particularly with headspace sampling, is the preferred analytical technique for monitoring residual solvents due to its sensitivity, selectivity, and suitability for volatile compounds.
Classification of residual solvents
To ensure patient safety, the ICH Q3C(R9) guideline classifies residual solvents according to their potential toxicological risk and establishes acceptable exposure limits based on the Permitted Daily Exposure (PDE).1 This risk-based approach helps manufacturers select appropriate solvents during process development while ensuring that residual solvent levels in pharmaceutical products remain within acceptable limits.¹
The guideline categorizes residual solvents into three classes:
  • Class 1: Solvents to be avoided due to unacceptable toxicity or environmental hazards.1
  • Class 2: Solvents to be limited because of their inherent toxicity.1
  • Class 3: Solvents with low toxic potential that present minimal risk at the specified exposure limits.¹
How key regulatory guidelines define residual solvents
ICH Q3C(R9)
"Residual solvents in pharmaceuticals are defined as organic volatile chemicals that are used or produced in the manufacturing of drug substances or excipients, or in the preparation of drug products."1
USP <467>
"For pharmacopeial purposes, residual solvents in pharmaceuticals are defined as organic volatile chemicals that are used or produced in the manufacturing of drug substances, excipients, or dietary ingredients, or in the preparation of drug products or dietary supplement products."2
Why residual solvent testing is essential
Regulatory guidance consistently emphasizes three key principles for residual solvents:
  • Residual solvents do not contribute to the therapeutic efficacy of a drug product.
  • Residual solvent levels should be minimized through appropriate manufacturing processes whenever practical.
  • Residual solvent concentrations should not exceed safety-based limits established from toxicological risk assessments.¹,²
Residual solvent testing is therefore an integral part of pharmaceutical quality control, helping manufacturers demonstrate compliance with regulatory requirements while ensuring patient safety and product quality.
Risks associated with inadequate residual solvent control
Failure to adequately monitor and control residual solvents can have significant implications for both patient safety and pharmaceutical manufacturers. Excess residual solvent levels may increase toxicological risk, lead to product quality concerns, result in regulatory observations or product recalls, and delay product approval or release. Table 1 summarizes reported incidents involving residual solvents and the resulting regulatory or enforcement actions.
No.
Incident
Year(s)
Discovered solvent
(ICH Class)
Outcome category
1
Benzene in sunscreens and aerosol body sprays
2021–23
Benzene, class 1
Limit: 2 ppm 
Voluntary recall /
civil litigation
2
Benzene from benzoyl peroxide (BPO) acne products
2025
Benzene, class 1
Limit: 2 ppm
Voluntary recall /
civil litigation
3
Methanol in hand sanitizers (COVID-19 pandemic)
2020–21
Methanol, class 2
Limit: 3,000 ppm
Criminal conviction /
market withdrawal
4
Residual solvent specification failure and data falsification
2025
Unspecified API residual solvent per ICH Q3C specification
Regulatory warning /
pending enforcement
5
Benzene in carbomer excipients: FDA guidance action
2023
Benzene, class 1
Limit: 2 ppm
Pending enforcement
Table 1.Examples of reported incidents involving residual solvents and their associated regulatory, legal, or commercial outcomes
Regulatory framework overview
ICH Q3C(R9)
The International Council for Harmonisation (ICH) Q3C(R9) guideline, adopted at Step 4 on 24 January 2024, is the primary global standard for controlling residual solvents in pharmaceuticals.
The 2024 revision introduces an important update to Section 3.4 (Analytical Procedures), which states that the volatility of the residual solvent should be considered during analytical method validation. This update reinforces the need to select analytical methods that are appropriate for the physicochemical properties of the target solvents and highlights the importance of optimizing GC methods for reliable residual solvent analysis.¹
ICH Q3C(R9) classifies residual solvents into three risk-based categories:
Class 1 solvents are known or strongly suspected human carcinogens, and environmental hazards. Their use should be avoided whenever possible. But if their use is unavoidable, residual levels must be controlled within the limits specified in ICH Q3C(R9).¹
Solvent
Concentration limit (ppm)
Concern
Benzene
2
Carcinogen
Carbon tetrachloride
4
Toxic; environmental hazard
1,2-Dichloroethane
5
Toxic
1,1-Dichloroethene
8
Toxic
1,1,1-Trichloroethane
1500
Environmental hazard
Table 2.Class 1 residual solvents and their concentration limits according to ICH Q3C(R9)¹
Class 2 solvents include non-genotoxic animal carcinogens and agents of irreversible toxicity, such as neurotoxicity and teratogenicity. These solvents are controlled using permitted daily exposure (PDE) limits.¹
Solvent
PDE (mg/day)
Concentration limit (ppm)
Acetonitrile
4.1
410
Chlorobenzene
3.6
360
Chloroform
0.6
60
Cumene
0.7
70
Cyclohexane
38.8
3880
Cyclopentyl methyl ether
15.0
1500
1,2-Dichloroethene
18.7
1870
Dichloromethane (DCM)
6.0
600
1,2-Dimethoxyethane
1.0
100
N,N-Dimethylacetamide
10.9
1090
N,N-Dimethylformamide (DMF)
8.8
880
1,4-Dioxane
3.8
380
2-Ethoxyethanol
1.6
160
Ethyleneglycol
6.2
620
Formamide
2.2
220
Hexane
2.9
290
Methanol
30.0
3000
2-Methoxyethanol
0.5
50
Methylbutyl ketone
0.5
50
Methylcyclohexane
11.8
1180
Methylisobutylketone
45
4500
N-Methylpyrrolidone (NMP)
5.3
530
Nitromethane
0.5
50
Pyridine
2.0
200
Sulfolane
1.6
160
Toluene
8.9
890
Tetrahydrofuran (THF)
7.2
720
Tetralin
1.0
100
Xylene#
21.7
2170
Cyclopentyl Methyl Ether (CPME)*
15.0
1500
Tertiary-Butyl Alcohol (TBA)*
35.0
3500
2-Methyltetrahydrofuran (2-MTHF)*
-
Class 3
1,1,2-Trichloroethene
0.8
80
#usually 60% m-xylene, 14% p-xylene, 9% o-xylene with 17% ethyl benzene
*Added in Q3C(R8)/R9 revisions
Table 3.List of representative Class 2 solvents
Class 3 solvents have PDE values of ≥50 mg/day and are considered to have low toxic potential. They include solvents such as ethanol, acetone, ethyl acetate, isopropanol, and heptane. When only Class 3 solvents are present at ≤0.5%, a non-specific method, such as loss on drying, may be appropriate. However, gas chromatography is recommended to confirm solvent identity and quantify residual solvent levels.¹
Representative Class 3 solvents: Acetic acid, acetone, anisole, 1-butanol, 2-butanol, butyl acetate, tert-butyl methyl ether, dimethyl sulfoxide, ethanol, heptane, isobutyl acetate, isopropyl acetate, methyl acetate, 3-methyl-1-butanol, methyl ethyl ketone, 2-methyl-1-propanol, 2-methyltetrahydrofuran, and pentane.¹
GC workflow for residual solvent analysis 
Residual solvent analysis by gas chromatography involves a series of interconnected steps to ensure accurate and reliable results. A typical GC workflow includes: 
  • Sample preparation and introduction, typically using headspace sampling
  • Calibration and method verification using primary or secondary reference materials
  • Separation and analysis by gas chromatography
Sample preparation techniques
Two sample preparation techniques are commonly used for residual solvent analysis, each suitable for different sample types and analytical requirements.
Headspace GC is the preferred technique for residual solvent analysis and is recommended in USP <467>. The sample is equilibrated in a sealed vial at an elevated temperature, allowing volatile solvents to partition into the headspace. An aliquot of the headspace vapor is then injected onto the GC column for analysis.
Best suited for:
  • Class 1 and Class 2 volatile solvents
  • Highly volatile analytes that require no pre-concentration
  • Low-concentration samples requiring increased sensitivity via trap-and-release
In direct injection, a liquid sample or a pharmaceutical formulation dissolved in a suitable solvent is injected directly into the GC inlet for chromatographic separation.
Recommended for:
  • Solvents with limited headspace partitioning
  • When pre-concentration steps are not needed
  • DMSO-based matrices, where DMSO is the standard diluent for water-insoluble formulations in USP <467>¹
GC column selection for USP <467> procedures
USP <467> specifies three GC-based analytical procedures (Procedures A, B, and C) for the identification and quantification of residual solvents. Each procedure recommends a specific USP GC phase and corresponding column chemistry to achieve the required separation.¹
Selecting the appropriate USP <467> procedure
  • Begin with Procedure A for initial screening. If all residual solvent levels are below the specified limits, no further analysis is required.
  • If co-eluting peaks or reportable residual solvents are detected, perform Procedure B for confirmation and identification.
  • Once solvent identity is confirmed, use Procedure C for quantification, where applicable.
  • For Class 3 solvents only, identification is not required if the total solvent content is ≤0.5%; if it exceeds 0.5%, the solvents should be identified and quantified.¹
Procedure
Purpose
USP GC phase*
GC capillary column
Procedure A
Screening/identification
G43
OVI-G43/SPB®-624
Procedure B
Confirmation of co-eluting peaks
G16
Supelcowax® 10
Procedure C
Quantitation
G43 or G16
best performer
*G43 Phase: 6% cyanopropylphenyl / 94% dimethylpolysiloxane
G16 Phase: High molecular weight polyethylene glycol with a diepoxide linker (wax phase)
Table 4.Comparison of USP <467> analytical procedures and representative GC columns for residual solvent testing¹
USP stationary phases for residual solvent analysis
  • G43 stationary phase ((Figures 1 and 2A): The G43 stationary phase provides intermediate polarity and enables the separation of most Class 1 and Class 2 residual solvents in a single analysis. Certain solvent pairs, such as toluene and methyl isobutyl ketone (MIBK), intentionally co-elute on this phase. These compounds are subsequently resolved using Procedure B with a G16 column.¹
  • G16 stationary phase (Figure 2B): The G16 stationary phase is a high-polarity polyethylene glycol (PEG)-based wax phase (e.g., Supelcowax® 10). It is used in USP <467> Procedure B to resolve solvent pairs that co-elute on the G43 phase, particularly toluene and methyl isobutyl ketone (MIBK). 
The complementary selectivity of the G16 and G43 phases enables comprehensive analysis of Class 1 and Class 2 residual solvents.¹ Laboratories performing residual solvent analysis should therefore ensure they have access to both G43 and G16 column chemistries to support compendial testing.¹ 
GC-FID chromatogram of Residual Solvents Mixture Class I certified reference material PHR1063 on an SPB™-624 capillary column, showing five separated solvent peaks.
Figure 1.GC-FID chromatograms of Residual Solvents Mixture – Class I certified reference material (PHR1063) illustrating the chromatographic separation on A) SPB™-624 capillary column (25430) Peak assignments are as follow: 1) 1,1-Dichloroethene, 2) 1,1,1-Trichloroethane, 3) Carbon Tetrachloride, 4) Benzene, and 5) 1,2-Dichloroethane.
GC-FID chromatogram of Residual Solvents Mixture Class IIA certified reference material PHR1064 on an SPB™-624 capillary column, showing separation of 15 residual solvents.
GC-FID chromatogram of Residual Solvents Mixture Class IIA certified reference material PHR1064 on a Supelcowax™-10 capillary column, showing separation of 15 residual solvents.
Figure 2.GC-FID chromatograms of Residual Solvents Mixture – Class IIA certified reference material, (PHR1064) illustrating the stationary phase selectivity differences between A) SPB™-624 capillary column (25430) and B) Supelcowax™-10 capillary column (24080-U). Four Class IIA solvents are highlighted in chromatograms A and B. No single column is capable of chromatographically resolving all residual solvents; therefore, the use of orthogonal stationary phases is necessary for confirmatory identification of potential contaminants. Peak assignments are as follows: 1) Methanol, 2) Acetone, 3) Methylene Chloride, 4t) trans-1,2-Dichloroethene, 4c) cis-1,2-Dichloroethene, 5) Tetrahydrofuran, 6) Cyclohexane, 7) Methylcyclohexane, 8) 1,4-Dioxane, 9) Methylisobutylketone, 10) Toluene, 11) Chlorobenzene, 12) Ethylbenzene, 13m) m-xylene, 13p) p-xylene, 13o) o-xylene, 14) acetonitrile, and 15) cumene.
Our solutions for residual solvent testing GC workflows
GC columns
The selection of an appropriate GC column is essential for achieving the selectivity required by USP <467>. Procedure A utilizes a G43 stationary phase for primary screening, while Procedure B employs a G16 stationary phase for confirmation to resolve co-eluting solvent pairs. Table 5 lists representative GC columns suitable for these compendial procedures.
USP code
Description
Cat. no.
G43
OVI-G43, 30m × 0.53mm × 3.0µm
25396
G43
SPB®-624, 30m × 0.32mm × 1.8µm
23323-U
G43
SPB®-624, 60m × 0.32mm × 1.8µm
24217-U
G16
SUPELCOWAX® 10, 30m × 0.53mm × 0.25µm
24079
G16
SUPELCOWAX® 10, 30m × 0.25mm × 0.25µm
24079
Table 5.Representative G43 and G16 GC capillary columns suitable for residual solvent analysis according to USP <467>
Explore our complete portfolio of USP-designated GC columns.
Guard columns for USP <467>
Guard columns are short lengths of deactivated, uncoated fused silica tubing installed between the GC inlet and the analytical column. They protect the analytical column from nonvolatile sample matrix residues, involatile contaminants, and accumulated column bleed that could otherwise deposit on and degrade the stationary phase. Periodic trimming or replacement of the guard column can extend analytical column lifetime and help maintain chromatographic performance. Guard columns are typically 2–10 m in length.
In headspace GC applications, the headspace matrix is primarily in the gas phase, which minimizes the risk of introducing involatile contaminants. But the deactivated surface must still be chemically compatible with both the analytical column stationary phase and the analyte solvent classes being analyzed.
Selection criteria for guard columns
The guard column's internal diameter should match that of the analytical column. An I.D. mismatch, particularly a larger-to-smaller step-down, can introduce dead volume at the connection, disrupt carrier gas flow, and cause peak broadening and tailing that may compromise system suitability. Hence a guard column with the same I.D. as the analytical column must be selected whenever possible.
Product no.
Product
Dimensions
Deactivation
For column I.D.
25708
Intermediate polar fused silica tubing 
1 m × 0.32 mm 
Phenyl/methyl 
0.32 mm columns 
25339
Intermediate polar fused silica tubing 
5 m × 0.53 mm 
Phenyl/methyl 
0.53 mm columns 
Table 6.Recommended guard columns for residual solvent analysis
Note: Procedure B (G16/WAX): A polar, PEG-type deactivated tubing is ideal for use as a guard column with a WAX/G16 analytical column. For static headspace GC, intermediate-polarity deactivation is also acceptable because the matrix is in the gas phase and analytes are present at trace concentrations.
Column connection hardware: Siltite µ-Union kits and replacement ferrules
Reliable, low-dead-volume connections between the guard and analytical columns are essential for maintaining chromatographic performance, particularly system suitability parameters such as resolution and peak symmetry. The Siltite µ-Union system provides a chemically deactivated stainless steel FingerTite connection designed to minimize dead volume and prevent leaks during GC thermal cycling, without requiring re-tightening after installation. 
Product no. 
Product 
Connects 
GU032-032-2 
Siltite µ-Union Kit, 0.32 mm × 0.32 mm
2 unions, 5 double-taper ferrules, and installation tools
GU053-053-2
Siltite µ-Union Kit, 0.53 mm × 0.53 mm 
2 unions, 5 double-taper ferrules, and installation tools
Table 7.Siltite µ-Union kits for guard column to analytical column connections
Replacement packs provide ferrules for maintaining or replacing connections in the Siltite µ-Union system. Ferrules are supplied separately without unions or installation tools, making these packs suitable for routine maintenance and replacement of worn or damaged ferrules. 
Product no. 
Product 
For Column I.D. 
Pack 
GUF032-032-10 
Siltite replacement ferrules 
0.32 mm × 0.32 mm 
pk of 10 
GUF053-053-10
Siltite replacement ferrules 
0.53 mm × 0.53 mm 
pk of 10 
Table 8.Replacement Siltite ferrules for guard column connections
Solvents for residual solvent analysis
SupraSolv® solvents for headspace GC
Headspace GC relies on high-purity solvents as sample diluents, making solvent quality crucial for minimizing background interference and supporting accurate quantification of residual solvents. The SupraSolv® solvents for headspace GC (HS-GC) are optimized for the quantitative analysis of residual solvents in drug substances, excipients, and drug products in accordance with ICH Q3C guidelines. The solvents are application-tested to ensure low intrinsic volatile impurities and batch-to-batch consistency, helping improve the reliability of residual solvent analysis.
Residual solvent class
Specification
Every residual solvent of class 1 acc. ICH
≤ 1 µg/g
Every residual solvent of class 2 acc. ICH
≤ 10 µg/g
Every residual solvent of class 3 acc. ICH
≤ 50 µg/g
Note: The values shown represent the product specifications for SupraSolv® solvents for headspace GC and should not be interpreted as the acceptable residual solvent limits for pharmaceutical products specified in ICH Q3C(R9).
Table 9.Residual solvent specifications for SupraSolv® solvents for headspace GC
Description
Pack size
Cat. no.
SupraSolv® solvents for headspace gas chromatography (HS-GC)
N,N-Dimethylformamide, for headspace gas chromatography Suprasolv®
1 L; 2.5 L
1.00202
N,N-Dimethylacetamide, for headspace gas chromatography Suprasolv®
1 L
1.00399
Water, for headspace gas chromatography Suprasolv®
1 L; 2.5 L
1.00577
Dimethyl sulfoxide, for headspace gas chromatography Suprasolv®
0.5 L; 1 L; 2.5 L
1.01900
1-Methyl-2-pyrrolidone, for headspace gas chromatography Suprasolv®
0.5 L; 1 L; 2.5 L
1.02497
Benzyl alcohol, for headspace gas chromatography Suprasolv®
1 L; 2.5 L
1.02695
Table 10.Representative SupraSolv® solvents for headspace GC applications
SupraSolv® solvents for GC-MS
SupraSolv® solvents for GC-mass spectrometry (GC-MS) are designed to provide the high purity and MS compatibility required for residual solvent analysis. Their low impurity profile helps optimize ionization efficiency while minimizing matrix effects and spectral interferences across the MS detection range, supporting reliable mass spectral identification and reproducible quantification. Batch release testing includes GC-MS performance criteria to help ensure consistent signal-to-noise ratios and retention-time stability across analyses.
Water, dimethyl sulfoxide (DMSO), and dimethylformamide (DMF) are commonly used diluents for residual solvent analysis, depending on sample solubility. According to USP <467> Section 8.3, DMSO is the recommended diluent for water-insoluble formulations, while water is used for water-soluble formulations. Other solvents commonly used in residual solvent analysis include dichloromethane (DCM), n-hexane, and methanol.
Description
Pack size
Cat. no.
Suprasolv® Solvents for gas chromatography-mass spectrometry (GC-MS)
Acetone, for gas chromatography MS Suprasolv®
1 L; 2.5 L
1.00658
Acetonitrile, for gas chromatography MS Suprasolv®
1 L; 2.5 L
1.00665
Cyclohexane, for gas chromatography MS Suprasolv®
1 L; 2.5 L
1.00667
Dichloromethane, for gas chromatography MS Suprasolv®
1 L; 2.5 L
1.00668
Ethyl acetate, for gas chromatography MS Suprasolv®
1 L; 2.5 L
1.00789
n-Hexane, for gas chromatography MS Suprasolv®
1 L; 2.5 L
1.00795
Methanol, for gas chromatography MS Suprasolv®
1 L; 2.5 L
1.00837
Toluene, for gas chromatography MS Suprasolv®
2.5 L
1.00849
Water, for gas chromatography MS Suprasolv®
2.5 L
1.03702
Table 11.Representative SupraSolv® solvents for GC-MS applications
Reference materials
Accurate residual solvent testing requires a well-structured, traceable reference material (RM) program to support method development, calibration, system suitability, and routine quality control. USP <1467> recommends that standard solutions used for verification demonstrate defined analytical performance, including 80–120% recovery and ≤20% relative standard deviation (RSD), helping ensure the accuracy and precision of residual solvent measurements.¹
Refer to Choosing the correct reference material quality grade for additional information.
Our portfolio includes both compendial primary reference standards and pharmaceutical secondary Certified Reference Materials (CRMs) to support residual solvent testing in accordance with pharmacopeial and regulatory requirements.
Primary reference standards
Our portfolio includes USP primary reference standards for residual solvent testing, providing the official compendial materials specified for method verification and applications where pharmacopeial reference standards are required.
Description
Cat. no.
Primary Reference Standards
Residual Solvents Mixture - Class 1
United States Pharmacopeia (USP) Reference Standard
1601102
Residual Solvents Class 2 - Mixture A
United States Pharmacopeia (USP) Reference Standard
1601281
Residual Solvents Class 2 - Mixture B
United States Pharmacopeia (USP) Reference Standard
1601292
Residual Solvents Class 2 - Mixture C
United States Pharmacopeia (USP) Reference Standard
1601306
Table 12.USP primary reference standards for residual solvent analysis
Pharmaceutical secondary standards (CRMs)
Our pharmaceutical secondary standards are Certified Reference Materials (CRMs) that provide documented traceability to currently valid USP, Ph. Eur., and BP primary reference standards, making them suitable for routine analytical applications.
Key features
  • Traceable to current pharmacopeial primary reference standards throughout the product shelf life
  • Certified values with expanded uncertainty documented in the Certificate of Analysis (CoA)
  • Purity assigned by orthogonal characterization using multiple independent analytical techniques
  • Produced under ISO 17034 and tested in accordance with ISO/IEC 17025
  • Monthly CoA batch updates available
  • Designed for use alongside pharmacopeial primary reference standards as part of a comprehensive reference material program 
Description
Cat. no.
Pharmaceutical Secondary Standards /CRMs
Residual Solvents Mixture - Class I
Pharmaceutical Secondary Standard; Certified Reference Material
PHR1063
Residual Solvents Mixture - Class IIA
Pharmaceutical Secondary Standard; Certified Reference Material
PHR1064
Residual Solvents Mixture - Class IIB
Pharmaceutical Secondary Standard; Certified Reference Material
PHR1065
Residual Solvents Mixture - Class IIC
Pharmaceutical Secondary Standard; Certified Reference Material
PHR1066
Table 13.Pharmaceutical secondary standards (CRMs) for residual solvent analysis
High-purity gas solutions for GC
High-purity carrier and detector gases are essential for reliable residual solvent analysis, as they directly influence baseline stability, detector performance, sensitivity, and GC column lifetime. Maintaining a consistent supply of clean, contaminant-free gases helps ensure robust chromatographic performance throughout routine testing.
Parker ChromGas™ gas generators
Our portfolio includes Parker ChromGas™ gas generators, which provide a continuous on-demand supply of high-purity hydrogen, nitrogen, and zero air for GC applications. By eliminating the need for compressed gas cylinders, they help improve laboratory safety, reduce operating costs, and provide a consistent gas supply for routine GC analyses.
Gas purification products
Even when high-purity gases are used, trace contaminants such as hydrocarbons, moisture, and oxygen can adversely affect chromatographic performance. Appropriate gas purification helps protect GC columns and detectors while improving baseline stability and analytical reproducibility.
Carrier gases (He, N₂, or H₂) should be free of hydrocarbons, moisture, and oxygen before entering the GC system.
Gas
Required purification
Recommended products
Carrier gas (He/N₂)
Hydrocarbons + moisture + oxygen
Supelpure™ HC + Mol. Sieve 5Å + Supelpure™-O + OMI®-2
Carrier gas (He)
Full purification
Supelco® Helium Purifier + OMI®-2
FID air (compressed)
Hydrocarbons + moisture
Supelpure™ HC + Mol. Sieve 5Å
FID H₂ fuel
Hydrocarbons
Supelpure™ HC
Table 14.Recommended gas purification products for common GC gas streams
OMI®-2 indicator tube
The OMI®-2 indicator tube is an oxygen and moisture indicating purifier containing a color-change resin that provides a visual indication when the purifier is saturated and requires replacement. This enables timely replacement while helping prevent undetected contamination of the GC gas supply.
Description
Cat. no.
OMI®-2 purifier tube
23906
OMI®-2 holder, 1/8 inch stainless steel fittings
23921
Supelpure HC hydrocarbon trap, 120 cc, ¼ in fittings
22446
Molecular Sieve 5A, water vapor trap, 200 cc, ¼ in fittigns
20618
Supelpure-O, oxygen trap, 120 cc, ¼ in fittings
22450-U
Supelco® Helium Purifer with ¼ in fittings
27601-U
Table 15.Representative gas purification products for GC
Headspace vials
Headspace GC is the preferred technique for residual solvent analysis due to its suitability for volatile compounds and reduced matrix interference. Selecting the appropriate headspace vial is therefore essential to ensure compatibility with the GC system and maintain reliable, reproducible analytical performance.
We offer a comprehensive range of headspace vials engineered for GC headspace analysis and compatible with leading autosampler platforms (Perkin-Elmer, Agilent/HP, CTC Combi PAL, Carlo Erba, Shimadzu, Tekmar, and Varian). Vials are manufactured from Type I borosilicate glass and are available in multiple closure styles, volumes, bottom geometries, and glass tints. Tables below summarise all current Supelco headspace vial SKUs by product family.
Choosing the right vial
  • Flat bottom: Maximizes heating uniformity on heated trays or blocks used with Carlo Erba, Agilent, and Shimadzu instruments.
  • Rounded-bottom: Preferred for robotic-arm autosamplers that lift vials from a tray, including PerkinElmer, Tekmar, Varian, and CTC PAL systems.
  • Beveled top: Reduces the sealing contact area with the crimp seal, helping improve seal integrity under pressure.
  • Long-neck/flat-top: Suitable for instruments that grip the vial at the neck, as commonly found on Carlo Erba and Shimadzu platforms.
  • Amber glass: Protects light-sensitive analytes, while clear glass allows visual inspection of the sample.
  • Screw-top: Features an 18 mm thread and requires magnetic screw caps, sold separately.
Standard flat-bottom vials with a beveled finish that reduce the crimp-seal and are compatible with most autosamplers using 20 mm crimp seals.
Product no. 
Volume 
Glass 
Top Finish 
Bottom 
O.D. × H (mm) 
I.D. (mm) 
Pkg Qty 
Closure 
27197 
6 mL 
Clear 
Beveled 
Flat 
23 × 38 
12.5 
100 
20 mm crimp 
27198 
10 mL 
Clear 
Beveled 
Flat 
23 × 46 
12.5 
100 
20 mm crimp 
27199
20 mL 
Clear 
Beveled 
Flat 
23 × 75.5 
12.5 
100 
20 mm crimp 
27293* 
6 mL 
Clear 
Beveled 
Flat 
22 × 38 
 -
1,000 
20 mm Crimp 
*27293 is the 1,000-count bulk version of the 6 mL beveled-top, flat-bottom vial. The slight O.D. difference (22 mm vs. 23 mm) may affect autosampler compatibility.  Verify compatibility before ordering.
Table 16Specifications for headspace vials with beveled tops, flat bottoms, and 20 mm crimp closures
Rounded-bottom design for robotic-arm autosamplers, with a beveled top that helps preserve seal integrity under pressure. Compatible with CTC Combi PAL systems.
Product no. 
Volume 
Glass 
Top finish 
Bottom 
O.D. × H (mm) 
I.D. (mm) 
Pkg qty 
Closure 
27294
10 mL 
Clear 
Beveled 
Rounded 
22.6 × 46 
-
100 
20 mm Crimp 
Table 17.Specifications for a headspace vial with a beveled top, rounded bottom, and 20 mm crimp closure
Long-neck, flat-top vials designed for instruments that grip the vial at the neck, typically Carlo Erba and Shimadzu headspace autosamplers. Also compatible with Agilent platforms. 
Product no. 
Volume 
Glass 
Top finish 
Bottom 
O.D. × H (mm) 
I.D. (mm) 
Pkg qty 
Closure 
SU860029 
10 mL 
Clear 
Flat 
Flat 
22.5 × 46 
12.5 
100 
20 mm Crimp 
SU860030 
20 mL 
Clear 
Flat 
Flat 
22.5 × 75.5 
12.5 
100 
20 mm Crimp 
Table 18.Specifications for headspace vials with flat tops, flat bottoms, long necks, and 20 mm crimp closures
Flat-top, rounded-bottom design that balances heating efficiency and robotic handling. Compatible with PerkinElmer, Tekmar, and Varian autosamplers.
Product no. 
Volume 
Glass 
Top finish 
Bottom 
O.D. × H (mm) 
I.D. (mm) 
Pkg qty 
Closure 
854180U
10 mL 
Clear 
Flat 
Rounded 
22.5 × 46 
-
100 
20 mm Crimp 
Table 19.Specifications for a headspace vial with flat top, rounded bottom, and 20 mm crimp closure
Screw-cap closure provides simple, torque-controlled sealing without a crimping tool. Requires magnetic screw caps SU860101 (PTFE/silicone, 1.3 mm), SU860102 (PTFE/butyl, 1.6 mm), or SU860103 (PTFE/silicone, 1.5 mm), sold separately. Rounded-bottom design is compatible with robotic-arm autosamplers.  
Product no. 
Volume 
Glass 
Top finish 
Bottom 
O.D. × H (mm) 
Thread 
Pkg qty 
Closure 
SU860099 
10 mL 
Clear 
Screw 
Rounded 
22.5 × 46 
18 mm 
100 
18 mm Screw 
SU860100
10 mL 
Amber 
Screw 
Rounded 
22.5 × 46 
18 mm 
100 
18 mm Screw 
SU860097 
20 mL 
Clear 
Screw 
Rounded 
22.5 × 75.5 
18 mm 
100 
18 mm Screw 
SU860098 
20 mL 
Amber 
Screw 
Rounded 
22.5 × 75.5 
18 mm 
100 
18 mm Screw 
Note: Screw-top vials require a compatible cap with an 18 mm thread and do not require a crimping tool. Amber glass versions help protect light-sensitive analytes.
Table 20.Specifications for headspace vials with screw tops, rounded bottoms, and 18 mm screw threads.
Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
References
1.
2024. International Council for Harmonisation of Technical Requirements for Pharmaceuticals for Human Use (ICH). ICH Harmonised Guideline Q3C(R9): Impurities: Guideline for Residual Solvents. [Internet]. Geneva, Switzerland: Available from: https://database.ich.org/sites/default/files/ICH_Q3C%28R9%29_Guideline_MinorRevision_2024_2024_Approved.pdf
2.
2020. 〈467〉 Residual Solvents. United States Pharmacopeia. https://doi.org/10.31003/uspnf_m99226_08_01
3.
2020. 〈1467〉 Residual Solvents—Verification of Compendial Procedures and Validation of Alternative Procedures. United States Pharmacopeia. https://doi.org/10.31003/uspnf_m11016_05_01