Hibar® RT HPLC Columns and Guard-Column Holder
Introduction
The Hibar® RT is a traditional, ready-to-use HPLC column designed for easy incorporation into analytical workflows. It has threaded ends that allow for easy connection to capillaries via reducers, providing reliability across a wide range of applications.
The Hibar® RT HPLC column hardware is available for Purospher® columns, Purospher® STAR columns, LiChrospher® columns, Superspher® columns, ChiraDex® columns, LiChrosorb® columns, and for customized products on demand.
Guard cartridges can be connected to the analytical column in two ways:
- With a pre-column holder for 4-4 LiChroCART® cartridges for direct coupling to Hibar® RT column
- With a separate guard column holder 4-4 LiChroCART® cartridges for capillary connection. This option is suitable for Hibar® RT columns as well as any other analytical HPLC column
Mounting a LiChroCART® 4-4 guard-column with direct connection to a Hibar® RT HPLC column
- Remove the endfitting of the Hibar® RT columns at the inlet side of the column (in front of flow direction).
- Screw the holder with seat for the pre-column (1.51487.0001) onto the Hibar® RT column and tighten with tool to avoid leaking.
- Fit the LiChroCART® 4-4 guard-column into the holder seat.
- Fit the endfitting of the holder and tighten with tool to avoid leaking.
Guard column holder for Hibar® RT columns | ||
|---|---|---|
Product | Cat. No. | Contents of one package |
Precolumn holder for 4-4 LiChroCART® cartridges for capillary connection to Hibar® RT column | 1 piece | |
Precolumn holder for 4-4 LiChroCART® cartridges for direct coupling to Hibar® RT column | 1 piece | |