Albumin for Cell Culture – Selection Guide
What are Albumins?
Crucial proteins that bind, stabilize, and sequester other proteins and small molecules in cells, albumins are proteins that play a large role in cell culture applications such as:
- Preventing non-specific binding in immunoassays (including ELISA) and Western blotting
- As a stabilizer in enzymatic assays and in therapeutic development
- As a cell culture media supplement, providing cells with growth factors and other nutrients
The most commonly used albumins include bovine serum albumin (BSA), human serum albumin (HSA), and recombinant albumins, but can be sourced from chicken, mouse, porcine, and animal component free systems. Not all albumins have the same efficacy in culture media, so it is important to consider the purity and if the product is tested for cell culture applications. The variety of albumins makes it difficult to determine which protein is right for your application.
Here we present a selection guide for users to determine the ideal albumin for their cell culture application, as albumin selection can drastically affect experimental outcomes.
Enzyme Immunoassays (EIA, ELISA)
Product Number | Name | Method | M Clarity | Form |
|---|---|---|---|---|
Bovine Serum Albumin | lyophilized powder, ≥96% | 200 | Powder | |
Bovine Serum Albumin | protease free, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | lyophilized powder, 1× crystallized, ≥97% | 200 | Powder | |
Bovine Serum Albumin | cold ethanol fraction, pH 5.2, ≥96% | 300 | Powder | |
Bovine Serum Albumin | Essentially fatty acid free, NZ origin | 200 | Powder | |
Bovine Serum Albumin | Protease free, fatty acid free, essentially globulin free, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | Suitable for use as a gel filtration marker; pH <5.0 | 200 | Powder | |
Bovine Serum Albumin | Low endotoxin, pH 7, ≥98% | 200 | Powder |
Cell and Tissue Culture
Product Number | Name | Method | M Clarity | Form |
|---|---|---|---|---|
Bovine Serum Albumin | lyophilized powder, low endotoxin, suitable for cell culture, ≥98% | 200 | Powder | |
Bovine Serum Albumin | New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98% | 200 | Powder | |
Bovine Serum Albumin | lyophilized powder, essentially IgG-free, low endotoxin, suitable for cell culture | 300 | Powder | |
Bovine Serum Albumin | γ-irradiated, Globulin Free, suitable for cell culture | 200 | Powder | |
Bovine Serum Albumin | cell culture | embryo: suitable, ≥98% (agarose gel electrophoresis) | 200 | Powder | |
Bovine Serum Albumin | Essentially globulin free, suitable for cell culture | 200 | Powder | |
Bovine Serum Albumin | Essentially fatty acid free, NZ origin | 200 | Powder | |
Bovine Serum Albumin | Protease free, low endotoxin, suitable for cell culture, pH 7, ≥98% | 200 | Powder | |
Bovine Serum Albumin solution | 35% in DPBS, sterile-filtered, suitable for cell culture | 400 | Solution | |
Bovine Serum Albumin solution | 7.5% in DPBS, sterile-filtered, suitable for cell culture | 200 | Solution | |
Bovine Serum Albumin | fatty acid free, low endotoxin, lyophilized powder, suitable for cell culture, ≥96% | 300 | Powder | |
Bovine Serum Albumin | cell culture | mammalian: suitable, ≥96% | 200 | Powder | |
Bovine Serum Albumin solution | 30% in DPBS, sterile-filtered | 300 | Solution |
3D Cell Culture, Stem Cell & Drug Discovery
Drug Discovery | ||||
|---|---|---|---|---|
Product Number | Name | Method | M Clarity | Form |
Bovine Serum Albumin | Protease free, fatty acid free, essentially globulin free, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | Lyophilized powder, ≥96% | 200 | Powder | |
Bovine Serum Albumin | cold ethanol fraction, pH 5.2, ≥96% | 300 | Powder | |
Bovine Serum Albumin | protease free, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | pH 7, ≥98% | 300 | Powder | |
Molecular Biology
Product Number | Name | Method | M Clarity | Form |
|---|---|---|---|---|
Albumin, Acetylated from bovine serum | DNase free, protease free, NICKase free, RNase free | 200 | Powder | |
Bovine Serum Albumin | Protease free, suitable for hybridization, pH 7, ≥98% | 200 | Powder | |
Bovine Serum Albumin | Non-acetylated, Alkaline phosphatase and peroxidase free, DNase, RNase, and protease free, Peroxidase free | 300 | Powder | |
Albumin | Acetylated, DNase, RNase, NICKase and protease free | 200 | Solution |
Ligand Binding
RIA systems
Product Number | Name | Method | M Clarity | Form |
|---|---|---|---|---|
Bovine Serum Albumin | lyophilized powder, 1× crystallized, ≥97% | 200 | Powder | |
Bovine Serum Albumin | Protease free, fatty acid free, essentially globulin free, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | pH 5.2, ≥96% | 200 | Powder | |
Bovine Serum Albumin | heat shock fraction, pH 7, ≥98% | 300 | Powder |
Protein Base
Product Number | Name | Method | M Clarity | Form |
|---|---|---|---|---|
Bovine Serum Albumin | Lyophilized powder, ≥96% | 200 | Powder | |
Bovine Serum Albumin | pH 5.2, ≥96% | 200 | Powder | |
Bovine Serum Albumin | lyophilized powder, 1× crystallized, ≥97% | 200 | Powder | |
Bovine Serum Albumin | cold ethanol fraction, pH 5.2, ≥96% | 300 | Powder | |
Bovine Serum Albumin | Protease free, fatty acid free, essentially globulin free, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | pH 7, ≥98% | 300 | Powder |
Microbiological Cell Culture
Product Number | Name | Method | M Clarity | Form |
|---|---|---|---|---|
Bovine Serum Albumin | lyophilized powder, ≥96% | 200 | Powder | |
Bovine Serum Albumin | protease free, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | pH 5.2, ≥96% | 200 | Powder | |
Bovine Serum Albumin | cold ethanol fraction, pH 5.2, ≥96% | 300 | Powder | |
Bovine Serum Albumin | Essentially fatty acid free, ≥96% (agarose gel electrophoresis) | 300 | Powder | |
Bovine Serum Albumin | Essentially globulin free, ≥99% (agarose gel electrophoresis) | 300 | Powder | |
Bovine Serum Albumin | pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | pH 7, ≥96% | 200 | Powder | |
Bovine Serum Albumin | Low endotoxin, pH 7, ≥98% | 200 | Powder | |
Albumin | Acetylated, DNase, RNase, NICKase, protease free | 200 | Powder |
Enzyme Systems
Product Number | Name | Method | M Clarity | Form |
|---|---|---|---|---|
Bovine Serum Albumin | lyophilized powder, ≥96% | 200 | Powder | |
Bovine Serum Albumin | protease free, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | pH 5.2, ≥96% | 200 | Powder | |
Bovine Serum Albumin | lyophilized powder, 1× crystallized, ≥97% | 200 | Powder | |
Bovine Serum Albumin | cold ethanol fraction, pH 5.2, ≥96% | 300 | Powder | |
Bovine Serum Albumin | Protease free, fatty acid free, essentially globulin free, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | Essentially globulin free, ≥99% (agarose gel electrophoresis) | 300 | Powder | |
Bovine Serum Albumin | heat shock fraction, suitable for RIA, pH 5.2, ≥96% | 200 | Powder | |
Bovine Serum Albumin | heat shock fraction, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | Low endotoxin, pH 7, ≥98% | 200 | Powder |
Serology
Product Number | Name | Method | M Clarity | Form |
|---|---|---|---|---|
Bovine Serum Albumin | pH 5.2, ≥96% | 200 | Powder | |
Bovine Serum Albumin | lyophilized powder, 1× crystallized, ≥97% | 200 | Powder | |
Bovine Serum Albumin | cold ethanol fraction, pH 5.2, ≥96% | 300 | Powder | |
Bovine Serum Albumin | Protease free, fatty acid free, essentially globulin free, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | Low endotoxin, pH 7, ≥98% | 200 | Powder |
Immunoassay
Product Number | Name | Method | M Clarity | Form |
|---|---|---|---|---|
Bovine Serum Albumin | ≥96% , New Zealand origin | 200 | Powder | |
Bovine Serum Albumin | New Zealand origin, flow cytometry suitable, ≥98% | 200 | Powder | |
Bovine Serum Albumin solution | 35% in DPBS, sterile-filtered, suitable for cell culture | 400 | Solution | |
Bovine Serum Albumin solution | low endotoxin, 7.5% in DPBS, sterile-filtered | 200 | Solution | |
Bovine Serum Albumin solution | endotoxin tested, 30% in DPBS, sterile-filtered | 300 | Solution |
Blood Typing
Product Number | Name | Method | M Clarity | Form |
|---|---|---|---|---|
Bovine Serum Albumin | lyophilized powder, ≥96% | 200 | Powder | |
Bovine Serum Albumin | protease free, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | pH 5.2, ≥96% | 200 | Powder | |
Bovine Serum Albumin | cold ethanol fraction, pH 5.2, ≥96% | 300 | Powder | |
Bovine Serum Albumin | Protease free, fatty acid free, essentially globulin free, pH 7, ≥98% | 300 | Powder | |
Bovine Serum Albumin | heat shock fraction, suitable for RIA, pH 5.2, ≥96% | 200 | Powder |