MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
HomeMedia PreparationValidated Animal Component-Free Growth Factors for Cell Culture
Validated Animal Component-Free Growth Factors for Cell Culture
Animal component-free (ACF) growth factors help promote cell growth without the safety risks that are sometimes associated with animal products, making them ideal for use in applications such as stem and cancer cell culture, immunological experiments, and antibody and vaccine development. Our collection of cell culture-validated, recombinant ACF growth factors have multiple benefits for cell culture experiments including:
  • High bioactivity with low endotoxins
  • Comparable (or higher) bioactivity versus animal-component containing recombinant growth factors
  • Little to no lot-to-lot variations or import restrictions
  • Reduced performance variability and consistent results
Enable a smooth transition towards novel clinical, pharmaceutical, biotech, and manufacturing applications with ACF growth factors. Click here to view our complete offering of ACF growth factors and cell culture-related reagents.
Section Overview:
EGF
TNF-α
IL-6
IL-2
Epidermal Growth Factor (EGF) Protein, Human
  • Catalog Number: E5036
  • Source: Recombinant expressed in E. coli; Animal Component Free
  • Purity: >97% (SDS-PAGE)
  • Molecular Weight: ~6 kDa
  • Bioactivity: 0.08–0.8 ng/mL EC50
Fluorescence graph of mouse fibroblasts when EGF is added to the media. The Y axis shows FLU 590 nm from 20000 to 80000 in intervals of 10000; the X axis is EGF concentration from 0.001ng/ml to 100 ng/ml. The trend line is a S shaped curve.
The biological activity was measured by the ability of E5036 to stimulate the mouse fibroblast cell line BALB/3T3.
EGF Human
  • Catalog Number: E1235
  • Source: Recombinant expressed in HEK 293; Xenobiotic Free (animal component-free)
  • Purity: ≥95% (SDS-PAGE)
  • Molecular Weight: 7.3 kDa (His-tagged)
  • Bioactivity: 0.5-5 ng/mL EC50
Fluorescence graph of A431 when EGF is added to the media. The Y axis shows is from 0.4 to 1.6 in intervals of 0.2; the X axis is log EGF concentration from 0.001ng/ml to 1000 ng/ml. The trend line is a backwards S shaped curve.
E1235 biological activity was tested in culture using human epithelial A431 cells. The EC50 is defined as the effective concentration that elicits a 50% increase in cell growth.
Tumor Necrosis Factor-α (TNF-α) Human
  • Catalog Number: SRP3177
  • Source: Recombinant expressed in E. coli; Animal Component-Free
  • Purity: ≥95% (SDS-PAGE)
  • Molecular Weight: 17.4 kDa
  • Bioactivity: ≤ 0.10ng/ml EC50
Percent cytotoxicity graph of mouse fibroblasts when TNF alpha is added in the presence of actinomycin D. The Y axis shows percent cytotoxicity from 0 to 120 in intervals of 20; the X axis is TNF alpha  concentration from 0.001ng/ml to 10 ng/ml. The trend line is a S shaped curve.
The biological activity of SRP3177 was tested in culture by cytotoxicity assay using L-929 mouse fibroblast cells in the presence of metabolic inhibitor actinomycin D. The EC50 is defined as the effective concentration that causes 50% cytotoxicity.
Tumor Necrosis Factor-α Human
  • Catalog Number: H8916
  • Source: Recombinant expressed in HEK 293; Xenobiotic Free (animal component-free)
  • Purity: ≥95% (SDS-PAGE)
  • Molecular Weight: 17.4 kDa
  • Bioactivity: ≤1.0 ng/mL EC50
Percent cytotoxicity graph of mouse fibroblasts when TNF alpha is added in the presence of actinomycin D. The Y axis shows optical density at 450 nm from 0 to 3.5 in intervals of 0.5; the X axis is TNF alpha  concentration from 0.001ng/ml to 10 ng/ml. The trend line is a backwards S shaped curve.
The biological activity of H8916 was tested in culture by measuring its ability to stimulate cytotoxicity of the TNF-α sensitive cell line L-929 in the presence of actinomycin D. The EC50 is defined as the effective concentration of TNF-α that elicits a 50% decrease in cell growth in a cell-based bioassay.
Interleukin-6 (IL-6) Human
  • Catalog Number: SRP3096
  • Source: Recombinant expressed in E. coli; Animal Component-Free
  • Purity: ≥95% (SDS-PAGE)
  • Molecular Weight: 20.9 kDa
  • Bioactivity: ≤2.0 ng/mL EC50
Proliferation graph of T1165 cells when IL-6 is added. The Y axis shows optical density at 580 nm from 0 to 0.3 in intervals of 0.05; the X axis is IL-6  concentration from 0.1ng/ml to 100 ng/ml. The trend line is a S shaped curve.
The biological activity of SRP3096 was tested in culture by measuring its ability to stimulate the proliferation of IL-6- dependent T1165 cells. The EC50 is defined as the effective concentration that elicits a 50% increase in cell growth.
Interleukin-6 (IL-6) Human
  • Catalog Number: SCU0001
  • Source: Recombinant expressed in HEK 293; Carrier Free
  • Purity: ≥98% (SDS-PAGE)
  • Molecular Weight: 20.8 kDa
  • Bioactivity: 0.2-2 ng/mL EC50
Proliferation graph of T1165 cells when IL-6 is added. The Y axis shows optical density at 420 nm from 0 to 0.6 in intervals of 0.1; the X axis is IL-6 concentration from 0.1ng/ml to 10 ng/ml. The trend line is a S shaped curve.
The biological activity of SCU0001 was measured in a cell proliferation assay using T1165 cells. The EC50 is defined as the effective concentration that elicits a 50% increase in cell growth in a cell-based bioassay.
Interleukin-2, Human
  • Catalog Number: SRP3085
  • Source: Recombinant expressed in E. coli; Animal Component-Free
  • Purity: ≥95% (HPLC)
  • Molecular Weight: 15.5 kDa
  • Bioactivity: ≤0.5 ng/mL ED50
Fluorescence graph of CTLL-2 cells when IL-2 is added. The Y axis shows FLU from 2e+4 to 1.8e+5; the X axis is IL-2 concentration from 0.001ng/ml to 10 ng/ml. The trend line is a S shaped curve.
The biological activity of SRP3085 was tested in culture by measuring its ability to stimulate the proliferation of IL-2-dependent CTLL-2 cells. The EC50 is defined as the effective concentration that elicits a 50% increase in cell growth.
Transforming Growth Factor-β1 Human
  • Catalog Number: H8541
  • Source: Recombinant expressed in HEK 293; Xenobiotic Free (animal component-free)
  • Purity: ≥95% (SDS-PAGE)
  • Molecular Weight: 25 kDa
  • Bioactivity: ≤0.5 ng/mL EC50
Activity graph of IL-4 inhibited HT-2 cells when TGF-β1 is added. The Y axis is from 0 to 1 at 0.2 intervals; the X axis is TGF-β1 concentration from 0.001ng/ml to 100 ng/ml. The trend line is a backwards S shaped curve.
Biological activity of TGF-β1 was tested in culture measuring its ability to inhibit IL-4 induced proliferation of mouse HT-2 cells. The EC50 is defined as the effective concentration that elicits a 50% inhibition of cell proliferation.
Insulin Like Growth Factors (IGF-I) human
  • Catalog Number: SRP3069
  • Source: Recombinant expressed in E. coli; Animal Component-Free
  • Purity: ≥95% (HPLC)
  • Molecular Weight: ~7.6 kDa
  • Bioactivity: 0.1-10 ng/mL ED50/EC50
Fluorescence graph of Balb-3T3 cells when IGF-1 is added to serum-free medium. The Y axis is FLU from 0 to 4e+5 ; the X axis is IGF-1 concentration from 0.01ng/ml to 1000 ng/ml. The trend line is a S shaped curve.
The biological activity of SRP3069 was measured by its ability to stimulate the proliferation of Balb 3T3 cells in serum-free medium. Cell viability was measured using Resazurin-based assay.
Erythropoietin Human
  • Catalog Number: H5166
  • Source: Recombinant expressed in HEK 293; Animal Component-Free
  • Purity: ≥95% (SDS-PAGE)
  • Molecular Weight: dimer 36 kDa
  • Bioactivity: ≤5.0 ng/mL ED50
Proliferation graph of TF-1 cells when erythropoietin is added . The Y axis is optical density at 450 nm ; the X axis is erythropoietin concentration from 0.002ng/ml to 20 ng/ml. The trend line is a S shaped curve.
EC50 is defined as the effective concentration of H5166 that elicits a 50% cell proliferation in TF1 cell-based bioassay.
Granulocyte-Macrophage Colony-Stimulating Factor Human
  • Catalog Number: SRP3050
  • Source: Recombinant expressed in E. coli; Animal Component-Free
  • Purity: ≥98% (SDS-PAGE)
  • Molecular Weight: 14.6 kDa
  • Bioactivity: ≤ 0,100 ng/ml ED50
Proliferation in a TF1 cell-based bioassays when GMCSF is added. The Y axis is optical density at 570 nm; the X axis is GMCSF concentration from 0.004ng/ml to 4 ng/ml. The trend line is a S shaped curve.
The EC50 is defined as the effective concentration of SRP3050 that elicits a 50% cell growth in TF1 cell-based bioassay.
Granulocyte-Macrophage Colony-Stimulating Factor Human
  • Catalog Number: G5035
  • Source: Recombinant expressed in E. coli; Carrier Free
  • Purity: ≥98% (SDS-PAGE)
  • Molecular Weight: 14.6 kDa
  • Bioactivity: ≤0.1 ng/mL EC50
Alkaline phosphatase activity in a TF1 cell-based bioassays when GMCSF is added. The Y axis is optical density at 570 nm; the X axis is GMCSF concentration from 0.001 ng/ml to 1 ng/ml. The trend line is a S shaped curve.
The EC50 is defined as the effective concentration of G5035 that elicits a 50% increase of alkaline phosphatase in TF1 cell-based bioassay.