MilliporeSigma
HomeInsulin and Dexamethasone in Cell Culture
Insulin and Dexamethasone in Cell Culture
Human Insulin 
Insulin is a hormone peptide that plays an essential role for many biological processes in vertebrates. It is involved in glucose uptake and homeostasis, helping to maintain the overall metabolic balance. Insulin regulates metabolism and gene expression in the cell, coordinating cellular responses to nutrients and energy availability. It is also known to have anti-apoptotic effects and to stimulate protein synthesis, helping to maintain proper tissue health and biological functions.
Human Insulin Structure
Human insulin is composed of two polypeptide chains, labeled A and B. The structure includes two inter-chain disulfide bonds and one intra-chain di-sulfide bond on the A chain. With a total molecular weight of 5807 Da, the A chain of human insulin contains 21 amino acids and the B chain contains 30 amino acids. 
Molecular structure of insulin protein. The structure includes two polypeptide chains, labeled as the “A” and “B” chains. A visual representation of the two disulfide bonds that link the chains together is shown. The B chain shows the 30 amino acids it’s comprised of, while the A chain shows 21 amino acids.
Figure 1.Insulin structure
Human Insulin in Cell Culture 
Insulin is a component of the culture media supplement fetal bovine serum (FBS) in cell culture applications. Insulin plays several important roles in culture, as it supports cellular metabolism, growth, and differentiation. For many commonly used cell lines, such as HeLa cells, the insulin in FBS is sufficient to support healthy cell growth. There are, however, several cell lines that benefit from additional insulin supplements in the media to support optimal growth and differentiation. These cell lines include those commonly used in research related to diabetes, obesity, and metabolic disorders, as human insulin plays a critical role in cell metabolism and function.
In specific cases where cells require additional insulin, including culturing pluripotent and neural stem cells, hepatocytes and hepatoma cells, CHO cells, and HEK293 cell lines, supplementing the culture media with the required levels of insulin is common. This adjustment helps to ensure that these cells can grow and maintain their desired characteristics effectively. Insulin can be added to media for serum-free cell cultures.
Insulin is also a common supplement in biopharmaceutical applications, as it helps improve the performance of cell lines to achieve higher yields of the desired protein. 
Product No.DescriptionCommon ApplicationsCommon Assays
I0516Insulin solution from bovine pancreasCell culture supplement
Glucose uptake studies
Metabolic research
Glucose uptake assays
Insulin receptor phosphorylation
Adipocyte differentiation assays
I1882Insulin from bovine pancreas, γ-irradiatedSterile applications in cell culture
Diabetes research
Metabolic assays
Cell proliferation assays in serum-free media
Insulin signaling pathway analysis
Glucose transporter translocation assays
I3536Insulin, human recombinant, zinc solutionSerum-free media
Diabetes and insulin receptor studies
Metabolic pathway research
Insulin receptor binding assays
ELISA for insulin quantification β-cell function assays
I4011Insulin from bovine pancreas, crystallineCell proliferation studies
Insulin signaling
Glucose metabolism
Metabolic flux analysis
Lipogenesis assays in hepatocytes
Insulin-stimulated gene expression profiling
I6634Insulin from bovine pancreas, lyophilized powderCell culture Insulin receptor binding assays
Metabolic regulation
Serum-free cell culture optimization
Insulin dose-response assays
Glucose tolerance assays in vitro
I9278Insulin, human recombinant, lyophilized powderSerum-free media
Therapeutic research
Insulin signaling and receptor binding studies
Insulin bioactivity assays
Receptor internalization assays
Signal transduction pathway mapping
Human Dexamethasone
Dexamethasone is a synthetic glucocorticoid that is commonly added to cell culture media for its effects on cell differentiation, proliferation, and survival. The hormone is often used in cell culture to study apoptosis, cell signaling pathways, and gene expression. Several cell types require dexamethasone supplementation to the media, including adipocytes, chondrocytes, osteoblasts, lymphocytes, neural progenitor cells, macrophages, endothelial cells, and hepatocytes.
Human Dexamethasone Structure 
Human dexamethasone is not a water-soluble molecule. However, it is soluble in ethanol, methanol, acetone, dioxane, DMSO. It is also slightly soluble in chloroform.
Molecular structure of dexamethasone. The structure includes molecular representations of oxygen and fluoride, with hexagons representing the carbon molecules.
Figure 2.Molecular structure of dexamethasone.
Human Dexamethasone in Cell Culture
Dexamethasone supplementation in cell culture media varies depending on the type of cell line that is being cultured and the specific research goals of the experiment. While dexamethasone has specific benefits in certain contexts, it is not universally applicable to all cell lines and should not be used in all applications. The effects of dexamethasone can vary significantly depending on the cell type, concentration, and experimental conditions; researchers should carefully consider using it based on the specific characteristics or requirements of the cell lines they are working with.
Product No.DescriptionCommon ApplicationsCommon Assays
D2915Dexamethasone-Water Soluble*Anti-inflammatory studies
Glucocorticoid receptor research
Cell culture supplement
Glucocorticoid receptor binding assay
Anti-inflammatory cytokine profiling
Apoptosis assays
D4902Dexamethasone*Hormone signaling studies
Anti-inflammatory research
Apoptosis induction
Reporter gene assays
Cell viability assays
Osteogenic differentiation assays
D8893Dexamethasone*Gene expression modulation and cancer researchGene expression analysis
Western blot for GR signaling pathway proteins
In vitro cancer cell proliferation inhibition assays
*Dexamethasone solutions in DMSO are stable for at least 30 days when stored at 4 °C and protected from light.
Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500