II. Preparation for Culturing
- Ensure the Class II Biological Safety Cabinet, with HEPA filtered laminar airflow, is in proper working condition.
- Sterilize the Biological Safety Cabinet with 70% alcohol.
- Turn the Biological Safety Cabinet blower on for 10 minutes before beginning cell culture work.
- Make sure all serological pipettes, pipette tips and reagent solutions are sterile.
- Follow the standard sterilization technique and safety rules:
a. Do not pipette by mouth.
b. Always wear gloves and safety glasses when working with animal cells even though all the animals have
been inspected by the USDA.
c. Handle all cell culture work in a sterile hood.
III. Culturing PAOEC
A. Preparing Culture Flask (optional)
- Swirl the Attachment Factor Solution (123-100) a few times to form a homogenous solution. If the solution is gelled, warm up in 56°C water bath for 10 minutes and swirl the bottle to mix the solution.
- Decontaminate the bottle with 70% alcohol in a sterile hood.
- Add 7.5 ml of Attachment Factor Solution (123-100)* to a T-75 flask (SIAL0641) and rock the flask gently to distribute the solution evenly to cover the whole culture surface.
- Coat the culture ware for 30 minutes at 37°C or 2 hours to overnight at room temperature.
- Remove the Attachment Factor Solution (123-100) by aspiration in a sterile hood.
- The coated flask can be used immediately or stored at 4°C for up to 2 weeks.
- Prepare the coated flask for seeding by pipetting 15 ml of Porcine Endothelial Growth Medium (P211-500) into this coated T-75 flask (SIAL0641) and wait for seeding.
*The coating concentration is 1 ml per 10cm2 surface area of culture ware.
- 2.5 ml for coating a T-25 flask or a 60mm culture dish
- 7.5 ml for coating a T-75 flask or a 100mm culture dish
B. Preparing Cell Culture Flasks for Culturing PAOEC
- Take the Porcine Endothelial Growth Medium (P211-500) from the refrigerator. Decontaminate the bottle with 70% alcohol in a sterile hood.
- Pipette 15 ml of Porcine Endothelial Growth Medium (P211-500)* to a T-75 flask (SIAL0641).
* Keep the medium to surface area ratio at 1ml per 5 cm2.
For example:
- 5ml for a T-25 flask (SIAL0639) or a 60 mm tissue culture dish (SIAL0166).
- 15 ml for a T-75 flask (SIAL0641) or a 100 mm tissue culture dish (SIAL0167).
C. Thawing and Plating PAOEC
- Remove the cryopreserved vial of PAOEC from the liquid nitrogen storage tank using proper protection for your eyes and hands.
- Turn the vial cap a quarter turn to release any liquid nitrogen that may be trapped in the threads, then re-tighten the cap.
- Thaw the cells quickly by placing the lower half of the vial in a 37°C water bath and watch the vial closely during the thawing process.
- Remove the vial from the water bath when only a small amount of ice is left in the vial. Do not let cells thaw completely.
- Decontaminate the vial exterior with 70% alcohol in a sterile Biological Safety Cabinet.
- Remove the vial cap carefully. Do not touch the rim of the cap or the vial with your hands to avoid contamination.
- Resuspend the cells in the vial by gently pipetting the cells 5 times with a 2 ml pipette. Be careful not to pipette too vigorously as to cause foaming.
- Pipette the cell suspension (1ml) from the vial into the T-75 flask (SIAL0641) containing 15 ml of Porcine Endothelial Growth Medium (P211-500).
- Cap the flask and rock gently to evenly distribute the cells.
- Place the T-75 flask (SIAL0641) in a 37oC, 5% CO2 humidified incubator. Loosen the cap to allow gas exchange. For best results, do not disturb the culture for 24 hours after inoculation.
- Change to fresh Porcine Endothelial Growth Medium (P211-500) after 24 hours or overnight to remove all traces of DMSO.
- Change Porcine Endothelial Growth Medium (P211-500) every other day until the cells reach 60% confluency.
- Double the Porcine Endothelial Growth Medium (P211-500) volume when the culture is >60% confluent or for weekend feedings.
- Subculture the cells when the PAOEC culture reaches 80% confluency.
IV. Subculturing PAOEC
A. Preparing Subculture Reagents
- Remove the Trypsin-EDTA solution (T3924) and Trypsin Inhibitor (T6414) from the -20°C freezer and thaw overnight in a refrigerator.
- Make sure all the subculture reagents are thawed. Swirl each bottle gently several times to form homogeneous solutions.
- Store all the subculture reagents at 4°C for future use.
- Aliquot Trypsin/EDTA solution (T3924) and store the unused portion at -20°C if only a portion of the Trypsin/EDTA (T3924) is needed.
B. Preparing Culture Flask
- Add 10 ml of Attachment Factor Solution (123-100)* to a T-175 flask (SIAL1080) and rock the flask gently to distribute the solution evenly to cover the whole culture surface.
- Coat the culture ware for 30 minutes at 37°C or 2 hours to overnight at room temperature.
- Remove the Attachment Factor Solution (123-100) by aspiration in a sterile hood.
- Take the Porcine Endothelial Growth Medium (P211-500) from the refrigerator. Decontaminate the bottle with 70% alcohol in a sterile hood.
- Pipette 35 ml of Porcine Endothelial Growth Medium (P211-500) to a T-175 flask (SIAL1080).
C. Subculturing PAOEC
Trypsinize Cells at Room Temperature. Do Not Warm Any Reagents to 37°C.
- Remove the medium from culture flasks by aspiration.
- Wash the monolayer of cells with HBSS (H6648) and remove the solution by aspiration.
- Pipette 6 ml of Trypsin/EDTA Solution (T3924) into the T-75 flask (SIAL0641). Rock the flask gently to ensure the solution covers all the cells.
- Remove 5.5 ml of the solution immediately.
- Re-cap the flask tightly and monitor the trypsinization progress at room temperature under an inverted microscope. It usually takes about 1 to 2 minutes for the cells to become rounded.
- Release the rounded cells from the culture surface by hitting the side of the flask against your palm until most of the cells are detached.
- Pipette 8 ml of Trypsin Inhibitor Solution (T6414) to the flask to inhibit further tryptic activity.
- Transfer the cell suspension from the flask to a 50 ml sterile conical tube.
- Rinse the flask with an additional 5 ml of Trypsin Inhibitor Solution (T6414) and transfer the solution into the same conical tube.
- Examine the T-75 flask (SIAL0641) under a microscope. If there are >20% cells left in the flask, repeat Steps 2-9.
- Centrifuge the conical tube at 220 x g for 5 minutes to pellet the cells.
- Aspirate the supernatant from the tube without disturbing the cell pellet.
- Flick the tip of the conical tube with your finger to loosen the cell pellet.
- Resuspend the cells in 5 ml of Porcine Endothelial Growth Medium (P211-500) by gently pipetting the cells to break up the clumps.
- Count the cells with a hemocytometer or cell counter. Inoculate at 4,000 cells per cm2 for regular subculturing.