Alkoxysilanes for C–C Coupling Reactions
Chemfiles Volume 3 Article 5
The use of silicon compounds as transmetalation reagents has attracted much attention as a viable alternative to the popular Stille and Suzuki coupling reactions mainly due to the formation of nontoxic byproducts and stability to many reaction conditions.1-4 Silicon-based coupling reactions can be carried out using aryl-, heteroaryl-, or alkenyl- halides and alkoxysilanes in the presence of palladium or rhodium catalysts. Among the various types of silicon compounds available, alkoxysilanes are more effective in coupling reactions.5-11
Silicon-based coupling scheme
Figure 1.Silicon-based coupling scheme
Recently, considerable attention has been paid to the rhodiumcatalyzed addition of aryl(trialkoxy)silanes to carbonyl compounds, such as aldehydes, α,β-unsaturated ketones and esters.12
Rhodium catalyzed scheme
Figure 2.Rhodium catalyzed scheme
Materials
Loading
References
1.
Hatanaka Y, Hiyama T. 1991. Highly Selective Cross-Coupling Reactions of Organosilicon Compounds Mediated by Fluoride Ion and a Palladium Catalyst. Synlett. 1991(12):845-853. http://dx.doi.org/10.1055/s-1991-20899
2.
Chuit C, Corriu RJP, Reye C, Young JC. 1993. Reactivity of penta- and hexacoordinate silicon compounds and their role as reaction intermediates. Chem. Rev.. 93(4):1371-1448. http://dx.doi.org/10.1021/cr00020a003
3.
Horn K. 1995. ibid. 95, 1317.
4.
Hiyama, T.; Shirakawa, E. In Topics of Current Chemistry; Miyaura, N., Ed.; Spinger-Verlag: Heidelberg, 2002; vol. 219, p 61.
5.
Denmark SE, Sweis RF. 2002. Design and Implementation of New, Silicon-Based, Cross-Coupling Reactions:  Importance of Silicon?Oxygen Bonds. Acc. Chem. Res.. 35(10):835-846. http://dx.doi.org/10.1021/ar020001r
6.
Tamao K, Kobayashi K, Ito Y. 1989. Palladium-catalyzed cross-coupling reaction of alkenylalkoxysilanes with aryl and alkenyl halides in the presence of a fluoride ion. Tetrahedron Letters. 30(44):6051-6054. http://dx.doi.org/10.1016/s0040-4039(01)93852-3
7.
Shibata K, et al. 1997. Chem. Commun. 1309.
8.
Mowery ME, DeShong P. 1999. Cross-Coupling Reactions of Hypervalent Siloxane Derivatives:  An Alternative to Stille and Suzuki Couplings. J. Org. Chem.. 64(5):1684-1688. http://dx.doi.org/10.1021/jo982463h
9.
Mowery ME, DeShong P. 1999. Improvements in Cross Coupling Reactions of Hypervalent Siloxane Derivatives. Org. Lett.. 1(13):2137-2140. http://dx.doi.org/10.1021/ol991186d
10.
Lee H, Nolan S. 2000. ibid. 2 2053.
11.
Murata M, Shimazaki R, Watanabe S, Masuda Y. 2001. Palladium-Catalyzed Cross-Coupling Reaction of Aryltriethoxysilanes with Aryl Bromides under Basic Aqueous Conditions. Synthesis. 2001(15):2231-2233. http://dx.doi.org/10.1055/s-2001-18435
12.
Oi S, Honma Y, Inoue Y. 2002. Conjugate Addition of Organosiloxanes to ?,?-Unsaturated Carbonyl Compounds Catalyzed by a Cationic Rhodium Complex. Org. Lett.. 4(4):667-669. http://dx.doi.org/10.1021/ol025518h