Atom Transfer Radical Polymerization (ATRP) Tools & Techniques
Sebastian Grajales, Ph.D.
6000 N. Teutonia Ave. Milwaukee, WI 53209.
Introduction
Controlled radical polymerization (CRP) techniques have been reviewed in a number of articles1,2 and books,3 and include nitroxide-mediated polymerization (NMP),4-7 reversible addition fragmentation chain transfer (RAFT),8-10 and atom transfer radical polymerization (ATRP).11 All of these CRP techniques operate by rapidly establishing an equilibrium between a small fraction of active polymerizing chains and a majority of dormant moieties. ATRP has proven to be versatile and effective in providing a controlled polymerization environment for a wide range of vinyl monomers including styrenes, (meth)acrylates, acrylonitriles, and dienes.3 ATRP differs from conventional radical polymerization because of the ability of a metal complex to dictate the rate of monomer addition to the propagating polymer chain end. While the procedure to perform normal ATRP is simple with essentially three components (initiator, catalyst, and monomer) added to solvent, the equilibrium equation describing the reaction is more complex, and is shown in Figure 1.
ATRP equilibrium equation
Figure 1.The ATRP equilibrium equation, which resides predominantly to the left/dormant side, includes dormant initiator R-X (1), catalyst composed of a transition metal Cu with ligand L (2), oxidized catalyst (3), active radical or active initiator R• (4), and monomer M (5) and terminated polymer RR (6). The reaction rates are labeled with kx.
This equation may appear to have a high degree of complexity, but its meaning and importance can be illustrated by three situations: a polymer sitting dormant on the left side of the equilibrium; a polymer reacting with monomer on the right side of the equilibrium (and growing in molecular weight); and two radical end-groups combining to terminate and kill the reaction (this is neither reversible nor desired). In ATRP, the reaction resides on the left side of the equation most of the time. This minimizes the likelihood for two radicals to exist near each other at the same time and therefore minimizes termination. This control over termination is the primary benefit of choosing ATRP. Another important feature of ATRP is that only one radical is formed upon activation. This is particularly important if surface-initiated polymerization is desired because this minimizes solution polymerization.
The rate of reaction is marked by rate constants (kx) where the rate of activation is kact, the rate of deactivation is kdeact, the rate of monomer addition is kp, and the rate of termination is kt. The control over this equilibrium highly depends on the choice of catalyst.
ATRP: Choosing a Ligand for the Catalyst
The catalyst (complex 2 in Figure 1) is a transition metal ion with coordinated ligands, and complex 3 is the oxidized form of the complex. (The difference between the two is that the oxidized complex 3 has the halogen from the initiator and a charge of one higher than complex 2.) A wide variety of transition metals have been reported to work with ATRP, including Ti,12 Fe,13-17 Co,18 Ni,19-21 Mo,22-24 Ru,25-27 Rh,28 Pd,29 Re,30 Os,31 but the use of Cu dominates the literature. To simplify the discussion, the rest of this article will refer to Cu.
The catalyst complex 2 abstracts the halogen from the initiator (or chain end) to activate the polymerization, so it has an important influence on the reaction rates of the equilibrium. The ligand choice will have a profound effect on kact and kdeact, which will cause a difference in the rate of polymerization. Several ligands are shown plotted along the ratio of (kact/kdeact) in Figure 2.
Nitrogen-based ligands plotted against the ATRP
Figure 2.Nitrogen-based ligands plotted against the ATRP equilibrium constant (KATRP) with initiator EtBibb, in the presence of CuBr in MeCN at 22 °C. Bidentate ligands are blue diamonds, tridentate ligands are red triangles, and tetradentate are green squares. Adapted from Tang, W. et al.32
This shows that, in general, the speed of the polymerization in terms of the ligand structure is bidentate < tridentate < tetradentate. This can also be thought of in terms of how much catalyst is needed to perform the reaction. Less catalyst is needed with a large KATRP value.
Variations of ATRP
Returning to Figure 1, the components that are added to the polymerization solution for normal ATRP are initiator 1, catalyst 2, and monomer 5. Catalyst 2 is oxygen-sensitive, so in an effort to minimize handling of this compound, several variations of ATRP have been developed. Newer “variations” of ATRP were developed primarily to generate the air-sensitive catalyst 2 at the start of the reaction rather than requiring it to be handled and added.
  • Initiators for Continuous Activator Regeneration (ICAR)
  • Activators Generated by Electron Transfer (AGET)
  • Activators ReGenerated by Electron Transfer (ARGET)
  • Reverse ATRP
Detailing the competing reactions for all of these techniques is beyond the scope of this article; however, the following table summarizes the components added to perform the different ATRP reactions.
Table 1ATRP polymerization type and corresponding identity or concentration of popular reactants added.
All of the newer ATRP variations eliminate the need to handle air sensitive Cu1+. ICAR, AGET, and ARGET methods utilize substantially lower concentrations of Cu catalyst over the other types. Table 2 summarizes the benefits and limitations of the various ATRP techniques:
Table 2Benefits and limitations for ATRP techniques.
The relatively high levels of residual Cu by traditional ATRP are problematic for biomedical and food packaging applications where it is required to minimize or eliminate toxins, such as heavy metals. Polymers generated by traditional ATRP contain substantial residual Cu catalyst if not properly purified. ICAR, AGET, and ARGET operate at low levels of Cu during polymerization and therefore need minimal subsequent purification for catalyst removal. These new versions of ATRP enable their utility to prepare well-defined copolymers for these high value applications.
The appropriate ATRP technique can be chosen based on the needs of the application. Figure 2 can be used to select a ligand based on catalyst activity; however, the effect of the ligand on the resulting molecular weight distribution is not illustrated. The plot of the KATRP vs. kdeact is shown in Figure 3 and illustrates catalyst activity as a function of control over the polymerization.32 Wei Tang and co-workers developed this plot which shows the PDI better correlates to the rate of deactivation kdeact rather than the value of KATRP found in Figure 2.32
The equilibrium constant
Figure 3.The equilibrium constant KATRP plotted against kdeact illustrates the correlation between catalyst activity and control. This should only be used as a reference, and was adapted from Tang, W. et al.32 Each ligand is labeled with an acronym, as well as the product number. Bidentate ligands are represented by blue diamonds, red triangles represent tridentate ligands, and green squares represent tetradentate ligands. *At a given concentration of catalyst, the activity axis shows the relative speed of the polymerization reaction.
The perfect ATRP catalyst would be both fast and well controlled, but this figure can help illustrate the relationship. If minimizing copper concentration is the highest priority, then selecting catalysts from the top of Figure 3 would be ideal. If a narrow distribution of chain lengths (low PDI) is the highest priority, then the ideal catalyst would be selected from the right side of the Figure 3.
The various ATRP techniques provide an opportunity to select a polymerization environment that is well suited to meet applicationspecific needs. When used in combination with the ideal catalyst for a given polymerization, well-defined polymers can be conveniently prepared.
Materials
Loading
Materials
Loading
References
1.
Kamigaito M, Ando T, Sawamoto M. 2004. Metal-catalyzed living radical polymerization: discovery and developments. Chem. Record. 4(3):159-175. http://dx.doi.org/10.1002/tcr.20011
2.
Edmondson S, Osborne VL, Huck WTS. 2004. Polymer brushes via surface-initiated polymerizations. Chem. Soc. Rev.. 33(1):14. http://dx.doi.org/10.1039/b210143m
3.
Matyjaszewski K. 2003. Advances in Controlled/Living Radical Polymerization. http://dx.doi.org/10.1021/bk-2003-0854
4.
Hawker CJ, Bosman AW, Harth E. 2001. New Polymer Synthesis by Nitroxide Mediated Living Radical Polymerizations. Chem. Rev.. 101(12):3661-3688. http://dx.doi.org/10.1021/cr990119u
5.
Benoit D, Chaplinski V, Braslau R, Hawker CJ. 1999. Development of a Universal Alkoxyamine for ?Living? Free Radical Polymerizations. J. Am. Chem. Soc.. 121(16):3904-3920. http://dx.doi.org/10.1021/ja984013c
6.
Hawker CJ, Barclay GG, Orellana A, Dao J, Devonport W. 1996. Initiating Systems for Nitroxide-Mediated ?Living? Free Radical Polymerizations:  Synthesis and Evaluation. Macromolecules. 29(16):5245-5254. http://dx.doi.org/10.1021/ma951905d
7.
Fukuda T, Terauchi T, Goto A, Ohno K, Tsujii Y, Miyamoto T, Kobatake S, Yamada B. 1996. Mechanisms and Kinetics of Nitroxide-Controlled Free Radical Polymerization. Macromolecules. 29(20):6393-6398. http://dx.doi.org/10.1021/ma960552v
8.
Chong )YK, Krstina J, Le TPT, Moad G, Postma A, Rizzardo E, Thang SH. 2003. Thiocarbonylthio Compounds [SC(Ph)S?R] in Free Radical Polymerization with Reversible Addition-Fragmentation Chain Transfer (RAFT Polymerization). Role of the Free-Radical Leaving Group (R). Macromolecules. 36(7):2256-2272. http://dx.doi.org/10.1021/ma020882h
9.
Coote ML. 2005. The Kinetics of Addition and Fragmentation in Reversible Addition Fragmentation Chain Transfer Polymerization:  An ab Initio Study. J. Phys. Chem. A. 109(6):1230-1239. http://dx.doi.org/10.1021/jp046131u
10.
Achilleos M, Legge TM, Perrier S, Patrickios CS. 2008. Poly(ethylene glycol)-based amphiphilic model conetworks: Synthesis by RAFT polymerization and characterization. J. Polym. Sci. A Polym. Chem.. 46(22):7556-7565. http://dx.doi.org/10.1002/pola.23061
11.
Wang J, Matyjaszewski K. 1995. Controlled/"Living" Radical Polymerization. Halogen Atom Transfer Radical Polymerization Promoted by a Cu(I)/Cu(II) Redox Process. Macromolecules. 28(23):7901-7910. http://dx.doi.org/10.1021/ma00127a042
12.
Kabachii YA, Kochev SY, Bronstein LM, Blagodatskikh IB, Valetsky PM. 2003. Atom Transfer Radical Polymerization with Ti(III) Halides and Alkoxides. Polymer Bulletin. 50(4):271-278. http://dx.doi.org/10.1007/s00289-003-0157-9
13.
Onishi I, Baek K, Kotani Y, Kamigaito M, Sawamoto M. 2002. Iron-catalyzed living radical polymerization of acrylates: Iodide-based initiating systems and block and random copolymerizations. J. Polym. Sci. A Polym. Chem.. 40(12):2033-2043. http://dx.doi.org/10.1002/pola.10299
14.
Kotani Y, Kamigaito M, Sawamoto M. 1999. FeCp(CO)2I:  A Phosphine-Free Half-Metallocene-Type Iron(II) Catalyst for Living Radical Polymerization of Styrene1. Macromolecules. 32(20):6877-6880. http://dx.doi.org/10.1021/ma9906130
15.
Matyjaszewski K, Wei M, Xia J, McDermott NE. 1997. Controlled/?Living? Radical Polymerization of Styrene and Methyl Methacrylate Catalyzed by Iron Complexes1. Macromolecules. 30(26):8161-8164. http://dx.doi.org/10.1021/ma971010w
16.
O?Reilly RK, Gibson VC, White AJ, Williams DJ. 2004. Five-coordinate iron(II) complexes bearing tridentate nitrogen donor ligands as catalysts for atom transfer radical polymerisation. Polyhedron. 23(17):2921-2928. http://dx.doi.org/10.1016/j.poly.2004.09.001
17.
Ishio M, Katsube M, Ouchi M, Sawamoto M, Inoue Y. 2009. Active, Versatile, and Removable Iron Catalysts with Phosphazenium Salts for Living Radical Polymerization of Methacrylates(1). Macromolecules. 42(1):188-193. http://dx.doi.org/10.1021/ma801762k
18.
Wang B, Zhuang Y, Luo X, Xu S, Zhou X. 2003. Controlled/?Living? Radical Polymerization of MMA Catalyzed by Cobaltocene. Macromolecules. 36(26):9684-9686. http://dx.doi.org/10.1021/ma035334y
19.
Granel C, Dubois P, Jérôme R, Teyssié P. 1996. Controlled Radical Polymerization of Methacrylic Monomers in the Presence of a Bis(ortho-chelated) Arylnickel(II) Complex and Different Activated Alkyl Halides. Macromolecules. 29(27):8576-8582. http://dx.doi.org/10.1021/ma9608380
20.
Uegaki H, Kamigaito M, Sawamoto M. 1999. Polym. Sci., Part A: Polym. Chem.. 373003. https://doi.org/10.1002/(SICI)1099-0518(19990801)37:15<3003::AID-POLA33>3.0.CO;2-K
21.
Uegaki H, Kotani Y, Kamigaito M, Sawamoto M. 1998. NiBr2(Pn-Bu3)2-Mediated Living Radical Polymerization of Methacrylates and Acrylates and Their Block or Random Copolymerizations1. Macromolecules. 31(20):6756-6761. http://dx.doi.org/10.1021/ma9805957
22.
Brandts JA, van de Geijn P, van Faassen EE, Boersma J, van Koten G. 1999. Controlled radical polymerization of styrene in the presence of lithium molybdate(V) complexes and benzylic halides. Journal of Organometallic Chemistry. 584(2):246-253. http://dx.doi.org/10.1016/s0022-328x(99)00149-7
23.
Le Grognec E, Claverie J, Poli R. 2001. Radical Polymerization of Styrene Controlled by Half-Sandwich Mo(III)/Mo(IV) Couples:  All Basic Mechanisms Are Possible. J. Am. Chem. Soc.. 123(39):9513-9524. http://dx.doi.org/10.1021/ja010998d
24.
Maria S, Stoffelbach F, Mata J, Daran J, Richard P, Poli R. 2005. The Radical Trap in Atom Transfer Radical Polymerization Need Not Be Thermodynamically Stable. A Study of the MoX3(PMe3)3Catalysts. J. Am. Chem. Soc.. 127(16):5946-5956. http://dx.doi.org/10.1021/ja043078e
25.
Kamigaito M, Watanabe Y, Ando T, Sawamoto M. 2002. A New Ruthenium Complex with an Electron-Donating Aminoindenyl Ligand for Fast Metal-Mediated Living Radical Polymerizations. J. Am. Chem. Soc.. 124(34):9994-9995. http://dx.doi.org/10.1021/ja0270470
26.
Terashima T, Ouchi M, Ando T, Sawamoto M. 2010. Thermoregulated phase-transfer catalysis via PEG-armed Ru(II)-bearing microgel core star polymers: Efficient and reusable Ru(II) catalysts for aqueous transfer hydrogenation of ketones. J. Polym. Sci. A Polym. Chem.. 48(2):373-379. http://dx.doi.org/10.1002/pola.23794
27.
Yoda H, Nakatani K, Terashima T, Ouchi M, Sawamoto M. 2010. Ethanol-Mediated Living Radical Homo- and Copolymerizations with Cp*-Ruthenium Catalysts: Active, Robust, and Universal for Functionalized Methacrylates. Macromolecules. 43(13):5595-5601. http://dx.doi.org/10.1021/ma100589b
28.
Percec V, Barboiu B, Neumann A, Ronda JC, Zhao M. 1996. Metal-Catalyzed ?Living? Radical Polymerization of Styrene Initiated with Arenesulfonyl Chlorides. From Heterogeneous to Homogeneous Catalysis. Macromolecules. 29(10):3665-3668. http://dx.doi.org/10.1021/ma960061a
29.
Lecomte P, Drapier I, Dubois P, Teyssié P, Jérôme R. 1997. Controlled Radical Polymerization of Methyl Methacrylate in the Presence of Palladium Acetate, Triphenylphosphine, and Carbon Tetrachloride. Macromolecules. 30(24):7631-7633. http://dx.doi.org/10.1021/ma970890b
30.
Kotani Y, Kamigaito M, Sawamoto M. 2000. Living Radical Polymerization of Para-Substituted Styrenes and Synthesis of Styrene-Based Copolymers with Rhenium and Iron Complex Catalysts1. Macromolecules. 33(18):6746-6751. http://dx.doi.org/10.1021/ma000624p
31.
Braunecker WA, Itami Y, Matyjaszewski K. 2005. Osmium-Mediated Radical Polymerization. Macromolecules. 38(23):9402-9404. http://dx.doi.org/10.1021/ma051877r
32.
Tang W, Kwak Y, Braunecker W, Tsarevsky NV, Coote ML, Matyjaszewski K. 2008. Understanding Atom Transfer Radical Polymerization: Effect of Ligand and Initiator Structures on the Equilibrium Constants. J. Am. Chem. Soc.. 130(32):10702-10713. http://dx.doi.org/10.1021/ja802290a