Welding Processes - Air Monitoring Products
welding-processes
Welding is the process by which pieces of metals or thermoplastics are combined or joined by applying heat, pressure, or a combination of both. It is one of the oldest industrial processes in practice today. The earliest examples of welded metals date back to the Bronze and Iron Ages. Welding processes occur in approximately 2% of industrialized working populations; and workers who perform welding tasks are at risk for exposure to various fumes and gases including carbon monoxide, nitrogen dioxide, ozone; also, metal fumes from zinc, cadmium, hexavalent chromium, and mercury among others at dangerous levels. Studies show that welders are at risk for developing lung-related ailments such as bronchitis, pulmonary infections such as pneumonia, and lung cancer.
There are many types of welding processes but the most commonly practiced are Shielded Metal Arc Welding (SMAW) and Gas Metal Arc Welding (GMAW), which comprise roughly 70% of all welding methods. The most commonly welded metals are carbon steel, stainless steel, and aluminum. Exposure to hazardous fumes and gases depend on the welding method and materials involved, the composition of the welding rod (electrode), filler and base metals, paints and coatings being welded, and area ventilation.
Although personal protective equipment and controls are mandated by OSHA and NIOSH for welding processes, welders remain at risk for occupationally-induced lung cancer and other health-related conditions.
Did you know? OSHA has confirmed the effective date of June 15, 2010, for the direct final rule requiring employers to notify their workers of all hexavalent chromium exposures. The rule revises a provision in OSHA's Hexavalent Chromium standard that required workers be notified only when they experienced exposures exceeding the permissible exposure limit.
Types of Sampling
Indoor Air
Personal sampling
Ambient Air
Sampling Media
Sorbent Tubes
Passive Sampling Devices
Thermal Desorption Tubes
Filters
Contaminants of Interest:
Welding Gases
  • Ozone
  • Nitrogen Oxides
  • Carbon Monoxide
    & Carbon Dioxide
Metals & Metal Fumes
Metals
  • Stainless Steel, Carbon Steel, and Zinc
  • Mercury
  • Nickel
  • Lead
  • Chromium-Hexavalent
Metal Fumes
  • Oxides- Iron, Zinc, and Aluminum
  • Cadmium
  • Manganese
  • Magnesium
Other Common Contaminants in Welding
  • Paint & Coating Residues
  • Isocyanates
  • Chlorinated Hydrocarbon Solvents
  • Phosgene
  • Fluorides
  • Asbestos
  • Arsenic
Air Sampling Media by Regulatory Method