MilliporeSigma
HomeChemical Analysis for Food and Beverage TestingDetermination of Glyphosate, Glufosinate, and AMPA in tea
Green tea analysis for determination of glyphosate, glufosinate, and AMPA by LC-MS/MS with QuEChERS cleanup
Jack Wang
R&D APAC lab, Shanghai, China
Abstract
A QuEChERS-liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was established for the determination of glyphosate, glufosinate, and AMPA in green/fresh tea. The samples were extracted by acetonitrile/water solution, and the extracts cleaned up by QuEChERS using a dedicated adsorbent mix for green samples. Analysis and quantification were performed by LC-MS/MS with multi-reaction monitoring mode (MRM) and matrix external standard method. Glyphosate, glufosinate, and AMPA compounds showed a good linear relationship in the range of 1-150 µg/L with correlation coefficients greater than 0.9995, recoveries in the range of 80.0% to 107%, and the relative standard deviations (RSD) were 2.9-5.0%, meeting the requirements of GB 23200.13-2016. The detection limits of the method met the more stringent requirements of SN/T 1923-2007 and were as follows for LOQs (µg/kg): glyphosate: 15.0; glufosinate: 27.0; AMPA:33.0. An inter-lab comparison fulfilled the requirements of GB/T 27404-2008. The developed method therefore offers the advantages of simple sample pretreatment, short time demand, high sensitivity, and recovery rate, and is suitable for the analysis of the three compounds in food. 
Section overview
Introduction
Glyphosate is a broad-spectrum herbicide with toxicity similar to organophosphorus compounds and is one of the most frequently used herbicides in the world. Furthermore, the chemicals’ usage increased after the introduction of genetically modified, glyphosate-tolerant crops such as corn, soybeans, and cotton. In the USA, the US Environmental Protection Agency (EPA) regulation document 40 CFR 180 sets the tolerance levels for the occurrence of glyphosate in food commodities and products. The EPA tolerance for glyphosate residues in dried tea is set at 1 ppm1. In rice, the tolerance is 0.1 ppm, whereas in sweet corn it is 3.5 ppm. For glufosinate, an herbicide that is often analyzed alongside glyphosate, the tolerance values are 1.1 ppm for canola meal and 1.0 ppm for rice. These tolerance values include metabolites and degradation products. The EU describes a Maximum Residue Limit (MRL) for tea of 2 mg/kg.2 (Aminomethyl)phosphonic acid (AMPA) is the main degradation product of glyphosate and is also included in most analyses targeting glyphosate determination.3-5
The detection methods reported so far include ion chromatography (IC), gas chromatography (GC), gas chromatography-mass spectrometry (GC-MS), high-performance liquid chromatography (HPLC), and high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS). Because glyphosate and its main metabolite AMPA are highly water-soluble and highly polar amphoteric compounds, the above-mentioned methods display some drawbacks. HPLC and particularly GC methods typically require derivatization, which is adding complexity and handling hazardous reagents. These also easily react with water and matrix components and form by-products, making quantitative analysis more challenging. Although GC-MS technology is qualitatively accurate and has little interference, excessive use of derivatization reagents may contaminate ion sources.6-7
The tea matrix is complex in composition and contains polar impurities such as polyphenols, amino acids, sugars, and pigments. Conventional sample preparation methods for glyphosate detection are mainly divided into solid-phase extraction purification and derivatization. Both method steps are tedious and add costs. In contrast, a cleanup by QuEChERS is simple and easy to apply. Traditional materials for QuEChERS might tend to over-retain certain pesticides. The Supel™ QuE Verde, a new and improved adsorption and purification mix, allows for a balanced matrix removal and analyte recovery, in particular for green samples.
The Supel™ QuE Verde clean-up tube contains Supelclean™ ENVI-Carb™ Y graphitized carbon, PSA (primary secondary amine), and Z-Sep+, a zirconia-coated silica adsorbent packing. This mix allows efficient removal of main matrix components such as organic acids, polyphenolic polar compounds, lipidic impurities, and (green) pigments while maintaining good recovery of planar pesticides.
Here we describe a method for the determination of glyphosate, AMPA, and glufosinate-ammonium in green tea with QuEChERS sample cleanup, using the Supel™ QuE Verde for efficient matrix removal and analyte recovery. The HPLC-MS/MS analysis is performed using a porous graphitic carbon (PGC)-based HPLC column, the Supel™ Carbon LC, allowing for simpler mobile phase conditions (reversed phase, RP) and omitting HILIC conditions.
The results are evaluated against the quality criteria described by two Chinese national standards. For linearity of calibration, reproducibility, and recovery, the criteria stated in GB 23200.13-2016, “National Standard for Food Safety—Determination of 448 pesticides and related chemical residues in tea by liquid chromatography-mass spectrometry”8, which is the most authoritative national standard for pesticide detection in tea in China at present, were applied. Regarding sensitivity (LOD & LOQ) the criteria described in SN/T 1923-2007, “Method for the determination of glyphosate residues in food for imported and exported food—the HPLC-MS/MS method”3 were used as a benchmark, as these limit values for glyphosate, glufosinate-ammonium, and AMPA are stricter than those stated in GB23220.13-2006, ensuring the developed method also meets this criteria.
Chemical structures of Glyphosate (left), (aminomethyl)phosphonic acid (AMPA) (middle) and glufosinate-ammonium (right)
Structural formulas of glyphosate (1), (aminomethyl)phosphonic acid (AMPA, 2) and glufosinate-ammonium (3).
Experimental
Standard, sample, and reagent preparation
Sample pretreatment
  • About 500 g of a green tea sample was crushed/homogenized with a grinder, screened through a 20 mesh (0.85 mm) sieve, mixed thoroughly, and the resulting coarse powder was stored in a plastic container.
Reagent preparation
  • Extraction solvent: 400 mL of acetonitrile and 600 mL of water were combined to obtain a solution of acetonitrile/water 40:60 (v:v).
  • Mobile phase A: 1.9217 g ammonium carbonate was weighed into a 1 L volumetric flask, and water was filled to the mark to prepare a 20 mmol/L ammonium carbonate solution.
Standard preparation
  • Glyphosate, glufosinate, and AMPA stock solution I (10 mg/mL): Weigh 0.20 g of each glyphosate, glufosinate, and AMPA standards into a 20 mL amber glass volumetric flask, dissolve with water, and fill up to the mark with water. The resulting stock solution with a concentration of 10.0 mg/mL of each glyphosate, glufosinate, and AMPA has to be stored at 0-8 °C and protected from light.
  • Glyphosate, glufosinate, and AMPA stock solution II (100 mg/L): Pipette 0.1 mL of glyphosate, glufosinate, and AMPA stock solution I into a 10 mL amber glass volumetric flask and fill up to the mark with water to obtain stock solution II containing 100 mg/L of glyphosate, glufosinate, and AMPA. Store at 0-8°C and protected from light.
  • Glyphosate, glufosinate, and AMPA stock solution III (1.00 mg/L): Pipette 0.1 mL of glyphosate, glufosinate, and AMPA stock solution II into a 10 mL amber glass volumetric flask and fill up to the mark with water to obtain stock solution III containing 1.00 mg/L of glyphosate, glufosinate, and AMPA. Store at 0-8°C and protected from light.
  • Glyphosate, glufosinate, and AMPA standard solutions 1-6: Prepare a total of five standard solutions by pipetting 10 μL, 100 μL, 200 μL, 400 μL, 800 μL and 1500 μL, respectively, of glyphosate, glufosinate, and AMPA stock solution III into five individual 10 mL volumetric bottles. Fill up to the mark with acetonitrile/water 40:60 (v:v). Glyphosate, glufosinate, and AMPA concentrations of the resulting six standard solutions are 1.0 μg/L, 10.0 μg/L, 20.0 μg/L, 40.0 μg/L, 80.0 μg/L, and 150 μg/L, respectively.
Sample preparation
  • Extraction: Weigh 2 g ± 0.005 g of tea sample (accurate to 0.001 g) into a 50 mL centrifuge tube, add 20.0 mL of a mixture of acetonitrile/water (40:60, v:v), vortex at 3500 rpm for 2 min, perform ultrasonic extraction for 30 min, and centrifuge at 10000 rpm for 5 min to obtain a tea extract.
  • Cleanup: Pipette 2.0 mL of the extract into a 15 mL Supel™ QuE Verde (55442-U) centrifuge tube, vortex for 3 min at 2500 rpm, centrifuge for 3 min at 10,000 rpm, and filter the supernatant using a 0.22 μm syringe filter.
  • Spiking experiments: For the determination of recovery and precision, two samples were prepared by combining 2.0 g of a blank tea sample with 100 μL and 200 μL of glyphosate, glufosinate, and AMPA stock solution III, respectively. The glyphosate, glufosinate, and AMPA concentrations in the two resulting solutions are representing spiked samples of 50 µg/kg and 100 µg/kg, respectively.
    The tea sample spiked with a glyphosate, glufosinate, and AMPA concentration of 100 µg/kg was applied to analyze precision, and the one with 50 µg/kg was utilized for the determination of recovery.
LC-MS/MS analysis
The purified extracts were analyzed by HPLC-MS/MS (Table 1) using a Supel™ Carbon LC column with a volatile bicarbonate buffer in the mobile phase. This column possesses a unique mixed-mode retention mechanism that allows better retention of polar analytes without the need for HILIC conditions.
Note: Before first use, it is recommended to prime the Supel™ Carbon LC columns with sample matrix extracts (20-50 injections) to cover certain active sites on the surface for improved repeatability. 
HPLC Parameters
Instrument:
Waters™ Acquity I-class plus and Waters™ Xevo TQS UPS with electrospray ion source (ESI source)
Column:
Supel™ Carbon LC, 2.7 μm, 10 cm × 2.1 mm I.D. (59986-U)
Mobile phase:
[A] 20mM Ammonium carbonate (pH=9) (Add 1mL InfinityLab deactivating agent;
[B] Acetonitrile:water (95:5, v:v) (Add 1mL InfinityLab deactivating agent)
Gradient:
Time (min)
% A
% B
0
100
0
1.0
95
5
3.0
0
100
13.0
0
100
13.1
95
5
15.0
95
5
Flow rate:
0.30 mL/min
Column Temp.:
30 °C
Detector:
MSD, ESI-, MRM (See Table 2)
Injection:
5.0 μL
Table 1.HPLC-MS/MS conditions
MS conditions
Instrument:
Waters™ Xevo TQS UPS
Capillary voltage:
4.0 kV
Nebulizing gas flow:
3.0 mL/min
Drying gas flow:
10.0 mL/min
Heat block temperature:
400 °C
DL tube temperature:
250 °C
Interface temperature:
300 °C
Ionization mode:
ESI-
Scan:
MRM (see below)
MRM transitions
Compound
Parent (m/z)
Daughter (m/z)
Cone (V)
Collision (V)
Glyphosate 
Quant
168
63
-30
-26
Qual
168
124
-30
-16
AMPA 
Quant
110
63
-15
-28
Qual
110
80
-15
-36
Glufosinate 
Quant
180
63
-50
-56
Qual
180
95
-50
-24
Note: Method can be modified based on make and model of the Mass spectrometry and Mass parameters can be modified to obtain optimum response.
Table 2.MS instrument parameters
Results and discussion
After extraction and QuEChERS cleanup, the tea sample was submitted to HPLC-MS/MS analysis (multiple reaction monitoring mode). The quantification was performed by an external calibration approach. A chromatogram for the analysis of glyphosate, glufosinate, and AMPA standard solution (10 µg/L) is displayed in Figure 1, showing good resolution. The separation for a spiked tea sample (50 µg/kg) is shown in Figure 5.
A LC-MS/MS chromatogram obtained from the analysis of a standard solution containing glyphosate, glufosinate, and AMPA at a concentration of 10 μg/L. The x-axis represents retention time in minutes, ranging from 3 to 15, while the y-axis represents intensity in counts per second (cps), ranging from 0 to 1,000. The chromatogram, shown as a green line, highlights three distinct peaks corresponding to the target compounds. The first peak, labeled "Glufosinate," appears at approximately 4.36 minutes and rises sharply to an intensity of just under 400 cps. The second peak, labeled "AMPA," occurs at approximately 7.48 minutes and reaches a maximum intensity of about 650 cps. The third peak, labeled "Glyphosate," is observed at approximately 9.41 minutes with an intensity of just over 400 cps. The baseline is relatively stable and flat, depicted in green, with no significant interference between peaks. The clear, sharp peaks indicate efficient chromatography and precise detection and quantification of the compounds in the standard solution.
Figure 1.LC-MS/MS chromatogram obtained by the analysis of glyphosate, glufosinate, and AMPA standard solution 1 (10 μg/L). 1: Glufosinate (tr= 4.36 min), MRM transitions: 180.0 → 63.0 (quantitative); 2: AMPA (tr= 7.48 min), MRM transitions: 110.0 → 63.0 (quantitative); 3: Glyphosate (tr= 9.41 min), MRM transitions: 168.0 → 63.0 (quantitative).
Calibration and sensitivity
The results of the external calibration experiments utilizing glyphosate, glufosinate, and AMPA standard solutions 1-6 (c=1.0, 10.0, 20.0, 40.0, 80.0, and 150 µg/L) showed good linearity for all compounds with R2 values between 0.9987 and 0.9995(GB 23200.13-2016 criteria R2>0.9950).8 The calibration data for glyphosate, glufosinate, and AMPA are shown in Figures 2-4 and Tables 3 and 4.
The image shows a calibration curve representing the response of glyphosate at varying concentrations. The x-axis is labeled "Concentration (μg/L)" and spans from 0 to 200, while the y-axis is labeled "Response" and ranges from 0 to 4,500. Six blue data points are plotted along the curve at specific concentrations: 1.0, 10.0, 20.0, 40.0, 80.0, and 150 μg/L. The data points follow a linear trend, and a dashed blue line represents the best-fit line for the calibration data. The equation of the line, "y = 25.932x - 73," and the coefficient of determination, "R² = 0.9991," are displayed above the curve. The chart has a white background, with clear blue data points and a dotted blue trendline indicating high linearity and accuracy in the calibration measurements.
Figure 2.Calibration curve for glyphosate obtained by the analysis of standard solutions 1-6: (c = 1.0, 10.0, 20.0, 40.0, 80.0, and 150 μg/L).
A calibration curve plotting the response of glufosinate acid at various concentrations. The x-axis, labeled "Concentration (μg/L)," ranges from 0 to 200, while the y-axis, labeled "Response," ranges from 0 to 4,000. Six blue data points are evenly spaced along the curve, corresponding to concentrations of 1.0, 10.0, 20.0, 40.0, 80.0, and 150 μg/L. The data points align closely with a blue dashed line, representing the linear regression fit. The equation of the line, "y = 23.753x - 65.355," and the coefficient of determination, "R² = 0.9987," are displayed in black text near the top right of the graph. The background is white, with the chart prominently featuring blue elements, including the data points and the dotted trendline, to highlight the linearity of the calibration measurements.
Figure 3.Calibration curve for glufosinate acid obtained by the analysis of standard solutions 1-6: (c = 1.0, 10.0, 20.0, 40.0, 80.0, and 150 μg/L).
A calibration curve graph representing the response of AMPA at varying concentrations. The x-axis, labeled "Concentration (μg/L)," ranges from 0 to 200, while the y-axis, labelled "Response," spans from 0 to 2500. Six blue circular data points are plotted along the graph, corresponding to concentrations of 1.0, 10.0, 20.0, 40.0, 80.0, and 150 μg/L. These points align closely with a blue dotted trendline, indicating a strong linear correlation. Above the curve, black text displays the linear regression equation "y = 15.21x + 9.8513" and the coefficient of determination "R² = 0.9995," highlighting the accuracy of the linear relationship. The graph has a white background, and the use of blue for the data points and trendline ensures clarity and visual focus on the calibration data.
Figure 4.Calibration curve for AMPA obtained by the analysis of standard solutions 1-6: (c = 1.0, 10.0, 20.0, 40.0, 80.0, and 150 μg/L).
Sample
Concentration (μg/L)
Glyphosate area
Glyphosinate area
AMPA
area
Standard solution 1
1.0
18.90
19.5
14.5
Standard solution 2
10.0
195.30
200.30
155.31
Standard solution 3
20.0
395.82
380.81
340.82
Standard solution 4
40.0
915.91
815.91
625.91
Standard solution 5
80.0
2011.68
1811.68
1200.58
Standard solution 6
150
3829.87
3529.87
2300.07
Table 3.Responses obtained for glyphosate, glufosinate, and AMPA for the external calibration solutions
Compound
Calibration
R2
Glyphosate
y = 25.932X -73
0.9991
Glufosinate
y =23.753X – 65.355
0.9987
AMPA
y =15.21X + 9.8513
0.9995
Table 4.Overview of the calibration data utilizing glyphosate, glufosinate, and AMPA standard solutions 1-6 (c = 1.0, 10.0, 20.0, 40.0, 80.0, & 150 µg/L)
Precision and recovery
Seven individual tea samples spiked with glyphosate, glufosinate, and AMPA at a concentration of 100 µg/kg (0.1 ppm) were applied to analyze precision (Table 5), and six individual tea samples spiked with 50 µg/kg (0.05 ppm) of each compound were utilized for the determination of recovery (Table 6). The precision/reproducibility was in the range of 2.9 to 5.0% RSD (n=7), meeting the GB 23200.13-2016 criteria of <15%; the average recoveries (n=6) ranged from 85.5 to 97.6%. (GB 23200.13-2016 criteria 60-120%).
Precision (RSD)
Glyphosate (µg/kg)
Glufosinate (µg/kg)
AMPA (µg/kg)
Theoretical value
100
100
100
Sample 1
106
90.1
89.5
Sample 2
104
89.3
89.1
Sample 3
99.5
88.9
88.2
Sample 4
104
89.7
88.9
Sample 5
102
89.6
89.3
Sample 6
106
89.0
91.3
Sample 7
98.3
91.1
88.7
Average
103
89.7
89.3
RSD (%)
2.9
4.1
5.0
Table 5.Precision (RSD) of method at a glyphosate, glufosinate, and AMPA concentration of 100 µg/kg in tea (seven tea samples were prepared as parallel samples for detection and analysis)
Recovery
Glyphosate (µg/kg)
Glufosinate (µg/kg)
AMPA (µg/kg)
Theoretical value
50.0
50.0
50.0
Sample 1
53.4
47.4
42.4
Sample 2
44.0
44.5
41.7
Sample 3
49.9
43.9
40.5
Sample 4
50.6
44.6
42.6
Sample 5
43.3
42.7
45.1
Sample 6
51.8
41.8
43.9
Average
48.8
44.2
42.7
% Recovery (average)
97.6%
88.4%
85.5%
RSD (%)
4.2
1.9
1.6
Table 6.Percent recovery of the method at a glyphosate, glufosinate, and AMPA concentration of 50.0 µg/kg in tea (six tea samples were prepared as parallel samples for detection and analysis)
Inter-lab comparison
As per the national standard GB/T 27404-2008 “Laboratory quality control specification—physical and chemical testing of food,9” the method was additionally tested in an external lab using a second Supel™ Carbon LC column. The results showed a deviation of 8.6%, well within the GB requirement for inter-lab comparison set at <15%.
Sample measurements
LC-MS/MS chromatogram obtained by the analysis of a tea sample utilized for the determination of the method recovery rate (spiked at 50 µg/kg) is displayed in Figure 5.
An LC-MS/MS chromatogram represented by a green line on a white background, depicting the analysis of a tea sample spiked with glyphosate, glufosinate, and AMPA. The x-axis, labelled "Retention time (min)," ranges from 3 to 15 minutes, while the y-axis, labelled "Intensity (cps)," spans from -10 to 950. Three distinct peaks are annotated with their corresponding analytes: the first peak at approximately 4.36 minutes is labelled "Glufosinate," the second peak at around 7.48 minutes is labelled "AMPA," and the third peak at approximately 9.41 minutes is labelled "Glyphosate." The chromatogram's green trace is consistent and smooth between the peaks, highlighting the detection of the specified compounds.
Figure 5.LC-MS/MS chromatogram obtained by the analysis of a tea sample spiked with glyphosate, glufosinate, and AMPA standard solution III at a concentration of 50.0 µg/kg. 1: Glufosinate (tr= 4.36 min), MRM transitions: 180.0 → 63.0 (quantitative); 2: AMPA (tr= 7.48 min), MRM transitions: 110.0 → 63.0 (quantitative); 3: Glyphosate (tr= 9.41 min), MRM transitions: 168.0 → 63.0 (quantitative).
Peak no.
Compound
Retention time (min)
Detected mass
m/z
1
Glufosinate
4.36
180.0
2
AMPA
7.48
110.0
3
Glyphosate
9.41
168.0
Table 7.Chromatographic data for the analysis of a tea sample spiked at 50 µg/kg
An LC-MS/MS chromatogram displayed as a green trace on a white background, represen an LC-MS/MS chromatogram displayed as a green trace on a white background, representing the analysis of an unspiked tea sample. The x-axis is labelled "Retention time (min)" and spans from 3 to 15 minutes, while the y-axis is labelled "Intensity (cps)" and ranges from -10 to 90. The green line exhibits a consistent baseline with minor fluctuations and small peaks across the retention time range. No prominent or sharp peaks are visible, indicating the absence of detectable target analytes.
Figure 6.LC-MS/MS chromatogram obtained by the analysis of an unspiked tea sample.
Sensitivity of the Method
The sensitivity was determined using the baseline noise of a blank tea sample, where 3N/X was used to determine LOD and 10N/X was used to determine LOQ. Determined values and comparison data from the SN/T 1923-2007 method are shown in Table 8.
Peak no.
Compound
Experimental results
SN/T 1923-2007
LOD (µg/kg)
LOQ (µg/kg)
LOD (µg/kg)
LOQ (µg/kg)
1
Glyphosate
5.0
15.0
15.0
50.0
2
Glufosinate
9.0
27.0
20.0
60.0
3
AMPA
11.0
33.0
20.0
60.0
Table 8.LOQ & LOD of glyphosate, glufosinate, and AMPA (determined using baseline noise) and comparison values defined in the SN/T 1923-2007 method
InfinityLab deactivator additive
Glyphosate, glufosinate, and AMPA are highly polar, weakly acidic, phosphonic acid-containing compounds with very low hydrophobicity and limited retention in conventional reversed-phase chromatography. Their phosphonic acid groups can interact with active metal sites in the LC flow path, which may result in adsorption, poor peak shape, reduced sensitivity, and unstable retention times.
The InfinityLab deactivator additive is therefore added to the mobile-phase system to minimize analyte interaction with metal surfaces and improve chromatographic performance. For gradient elution, the additive is added to both the aqueous and organic mobile phases at the same concentration to maintain a consistent deactivator concentration throughout the gradient. This helps improve peak shape, analyte response, and retention-time reproducibility.
Conclusion
A U/HPLC-MS/MS method combined with solvent extraction and QuEChERS purification was established to determine glyphosate, glyfosinate, and AMPA residues in green/fresh tea. The samples were purified using a Supel™ QuE Verde adsorbent mix for green sample matrices and analysed by LC-MS using a Supel™ Carbon LC PGC UHPLC column that enabled RP conditions. Samples were quantified against an external standard calibration method. The results showed linear responses in the range of 1.0 to 150 μg/L for the injected solution with correlation coefficients (R2) greater than 0.998 for all three analytes. The observed recoveries were ranging from 81.0% to 107%, and the precision (RSD) was 2.7-5.0%. Therefore, linearity, RSD, and recovery rate were meeting the GB 23200.13-2016 method criteria. The determined sensitivities for LODs (µg/kg): glyphosate: 5.0; glufosinate: 9.0; AMPA:11.0; and for LOQ (µg/kg): glyphosate: 15.0; glufosinate: 27.0; AMPA:33.0. These meet or exceed the criteria of the SN/T 1923-2007 method. An inter-lab comparison showed reproducibility meeting the criteria of GB/T 27404-2008.
These results confirm the accuracy and reliability of the method for the detection and analysis of glyphosate, glufosinate, and AMPA in green tea.
Find more applications on Food & Beverage Testing.
Related products
HPLC column and QuEChERS clean up
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Solvents, reagents, and accessories
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Reference materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
REFERENCES
1.
40 CFR 180.364- Glyphosate; tolerances for residues. [Internet]. Ecfr.gov.[cited 28 May 2024]. Available from: https://www.ecfr.gov/current/title-40/chapter-I/subchapter-E/part-180/subpart-C/section-180.364
2.
EU Pesticides Database. [Internet]. Europa.eu.[cited 30 Sep 2024]. Available from: https://ec.europa.eu/food/plant/pesticides/eu-pesticides-database/start/screen/mrls/details?lg_code=EN&pest_res_id_list=120
3.
SN/T 1923-2007 Method for the determination of glyphosate residues in food for import and export - HPLC-MS/MS method. [Internet]. Available from: http://down.foodmate.net/standard/yulan.php?itemid=14462
4.
HJ 1071-2019 Water quality determination of glyphosate by high performance liquid chromatography. [Internet]. Available from: http://down.foodmate.net/standard/yulan.php?itemid=68923
5.
Pires NL, Passos CJS, Morgado MGA, Mello DC, Infante CMC, Caldas ED. 2020. Determination of glyphosate, AMPA and glufosinate by high performance liquid chromatography with fluorescence detection in waters of the Santarém Plateau, Brazilian Amazon. Journal of Environmental Science and Health, Part B. 55(9):794-802. https://doi.org/10.1080/03601234.2020.1784668
6.
NY/T 1096-2006 Determination of glyphosate residues in food.. [Internet]. Available from: http://down.foodmate.net/standard/yulan.php?itemid=10826
7.
GB/T 23750-2009 Determination of glyphosate residues in plant products- Gas chromatography-mass spectrometry. [Internet]. Available from: http://down.foodmate.net/standard/yulan.php?itemid=20193
8.
GB 23200.13-2016 National Standard for Food Safety - Determination of 448 pesticide and related chemical residues in tea -- Liquid chromatography-mass spectrometry. [Internet]. Available from: http://down.foodmate.net/standard/yulan.php?itemid=50184
9.
GB/T 27404-2008 Laboratory quality control specification - Physical and chemical testing of food. [Internet]. Available from: http://down.foodmate.net/standard/yulan.php?itemid=15544