Determination of Impurities in Clozapine API using Purospher® Star RP-18 endcapped HPLC column acc. to Ph. Eur. Monograph
Abstract
The clozapine related substances/impurities determination following the European Pharmacopeia (Ph. Eur.) monograph is demonstrated using a Purospher® Star RP-18 endcapped column. This method showed compliance with the Ph. Eur. system suitability criteria for the clozapine related substances test.
Section Overview
Introduction
The presence of potential toxic chemicals and impurities in drugs is one of the biggest challenges in the manufacturing of an active pharmaceutical ingredient (API). Therefore, it is important to identify these impurities during the manufacturing process to avoid issues related to the quality, efficacy, and safety of drugs. Screening and quantitation of these impurities in APIs is useful to identify potential problems when evaluating new suppliers, changing manufacturing sites, or during production scale-up. An HPLC-UV method was developed on a Purospher® STAR RP-18 endcapped column (125 x 4.6 mm, 5 µm) for the simultaneous quantification of the impurities in API for clozapine as per the European Pharmacopoeia method.1 Beyond what is stated in the European Pharmacopoeia monograph, individual LOD and LOQ limits for the impurities were established by performing an additional calibration for each impurity and clozapine.
Clozapine impurities in the Ph. Eur. monograph are (see structures Figure 1):
- Clozapine Impurity A - 7-chloro-5, 10-dihydro-11H-dibenzo[b,e][1,4]diazepin-11-one
- Clozapine Impurity B - 11,11′-(piperazine-1,4-diyl)bis(8-chloro-5H-dibenzo[b,e][1,4]diazepine)
- Clozapine Impurity C - 8-chloro-11-(piperazin-1-yl)-5H-dibenzo[b,e][1,4]diazepine
- Clozapine Impurity D - 1-[2-[(2-amino-4-chlorophenyl)amino]benzoyl]-4-methylpiperazine
Figure 1.Structures of clozapine and related impurities acc. to European Pharmacopeia (Ph. Eur.).
Experimental
Mobile Phase Preparation
- Solution A: Dissolve 2.04 g of potassium dihydrogen phosphate in 1000 mL of water and adjust the pH to 2.4 + 0.05 with diluted phosphoric acid.
Solution A is mixed with acetonitrile and methanol according to shares described in Table 1 for the mobile phase [A] and [B]. After mixing each mobile phase, sonicate for 20 min and filter through a 0.45 µm membrane filter.
Standard and Sample Preparation
Standards and samples were prepared according to the below procedures:
- Solvent mixture: Methanol and water (80:20 v:v).
- Test Solution: 75 mg of substance to be examined dissolved in Add 80 mL of methanol and swirl gently to dissolve before diluting to 100 mL with water. Mix well and filter through a 0.45 µm membrane filter.
- Reference solution A: 1 mL of Test Solution diluted to 10 mL with solvent mixture. Then 1 mL further diluted to 100 mL with a solvent mixture.
- Reference solution B: Dissolve the contents of a vial of Clozapine for peak identification CRS (containing impurities A, B, C and D) in 1 mL of solvent mixture.
- Stock solution for linearity (15 µg/mL): Weigh and transfer about 1.5 mg of each individual clozapine impurity A, B, C, D, and clozapine standard to 100 mL volumetric flask, respectively, add 80 mL of methanol to each flask and swirl gently to dissolve before diluting to 100 mL with water, then shake and mix well thoroughly.
HPLC-UV Analysis
The standards and samples were analyzed by HPLC-UV at 277 nm using a Purospher® STAR RP-18 endcapped column (Table 1).
HPLC Parameters | |||
|---|---|---|---|
Column: | Purospher® STAR RP-18 endcapped, 5 µm, 125 x 4.6 mm (1.51914) |
| |
Mobile phase: | [A] Acetonitrile:methanol:solution A (1:1:8, v:v:v); [B] Acetonitrile:methanol:solution A (4:4:2, v:v:v) |
| |
Gradient | Time (min) | %B |
|
0-4 | 0 |
| |
| 4-24 | 0-100 |
|
| 24-32* | 100 |
|
Flow rate: | 1.2 mL/min |
| |
Pressure: | 180 bar (2600 psi) |
| |
Column temp.: | Ambient |
| |
Detector: | UV, 257 nm |
| |
Injection volume: | 20 µL |
| |
Samples: | as described above and in the figures below |
| |
Results & Discussion
System Suitability
The system suitability was assessed by comparing the chromatogram acquired with reference solution B (Figure 2) with the chromatogram supplied with Clozapine peak identification CRS (Y0000758; Figure 3), which have to be similar, and the resolution between impurity C and clozapine (>2.5) as indicated in the Ph. Eur. monograph. Both criteria were met. An overview of the suitability criteria and the achieved results is shown in Table 4.
Figure 2.Chromatogram recorded for clozapine reference solution B, prepared in 80% v/v of methanol in water, as per Ph. Eur. monograph for clozapine related substance method.
Figure 3.Example chromatogram provided with clozapine peak identification CRS (Y0000758, batch 3).
Peak No. | Name | RT (min) | RRT | RRT in Ph. Eur. | Peak Area | Resolution | Tailing Factor |
|---|---|---|---|---|---|---|---|
1 | Clozapine Imp C | 9.183 | 0.9 | about 0.9 | 39801 | - | 1.12 |
2 | Clozapine | 10.554 | 1.0 | - | 566548 | 4.80 | 1.20 |
3 | Clozapine Imp D | 13.449 | 1.3 | about 1.1 | 29520 | 11.58 | 1.07 |
4 | Clozapine Imp A | 17.306 | 1.6 | about 1.6 | 174282 | 18.79 | 1.06 |
5 | Clozapine Imp B | 17.993 | 1.7 | about 1.7 | 47975 | 2.95 | 1.09 |
Repeatability
The repeatability for the peak area and the retention time was assessed using the reference solution A (Figure 2 & Table 3)
Figure 4.Chromatogram for clozapine reference solution A, prepared in 80% v/v of methanol in water, as per Ph. Eur. monograph for clozapine related substance method (clozapine RT= 10.592 min).
Standard | Peak Area | Retention Time (min) |
|---|---|---|
STD_1 | 50105 | 10.592 |
STD_2 | 50332 | 10.591 |
STD_3 | 50267 | 10.593 |
STD_4 | 50357 | 10.594 |
STD_5 | 50571 | 10.591 |
Mean | 50326.4 | 10.5922 |
Standard Deviation | 168.3324 | 0.0013 |
% RSD | 0.33 | 0.01 |
Ph. Eur. Criteria %RSD | NMT 0.73% | - |
Summary of Suitability Criteria
A comparison of system suitability test (SST) criteria as per Ph. Eur monograph for clozapine with the observed value of reference solution A and reference solution B as well as applicability of Ph. Eur 2.2.46 general chapter criteria for % RSD of five replicate injections and tailing factor2 are provided in Table 4.
SST Parameters | EP Limits | Observed Value |
|---|---|---|
Resolution between impurity C and clozapine | Minimum 2.5 | 4.8 |
Chromatogram obtained with reference solution B | Chromatogram obtained with refence solution B should be similar to chromatogram supplied with clozapine for peak identification CRS. | Chromatogram obtained with refence solution B is similar to chromatogram supplied with clozapine peak identification |
Relative retention time with reference to clozapine (retention time about 11 min) | Imp A = about 1.6 | Imp A = about 1.63 |
Imp B = about 1.7 | Imp B = about 1.70 | |
Imp C = about 0.9 | Imp C = about 0.87 | |
Imp D = about 1.1 | Imp D = about 1.27 | |
% RSD of Peak Area reference solution A | NMT 0.73% | 0.33% |
Tailing factor (as per Ph. Eur 2.2.46) | 0.8 – 1.8 | Imp A=1.06 |
Imp B = 1.09 | ||
Imp C = 1.12 | ||
Imp D = 1.09 |
Calibration
In addition to what is described in the Ph. Eur. preparation of calibration level solutions were done for each individual impurity and clozapine to assess linearity of calibration and limit of detection and quantification (LOD & LOQ). The calibration levels for each individual impurities were run from 25 % to 150% of the limit mentioned in the Ph. Eur. monograph Table 5.
Impurity Name | Calibration Range (µg/mL) | Calibration Equation | R2 | LOD (µg/mL) | LOQ (µg/mL) |
|---|---|---|---|---|---|
Clozapine | 0.1875 -1.125 | y = 66137x - 1527.3 | 0.9945 | 0.1 | 0.29 |
Clozapine Impurity A | 0.1875 - 1.125 | y = 218340x + 10169 | 0.9974 | 0.07 | 0.2 |
Clozapine Impurity B | 0.375 - 2.250 | y = 33950x - 649.67 | 0.9953 | 0.18 | 0.54 |
Clozapine Impurity C | 0.560 - 3.380 | y = 18718x + 2969.6 | 0.9973 | 0.2 | 0.61 |
Clozapine Impurity D | 0.375 - 2.250 | y = 18041x + 1937.8 | 0.9952 | 0.18 | 0.54 |
Measurement of a Test Solution
Figure 5.Chromatogram for clozapine test solution, prepared in 80% v/v of methanol in water, as per EP monograph for Clozapine method for related substances.
Compound | % Impurity present in test solution |
|---|---|
Clozapine Impurity A | 4.48 |
Clozapine Impurity B | 0.51 |
Clozapine Impurity C | - |
Clozapine Impurity D | - |
Conclusion
The demonstrated method shows that with the Purospher® Star RP-18 endcapped, 5 µm, 125 x 4.6 mm column the system suitability criteria for the determination of clozapine related impurities following the Ph. Eur. monograph can be met. The resolution obtained between the peaks of clozapine and clozapine impurity C is 4.8. The relative retention time obtained for the impurities were also comparable with the values listed in the EP monograph.
Although the EP monograph method is used for impurity determination by comparing peak areas, this method here was taken one step ahead beyond the requirements of the Ph. Eur. monograph, by exactly calibrating each individual impurity with single compounds solutions in the concentration range of 25% -150% of the limits mentioned in the monograph. The method showed for the applied concentration ranges a linear behavior with R2 values ≥0.995. The determined LODs and LOQs for the HPLC method were between 0.07 - 0.20 mg/mL and 0.20 - 0.61 µg/mL respectively.