MilliporeSigma
HomeSmall Molecules Analysis & QCGlycol Analysis in Cough Syrup by GC MS
Glycol Analysis in Cough Syrup by GC-MS Using an Equity-1701 Capillary GC Column
Abstract
Gas chromatography coupled to mass spectrometry detection (GC-MS/MS) was used as an analytical tool to analyze glycols present in commercial cough syrup samples with an Equity-1701 Capillary GC Column 30 m × 0.25 mm, df 0.25 μm. The developed method is able to separate and quantify ethylene glycol, propylene glycol, diethylene glycol, glycerin, triethylene glycol, and sorbitol present in the commercial cough syrup preparations. For the quantification, certified reference materials (CRMs) were used. The method is partially validated for establishing the performance characteristics.
Section Overview
Introduction
Glycols are chemical compounds characterized by two hydroxyl groups attached to separate carbon atoms, commonly referred to as aliphatic diols.1 Various forms of glycols are utilized in industries for different purposes, such as liquid desiccants in air conditioning systems, antifreeze in car radiators2, and are also used as additives in hydraulic and brake fluids. Ethylene glycol is recognized as the simplest type. Diethylene, triethylene, and propylene glycols are oligomers of ethylene glycol. Ethylene glycol is a potent cause of acute toxicity in humans, in contrast, propylene glycol is a generally recognized as safe additive for foods and medications.3 Propylene glycol is generally considered safe, but when used in high doses or used for prolonged periods, propylene glycol toxicity can occur with reported adverse effects includes central nervous system (CNS) toxicity, hyperosmolarity, hemolysis, cardiac arrhythmia, seizures, agitation, and lactic acidosis.4 Typically, colorless, and odorless, ethylene glycol and diethylene glycol have a sweet taste5 and are often considered inexpensive adulterants, making them attractive for commercial sectors involved in drug formulation, including pediatric drugs.
However, when glycols are added to drug formulations beyond recommended limits, they can be toxic to human health, potentially leading to acute hepatotoxicity, renal failure, and even death in some cases. In recent times fatal incidences were reported e.g. from Indonesia and Gambia where of ethylene glycol and diethylene glycol were found in syrup medicines. 6,7,8 As a response the US Food and Drug Administration (US FDA) provided a guidance for high-risk drug components9 and the United States Pharmacopoeia (USP) revised the monograph for propylene glycol that addresses the toxicity concerns by specifying the limits for ethylene glycol and diethylene glycol in formulation components such as propylene glycol to be not more than (NMT) 0.1% to ensure quality standards for pharmaceutical products.10
This article will present a method for the simultaneous determination of five glycols and sorbitol in a cough syrup sample using GC-MS/MS. Two commercially available cough syrups are analyzed unspiked and spiked. The developed method was partially validated as per the ICH recommended guidelines11 and used for determination of minimum limit of detection and quantification of glycols. Furthermore, two commercially available cough syrups are analyzed and compared to the occupational health and safety limits and permissible limits in drug products stated by USP.
The Equity-1701 column fulfils USP G46 requirements with 14% cyanopropylphenyl - 86% dimethylpolysiloxane and the max temperature limit of this column is 280 ̊C. Even though this column is from the mid polar range, the Equity‑1701 stationary phase along with its temperature characteristics was found capable for additional determination of sorbitol along with other glycol present in the samples. Since Sorbitol is exhibiting a very high boiling point of 296 °C, in many cases, the reported method of choice for sorbitol is HPLC-RI/ELSD. Here it was included as additional analyte for the GC-MS/MS technique to further explore the capability of the developed method on the selected Equity-1701 phase. 
Molecular structures of six different glycols: ethylene glycol (EG), propylene glycol (PG), diethylene glycol (DEG), triethylene glycol (TEG), glycerin (Gly), and sorbitol (SOR). Each molecule is represented using black lines for chemical bonds and black letters for atoms, drawn on a white background. Ethylene glycol consists of a two-carbon chain with hydroxyl (-OH) groups attached to both ends. Propylene glycol has a three-carbon chain with hydroxyl groups on the first and second carbons. Diethylene glycol and triethylene glycol extend the ethylene glycol structure by incorporating ether (-O-) linkages between additional ethylene units. Glycerin consists of a three-carbon backbone, each carbon attached to a hydroxyl group. Sorbitol has a six-carbon backbone with hydroxyl groups attached to each carbon, forming a polyol structure. The names of each compound, along with their abbreviations, are labeled in black text below their respective structures. The molecules are neatly arranged in two rows, with four structures in the top row and two in the bottom row.
Structures of compounds analyzed in this study.
Experimental
Standard preparation
The glycol standards were prepared as single component solutions of ethylene glycol (EG), propylene glycol (PG), diethylene glycol (DEG), glycerin (Gly), triethylene glycol (TEG), Sorbitol (SOR) according to the following procedures using certified reference materials (CRMs) and methanol/water (95:5, v/v) as diluent.
  • Stock solutions glycols (500 µg/mL): Weigh and transfer about 10 mg of glycol substance into a 20 mL volumetric flask, add about 10 mL of diluent and sonicate for 5 minutes, make up to volume with diluent and shake to mix thoroughly. The concentration of the glycol in the resulting solution is 500 µg/mL.
  • Stock solutions sorbitol (1000 µg/mL): Weigh and transfer about 10 mg of sorbitol into a 10 mL volumetric flask, add about 10 mL of diluent and sonicate for 5 minutes, make up to volume with diluent and shake to mix thoroughly. The concentration of the sorbitol in the resulting solution is 1000 µg/mL.
  • Standard solution (SS, all glycols 20 µg/mL, sorbitol 200 µg/mL): Transfer 400 µL of each glycol stock solution and 2 mL of Sorbitol stock solution into a 10 mL volumetric flask, add 5 mL of diluent and sonicate for 5 minutes. Then make up to volume with diluent and mix thoroughly.
Calibration Solutions
Calibration solutions were prepared as single-component solutions using the following dilution schemes:
Transfer of 100, 250, 500, 750, and 1000 µL from each of the above-mentioned glycol standard stock solutions of ethylene glycol, propylene glycol, diethylene glycol, glycerin and triethylene glycol into 10 mL volumetric flask individually and to each individual flasks add 2, 2.5, 3, 3.5 and 4 mL of sorbitol stock solution and make up to volume with diluent, sonicate for 5 minutes and mix thoroughly. Resulting solutions have 5.0, 12.5, 25.0, 37.5, and 50.0 µg/mL for each glycol and 200.0, 250, 300, 350, and 400 µg/L sorbitol respectively.
Sample Preparation
Transfer 12 mg of commercially available cough syrup into a 10 mL volumetric flask, add 5 mL of diluent and sonicate for 5 minutes. Fill up to mark with diluent and mix thoroughly. Two commercially available syrups (A & B) were used for the assessments.
Spiked Samples
  • Spiked sample 1 PG & Gly (20.8 mg/g): Weigh 12 mg of syrup sample A into a 10 mL volumetric flask and add 500 µL of each of PG and Gly stock solutions. Make up to mark with diluent, mix thoroughly and sonicate for 5 minutes. Solution contains 25 µg/mL of each glycol.
  • Spiked sample 2 PG & Gly (41.7 mg/g): Weigh 12 mg of syrup sample A to a 10 mL volumetric flask and add 1000 µL of each of PG and Gly stock solutions. Make up to mark with diluent, mix thoroughly and sonicate for 5 minutes. Solution contains 50 µg/mL of each glycol.
  • Spiked Sample 3 PG, Gly (20.8 mg/g): Weigh 12 mg of syrup sample B into a 10 mL volumetric flask and add 500 µL of each of PG and Gly stock solutions. Make up to mark with diluent, mix thoroughly and sonicate for 5 minutes. Solution contains 50 µg/mL of each glycol.
  • Spiked sample 4 PG, Gly (41.7 mg/g): Weigh and transfer 12 mg of commercially available sample B to a 10 mL volumetric flask, and add 1000 µL of each of PG, Gly stock solutions. Make up to mark with diluent, mix thoroughly and sonicate for 5 minutes. Solution contains 50 µg/mL of each glycol.
GC-MS Analysis
The GC-MS analysis was performed on an intermediate polarity Equity-1701 column (Table 1). Depending upon the fragmentation recorded for each individual glycols, the selection of specific target ions and reference ions were selected for scanning at a defined retention time. Selection of the retention time was done as per the separation acquired by the developed method on Equity-1701 column (Table 2).
GC-MS Parameters
Instrument
Agilent 7000 Series Triple Quad GC-MS with ALS
Column:
Equity-1701, 30 m × 0.25 mm I.D., df 0.25 μm (28372-U)
Oven:
Initial temperature (°C)
Temp. ramp (°C/min)
Final temp. (°C)
Hold time at final temperature (min)
60
-
60
4
60
10
150
4
150
15
280
5
Inj. temp.:
250 ̊C
Carrier gas:
Helium, 1.3 mL/min
Detector:
MS/MS
MSD interface:
285 °C
Injection:
0.8 µL splitless
Liner
Spitless single taper
Run time
30.667
Samples:
Standards, unspiked and spiked samples all in methanol/water (95:5, v/v)
MS Conditions
Acquisition:
Automatic Liquid Sampler
Quench gas:
Helium
Solvent delay:
4.5 minutes
MS Source temperature
270 °C
Quad Temperature:
150 ̊C
Electron energy
70 eV
Dwell time:
100 ms
Quenching gas flow rate
2.25 mL/min
MS measurement mode
SIM
MS mode
Splitless
Solvent delay
4.5 min
Ion source
EI
Gain factor
10
Scan type
MS1 SIM (see Table 2)
Table 1.GC -MS conditions used for glycol determination
Compound
Retention time (min)
Target Ion m/z (Quantifier)
Reference Ion m/z (Qualifier)
Ethylene glycol
5.6
43.0
62.0
61.1
46.1
44.0
Propylene glycol
6.2
45.1
76.1
61.0
57.0
43.1
Diethylene glycol
10.8
75
76.0
45.1
44.1
43.1
Glycerin
11.9
61
66.0
62.1
44.1
43.1
Triethylene Glycol
15.1
89
87.0
75.0
58.0
45.1
Sorbitol
25.2
73
132.9
117.0
103.0
61.0
Table 2.MS SIM mode parameter (scan type MS1 SIM) used for glycol determination
Results and Discussion
The developed method applying an Equity-1701 (30 m × 0.25 mm I.D., df 0.25 μm) capillary GC column and splitless injection showed good retention and sufficient resolution of the analytes in scope. A representative chromatogram of an analyte mixture at 20 µg/mL each (sorbitol at 200 µg/mL) is shown in Figure 1.
GC-MS chromatogram using selected ion monitoring (SIM), showing the separation of six compounds in a standard solution mixture. The x-axis represents retention time in minutes, ranging from approximately 4.5 to 29.5 minutes, while the y-axis represents intensity in counts, ranging from -1.0 × 10⁷ to 2.9 × 10⁸. The chromatogram consists of six labelled peaks, each corresponding to a different compound eluted at specific retention times. The first peak, representing ethylene glycol, appears at the lowest retention time, followed by a second peak for propylene glycol. A significantly intense third peak, corresponding to diethylene glycol, is observed at around 10.5 minutes. The fourth peak, for glycerin, is smaller and appears between the third and fifth peaks. Triethylene glycol elutes at the fifth peak, which is another intense signal. The final peak, corresponding to sorbitol, appears at a much later retention time, around 25.5 minutes, with moderate intensity. The chromatogram is plotted with a fine black line on a white background, and the peaks are labeled with black numbers placed above them.
Figure 1.GC-MS chromatogram (SIM) for the standard solution (SS) mixture containing ethylene glycol (1), propylene glycol (2), diethylene glycol (3), glycerin (4), triethylene glycol (5), prepared in diluent at 20 µg/mL and sorbitol (6) at 200 µg/mL.
Calibration
The quantification was based on an external calibration using single reference standard solutions as described in the experimental section. The GC-MS method linearity for ethylene glycol, propylene glycol, diethylene glycol, glycerin and triethylene was established in the concentration range of 5 to 50 μg/mL, and for sorbitol 200 to 400 µg/mL. In Tables 3 & 4, the experimental results including the linearity (R2) and the limits of detection (LOD) and limits of quantification (LOQ) are summarized for all analytes. Based on the developed method the minimum concentrations of glycols that can be detected in the injected solutions as per the ICH guidelines11 by using this method’s linearity study is ranging for the five glycols from 3.33 µg/mL to 4.49 µg/mL, for sorbitol 50.12 µg/mL, and the quantification can be done in the range of 10.09 µg/mL to 13.6 µg/mL, for sorbitol 126.58 µg/mL. Derived from the linearity study data the estimated potential LODs of glycols present in the original cough syrup samples are in the range of 2.78 mg/g to 3.74 mg/g, for sorbitol 41.74 mg/g, and the LOQs in the range of 8.41 mg/g to 11.33 mg/g, for sorbitol 126.58 mg/g.
Conc (µg/mL)
Ethylene glycol (EG)
Peak area
Propylene glycol (PG)
Peak area 
Diethylene glycol (DEG) 
Peak area
Glycerin (Gly)
Peak area 
Triethylene glycol (TEG)
Peak area 
STEYX
906325
16718657
29236081
25861524
29214647
Slope
784013
15560508
28973976
19022370
21529257
R2
0.997
0.997
0.997
0.995
0.995
LOD (µg/mL)
3.81
3.55
3.33
4.49
4.48
Derived LOD in cough syrup (mg/g)
3.18
2.96
2.78
3.74
3.73
LOQ (µg/mL)
11.56
10.74
10.09
13.6
13.57
Derived LOQ in cough syrup (mg/g)
9.63
8.95
8.41
11.33
11.31
Table 3.Calibration and linearity data for peak responses recorded for single component solutions of glycols in concentration range of 5.0 μg/mL to 50 μg/mL and sensitivity data LOD & LOQ for the injected solutions of ethylene glycol, propylene glycol, diethylene glycol, glycerin and triethylene glycol.
Conc (µg/mL)
Sorbitol (SOR) Peak Area
Slope
5129714
R2
0.973
LOD (µg/mL)
50.12
Derived LOD in cough syrup (mg/g)
41.77
LOQ (µg/mL)
151.90
Derived LOQ in cough syrup (mg/g)
126.58
Table 4.Linearity data for peak responses recorded for single component solutions of glycols in concentration range of 200 μg/mL to 400 μg/mL and sensitivity data LOD&LOQ for the injected solutions of sorbitol
Reproducibility
The system repeatability was demonstrated by injecting five replicates of the standard solution (SS) containing mixture of glycol standards of ethylene glycol (EG), propylene glycol (PG), diethylene glycol (DG), glycerin (Gly), triethylene glycol (TG) and sorbitol (SOR) prepared in diluent (see Figure 1 and for diluent blank Figure 2) The determined RSD ranged from 3.63-8.89% (Table 5).
A chromatogram graph with a white background, showing a single black line tracing the intensity of a sample over a retention time of 5 to 30 minutes. The x-axis represents retention time in minutes, while the y-axis represents intensity in counts, ranging from approximately -1.0E+07 to 3.9E+08. The graph displays a mostly flat baseline with a very small peak just after the 5-minute mark and a gradual, slight rise in intensity beginning around 22 minutes, leveling off near 27 minutes. No significant peaks or sharp signals appear throughout the chromatogram, indicating a clean, low-activity result typical of a solvent blank.
Figure 2.GC-MS chromatogram recorded for diluent (blank) containing methanol and water (95:5 v/v).
Standard
EG
PG
DEG
GLY
TEG
SOR
Mean (n=5)
7505661
264558633
429784111
232578815
316372626
259053506
Standard deviation
272646
21108538
38225347
17822033
16963196
10853630
% RSD
3.63
7.98
8.89
7.66
5.36
4.19
Table 5.Repeatability data obtained for the standard solution mixture containing mixture of glycol standards of ethylene glycol (EG), propylene glycol (PG), diethylene glycol (DG), triethylene glycol (TG), glycerin (Gly), all at 20 µg/mL, and Sorbitol (SOR) at 200 µg/mL as depicted in Figure 1
Recovery Determination
For recovery determinations four solutions were prepared with 2 spike concentration levels 20.8 µg/g and 41.7 µg/g (25 and 50 µg/mL in the injection solutions) and two sets of compounds (Table 6 & 7) and 2 syrups used (A&B). For recovery calculation existing glycol background was subtracted.
Compound
Concentration determined (µg/mL)
Recovery
Spiked sample 1 (20.8 mg/g → 25 µg/mL) 
Propylene Glycol
20.5
82.0%
Glycerin
40.4
80.8%
Spiked sample 2 (41.7 mg/g → 50 µg/mL)
Propylene Glycol
24.5
98.0%
Glycerin
54.0
108.0%
Table 6.Results of recovery determination using syrup sample A spiked with PG and Gly at 20.8 µg/g and 41.7 µg/g (conc in injected solution 25 and 50 µg/mL) of each component.
Compound
Concentration determined (µg/mL)
Recovery
Spiked sample 3 (20.8 mg/g → 25 µg/mL)  
Propylene Glycol
18.4
73.6%
Glycerin
38.6
77.2%
Spiked sample 4 (41.7 mg/g → 50 µg/mL)
Propylene Glycol
21.6
86.4%
Glycerin
56.4
112.8%
Table 7.Results of recovery determination using syrup sample B spiked with PG, and Gly at 20.8 µg/g and 41.7 µg/g (conc. in injected solution 25 and 50 µg/mL) of each component
Original Sample Analysis
The method was applied for analyzing two different commercially available cough syrup samples (A & B) solutions (see Figures 3  & 4). Table 8 shows the results for the analysis of glycols present in the two commercially available cough syrup samples (A & B).
Gas chromatography-mass spectrometry (GC-MS) chromatogram displaying the separation of compounds in a sample. The x-axis represents retention time in minutes, ranging from approximately 5 to 30 minutes, while the y-axis represents intensity in counts, ranging from 0.00E+00 to approximately 6.00E+07. The chromatogram features two distinct peaks, labeled as 1 and 2, appearing at early retention times. The first peak is observed 6.216 minutes with moderate intensity, while the second peak, which is significantly higher, appears around 11.945 minutes. Following these peaks, the signal remains relatively low until a gradual increase is observed after 24 minutes, forming a broad elevation in intensity. The chromatogram is drawn with a fine black line on a white background, with peak labels in black text placed above their respective peaks.
Figure 3.GC-MS chromatogram recorded for commercially available cough syrup sample A prepared in diluent, showing the presence of propylene glycol (1) and glycerin (2).
Gas chromatography-mass spectrometry (GC-MS) chromatogram displaying the separation of compounds in a sample. The x-axis represents retention time in minutes, ranging from approximately 5 to 30 minutes, while the y-axis represents intensity in counts, ranging from 0.00E+00 to approximately 6.00E+07. The chromatogram features two distinct peaks, labeled as 1 and 2, appearing at early retention times. The first peak is observed 6.216 minutes with moderate intensity, while the second peak, which is significantly higher, appears around 11.945 minutes. Following these peaks, the signal remains relatively low until a gradual increase is observed after 24 minutes, forming a broad elevation in intensity. The chromatogram is drawn with a fine black line on a white background, with peak labels in black text placed above their respective peaks.
Figure 4.GC-MS chromatogram recorded for commercially available cough syrup sample B, prepared in diluent, showing the presence of propylene glycol (1), glycerin (2).
Sr. No.
Name
Retention Time
Glycols in sample A
mg/g
Glycols in sample B
mg/g
1
Ethylene glycol
5.602
Not detected
Not detected
2
Propylene glycol
6.216
7.23
5.73
3
Diethylene glycol
10.818
Not detected
Not detected
4
Glycerin
11.945
16.33
13.52
5
Triethylene glycol
15.117
Not detected
Not detected
6
Sorbitol
25.465
Not detected
Not detected
Table 8.Results depicted in the table below is the concentration of glycols present in two commercially available cough syrups samples represented in mg/g of sample
Conclusion
This study presents a GC-MS/MS method for quantifying ethylene glycol, propylene glycol, diethylene glycol, triethylene glycol, glycerin and sorbitol present in formulations for patients such as cough syrup. The method development utilized an Equity-1701 30m x 0.25 mm column with a film thickness of 0.25 µm. The % RSD for system repeatability with standard solution (SS) of the glycol mixture was well below 10% (3.63 to 8.89%). Derived from the external calibration data (R2≥0.962) the estimated potential LODs of glycols present in the original cough syrup samples for 5 compounds in the range of 2.78 mg/g to 3.74 mg/g, 50.12 mg/g for sorbitol), and the LOQs in the range of 8.41 mg/g to 11.33 mg/g, for sorbitol 126.5 mg/g. Analysis of the commercial sample A spiked with propylene glycol and glycerin showed recovery rates between 80.8% and 108 %. The second commercial sample B also spiked with propylene glycol and glycerin displayed recovery rates ranging from 73.6% to 112.8%. Sorbitol being more denser and possess higher boiling point as compared to other glycols, the development required higher concentration for sorbitol on GC-MS/MS technique, thus the studies of linearity and repeatability exhibits higher sorbitol concentrations.
Read more about Pharmeceutical Analysis & Quality Control
Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
References
1.
Forkner MW, Robson JH, Snellings WM, Martin AE, Murphy FH, Parsons TE, Albin BA, Burgess LM, Proulx G. 2022. Glycols. Encyclopedia of Chemical Technology.1-37. https://doi.org/10.1002/0471238961.0520082506151811.a01.pub3
2.
Rebsdat S, Mayer D. 2000. Ethylene Glycol. Ullmann’s Encyclopedia of Industrial Chemistry. https://doi.org/10.1002/14356007.a10_101
3.
2023 October 6. Ethylene Glycol and Propylene Glycol Toxicity. [Internet]. Agency for Toxic Substances and Disease Registry. Available from: https://archive.cdc.gov/www_atsdr_cdc_gov/csem/ethylene-propylene-glycol/propylene_glycol
4.
Lim TY, Poole RL, Pageler NM. 2014. Propylene Glycol Toxicity in Children. J Pediatric Pharmacology and Therapeutics. 19(4):277-282. https://doi.org/10.5863/1551-6776-19.4.277
5.
Schep LJ, Slaughter RJ, Temple WA, Beasley DMG. 2009. Diethylene glycol poisoning. Clinical Toxicology. 47(6):525-535. https://doi.org/10.1080/15563650903086444
6.
Fikri E, Firmansyah YW. 2003. A Case Report of Contamination and Toxicity of Ethylene Glycol and Diethylene Glycol on Drugs in Indonesia. Environment and Ecology Research. 11(2):378-384. https://doi.org/10.13189/eer.2023.110211
7.
MohanaSundaram A, Padhi BK, Mohanty A, Shrestha S, Sah R. 2023. The silent epidemic of substandard and falsified medicines in low- and middle-income countries: heed lessons from the tragic deaths of children in Indonesia. International journal of surgery (London, England). 109(3):523-525. https://doi.org/10.1097/js9.0000000000000059
8.
Saied AA, Metwally AA, Dhama K. 2023. Gambian children’s deaths due to contaminated cough syrups are a mutual responsibility. International journal of surgery (London, England). 109(2):115-116. https://doi.org/10.1097/js9.0000000000000112
9.
USP-NF Propylene Glycol Monograph: USP47-NF42. Pharmacopeial Forum: Volume No. PF 33(2). USP-NF Propylene Glycol. [Internet]. Available from: https://www.uspnf.com/sites/default/files/usp_pdf/EN/USPNF/propyleneGlycol.pdf
10.
Ballantyne B, Snellings WM. 2007. Triethylene glycol HO(CH2CH2O)3H. J of Applied Toxicology. 27(3):291-299. https://doi.org/10.1002/jat.1220
11.
ICH Harmonised Guideline ICH Q2 (R2) – Validation of Analytical Procedures. [Internet]. Available from: https://database.ich.org/sites/default/files/ICH_Q2-R2_Document_Step2_Guideline_2022_0324.pdf