MilliporeSigma
HomeSmall Molecules Analysis & QCGlycol Analysis using a GC Capillary Column
Glycol Analysis in Cough Syrup Using an SPB®-624 GC Capillary Column
Ajay Kaparwan
R&D APAC Lab, Jigani, Bangalore, India
Abstract
Gas chromatography coupled to mass spectrometry detection (GC-MS/MS) was used as an analytical tool to analyze glycols present in commercial cough syrup samples with an SPB®-624 Capillary GC Column (60 m × 0.32 mm, df 1.80 μm). The developed method is able to separate and quantify ethylene glycol, propylene glycol, diethylene glycol, glycerin, and triethylene glycol and was used for commercial cough syrup preparations. For the quantification, certified reference materials (CRMs) were used. The method is partially validated for establishing the performance characteristics.
Section Overview
Introduction
Glycols are chemical compounds characterized by two hydroxyl groups attached to separate carbon atoms, commonly referred to as aliphatic diols.1 Various forms of glycols are utilized in industries for different purposes, such as liquid desiccants in air conditioning systems, antifreeze in car radiators2, and are also used as additives in hydraulic and brake fluids. Ethylene glycol is recognized as the simplest type. Diethylene, triethylene, and propylene glycols are oligomers of ethylene glycol. Ethylene glycol is a potent cause of acute toxicity in humans, in contrast, propylene glycol is a generally recognized as safe additive for foods and medications.3 Propylene glycol is generally considered safe, but for use in high doses or for prolonged periods, adverse effects including central nervous system (CNS) toxicity, hyperosmolarity, hemolysis, cardiac arrhythmia, seizures, agitation, and lactic acidosis are reported.4 Typically, colorless, and odorless, ethylene glycol and diethylene glycol have a sweet taste5 and are often considered inexpensive adulterants, making them attractive for commercial sectors involved in drug formulation, including pediatric drugs.
However, when glycols are added to drug formulations beyond recommended limits, they can be toxic to human health, potentially leading to acute hepatotoxicity, renal failure, and even death in some cases. In recent times fatal incidences were reported, e.g., from Indonesia and Gambia where ethylene glycol and diethylene glycol were found in syrup medicines. 6,7,8. As a response, the US Food and Drug Administration (US FDA) provided guidance for high-risk drug components 9 and the United States Pharmacopoeia (USP) revised the monograph for propylene glycol that addresses the toxicity concerns by specifying the limits for ethylene glycol and diethylene glycol in formulation components such as propylene glycol to be not more than (NMT) 0.1% to ensure quality standards for pharmaceutical products.10
Here a method for the simultaneous determination of 5 glycols using GC-MS/MS and certified reference materials (CRMs) for calibration is presented. Two commercially available cough syrups are analyzed unspiked and spiked. The developed method was partially validated as per the ICH recommended guidelines11 and used for determination of the minimum limit of detection and quantification of glycols.
Five structural formulas of glycols, all drawn in black on a white background. Starting from the top left, ethylene glycol (EG) is depicted as a two-carbon chain with a hydroxyl group (OH) attached to each carbon. Next to it, propylene glycol (PG) has a three-carbon chain with hydroxyl groups on the first and second carbons. To its right, diethylene glycol (DEG) displays two ethylene units connected by an oxygen atom (an ether linkage), with hydroxyl groups at each end. On the bottom left, triethylene glycol (TEG) extends this pattern with three ethylene units linked by two oxygen atoms, again capped by hydroxyl groups at both ends. Finally, at the bottom right, glycerin (Gly) is shown as a three-carbon backbone, each carbon bearing a hydroxyl group.
Chemical structures of ethylene glycol, propylene glycol, diethylene glycol, glycerin, and triethylene glycol.
Experimental
Standard preparation
The glycol standards were prepared as single component solutions of ethylene glycol (EG), propylene glycol (PG), diethylene glycol (DEG), glycerin (Gly), triethylene glycol (TEG) according to the following procedures using certified reference materials (CRMs) and methanol/water (95:5, v/v) as a diluent.
  • Stock solutions (500 µg/mL): Weigh and transfer about 10 mg of glycol substance into a 20 mL volumetric flask, add about 10 mL of diluent and sonicate for 5 minutes, make up to volume with diluent and shake to mix thoroughly. The concentration of the glycol in the resulting solution is 500 µg/mL.
  • Standard solution (SS, 25 µg/mL): Transfer 500 µL of each stock solution into a 10 mL volumetric flask, add 5 mL of diluent, and sonicate for 5 minutes. Then make up the volume with the diluent and mix thoroughly.
Calibration Solutions
Calibration solutions were prepared as single component solutions using the following dilution schemes:
  • Ethylene Glycol: Transfer 50, 100, 250, 500, 750, and 1000 µL of ethylene glycol standard stock solution into individual 10 mL volumetric flasks and make up to volume with diluent. Sonicate for 5 minutes and mix thoroughly.
  • Other glycols: Transfer 5, 10, 50, 100, 250, 500, 750, and 1000 µL of glycol standard stock solution into individual 10 mL volumetric flasks and make up to volume with diluent. Sonicate for 5 minutes and mix thoroughly. 0.25, 0.5, 2.5, 5.00, 12.50, 25.00, 37.50, 50.00 µg/mL
Sample Preparation
Two commercially available syrups (A & B) were used for the assessments. Cough syrup formulation B was rich in texture and more viscous as compared to formulation A. The formulation A is a combination medication designed to relieve cough accompanied by mucus, whereas formulation B is a combination cough syrup formulated to relieve dry cough and associated symptoms such as nasal congestion, sneezing, and throat irritation.
Syrup sample: Transfer 12 mg of commercially available cough syrup into a 10 mL volumetric flask, add 5 mL of diluent and sonicate for 5 minutes. Fill up to the mark with diluent and mix thoroughly.
Spiked Samples
  • Spiked sample 1- PG & Gly (20.8 mg/g): Weigh 12 mg of syrup sample A into a 10 mL volumetric flask and add 500 µL of each of PG and Gly stock solutions. Make up to mark with diluent, mix thoroughly, and sonicate for 5 minutes. The solution contains 25 µg/mL of each glycol.
  • Spiked sample 2- PG & Gly (41.7 mg/g): Weigh 12 mg of syrup sample A into a 10 mL volumetric flask and add 1000 µL of each of the PG and Gly stock solutions. Make up to mark with diluent, mix thoroughly, and sonicate for 5 minutes. The solution contains 50 µg/mL of each glycol.
  • Spiked Sample 3- PG, DEG, TEG, Gly (20.8 mg/g): Weigh 12 mg of syrup sample B into a 10 mL volumetric flask, and add 500 µL of each of PG, DEG, TEG, and Gly stock solutions. Make up to mark with diluent, mix thoroughly, and sonicate for 5 minutes. The solution contains 50 µg/mL of each glycol.
  • Spiked sample 4- PG, DEG, TEG, Gly (41.7 mg/g): Weigh and transfer 12 mg of commercially available sample B to a 10 mL volumetric flask, and add 1000 µL of each of PG, DEG, TEG, and Gly stock solutions. Make up to mark with diluent, mix thoroughly, and sonicate for 5 minutes. The solution contains 50 µg/mL of each glycol.
GC-MS Analysis
The GC-MS analysis (Table 1) was performed on an intermediate polarity SPB®-624 column. Depending upon the fragmentation recorded for each individual glycols, the selection of specific target ions and reference ions was selected for scanning at a defined retention time. Selection of the retention time was done as per the separation acquired by the developed method on the selected column (Table 2).
GC-MS Parameters
Instrument
Agilent 7000 Series Triple Quad GC-MS with ALS
Column:
SPB®-624, 60 m × 0.32 mm I.D., df 1.80 μm (24251)
Oven:
Initial temperature (°C)
Temp. ramp (°C/min)
Final temp. (°C)
Hold time at final temperature (min)
60
-
60
3
60
10
150
3
150
15
250
5
Inj. temp.:
240 ̊C
Carrier gas:
Helium, 1.2 mL/min
Detector:
MS/MS
MSD interface:
285 °C
Injection:
0.6 µL splitless
Liner:
Spitless single taper, wool packed
Run time:
26.7 min
Samples:
Standards, unspiked and spiked samples all in methanol/water (95:5, v/v)
MS Conditions
Acquisition:
Automatic Liquid Sampler
Quench gas:
Helium
Solvent delay:
12 minutes
MS Source temperature
270 °C
Quad Temperature:
150 ̊C
Electron energy:
70 eV
Dwell time:
100 ms
Quenching gas flow rate:
2.25 mL/min
MS measurement mode:
SIM
MS mode:
Splitless
Solvent delay:
12 min
Ion source:
EI
Gain factor:
10
Scan type:
MS1 SIM (see Table 2)
Table 1.GC-MS conditions used for glycol determination
Peak No.
Compound
Retention time (min)
Target Ion m/z (Quantifier)
Reference Ion m/z
(Qualifier)
1
Ethylene glycol
12.0
43.0
62.0
61.1
46.1
44.0
2
Propylene glycol
13.5
45.1
76.1
61.0
57.0
43.1
3
Diethylene glycol
15.0
75
76.0
45.1
44.1
43.1
4
Glycerin
20.2
61
66.0
62.1
44.1
43.1
5
Triethylene Glycol
23.5
89
87.0
75.0
58.0
45.1
Table 2.MS SIM mode parameter (scan type MS1 SIM) used for glycol determination
Results & Discussion
The developed method applying an SPB®-624 (60 m × 0.32 mm I.D., df 1.80 μm) capillary GC column and splitless injection showed good retention and sufficient resolution of the analytes in scope. A representative chromatogram of an analyte mixture at 25 µg/mL each is shown in Figure 1.
A GC-MS chromatogram plotted with retention time in minutes on the x-axis, ranging from 12 to 27 minutes, and intensity in counts on the y-axis, scaling up to approximately 5.9 times ten to the power of eight. The chromatogram trace is a continuous line that rises to form five distinct peaks across the time axis. The first small peak, labeled “1,” appears just after 13 minutes and corresponds to ethylene glycol. A sharper, taller peak labeled “2,” for propylene glycol, follows at around 14 minutes. The largest peak on the chromatogram is labeled “3,” representing diethylene glycol, eluting near 20 minutes. Shortly after, another prominent peak labeled “4,” indicating glycerin, appears just before 21 minutes. The final notable peak, labeled “5,” stands out around 24 minutes and corresponds to triethylene glycol. Between and after these peaks, the trace returns close to the baseline, indicating minimal other detected compounds. The graph clearly demonstrates separation of the five glycol compounds in the standard mixture, each peak distinctly numbered above its apex to aid identification.
Figure 1.GC-MS chromatogram (SIM) for the standard solution (SS) mixture containing ethylene glycol (1), propylene glycol (2), diethylene glycol (3), glycerin (4), and triethylene glycol (5), prepared in diluent at 25 µg/mL each compound.
Calibration
The quantification was based on an external calibration using single reference standard solutions as described in the experimental section. The GC-MS method linearity for propylene glycol, diethylene glycol, glycerin, and triethylene glycol was established in the concentration range of 0.25 to 50 μg/mL, and for ethylene glycol 2.5-50 µg/mL. In Table 3 the experimental results, including the linearity (R2 ≥ 0.996) and the limits of detection (LOD) and limits of quantification (LOQ) are summarized. Based on the developed method’s linearity study, the minimum concentrations of glycols that can be detected in the injected solutions as per the ICH guidelines11 range from 2.46 µg/mL to 4.41 µg/mL, and the quantification can be done in the range of 7.46 µg/mL to 13.37 µg/mL. 
Glycols
Concentration range (μg/mL)
R2
LOD (µg/mL)
LOQ (µg/mL)
Ethylene Glycol (EG)
2.5 – 50.0
0.996
4.41
13.37
Propylene Glycol (PG)
0.25 –50.0
0.9974
3.44
10.43
Diethylene Glycol (DEG)
0.25 – 50.0
0.9969
3.73
11.31
Glycerin (Gly)
0.25 – 50.0
0.9987
2.46
7.46
Triethylene Glycol (TEG)
0.25 – 50.0
0.9983
2.8
8.48
Table 3.Calibration and sensitivity data for all glycols determined. 8 calibrants used for each analyte, except EG, with 6 calibrants
Recovery Determination
For recovery determinations, four solutions were prepared with two spike concentration levels, 20.8 mg/g and 41.7 mg/g (25 and 50 µg/mL in the resulting injection solutions), and two sets of compounds (Tables 4 & 5) and two cough syrups (A & B). For recovery calculations, the existing glycol background was subtracted. The recovery for spiked syrup A (PG & Gly) ranged from 84.4 to 93.2%, for spiked syrup B (PG, DEG, Gly, TEG) 77.0 to 103.6%.
Compound
Concentration determined (µg/mL)
Recovery
Spiked sample 1 (20.8 mg/g -> 25 µg/mL)
Propylene Glycol
23.3
93.2%
Glycerin
21.1
84.4%
Spiked sample 2 (41.7 mg/g -> 50 µg/mL)
Propylene Glycol
45.1
90.2%
Glycerin
42.8
85.6%
Table 4.Results of recovery determination using syrup sample A spiked with PG and Gly at 20.8 µg/g and 41.7 µg/g (conc in injected solution 25 and 50 µg/mL) of each component.
Compound
Concentration determined (µg/mL)
Recovery
Spiked sample 3 (20.8 mg/g -> 25 µg/mL) 
Propylene Glycol
21.0
84.0%
Diethylene Glycol
25.9
103.6%
Glycerin
19.3
77.2%
Triethylene Glycol
19.8
79.2%
Spiked sample 4 (41.7 mg/g -> 50 µg/mL)
Propylene Glycol
38.7
77.4%
Diethylene Glycol
44.2
88.4%
Glycerin
38.9
77.8%
Triethylene Glycol
38.5
77.0%
Table 5.Results of recovery determination using syrup sample B spiked with PG, DEG, Gly, and TEG at 20.8 µg/g and 41.7 µg/g (conc. in injected solution 25 and 50 µg/mL) of each component.
Reproducibility
The system repeatability was demonstrated by injecting five replicates of the standard solution (SS) containing mixture of glycol standards of ethylene glycol (EG), propylene glycol (PG), diethylene glycol (DG), glycerin (Gly), triethylene glycol (TG), prepared in diluent with 25 μg/mL each compound (see Figure 1 and for diluent blank Figure 2) The determined RSD ranged from 1.02-3.85% (Table 6).
GC-MS chromatogram plotted with retention time in minutes on the x-axis, ranging from 12 to 27, and intensity in counts on the y-axis, which scales up to approximately 5.9 times ten to the power of eight. A single thin line traces across the graph, mostly flat and close to the baseline, indicating very low signal intensity throughout the run. There are only two small, shallow peaks that rise slightly above the baseline: the first appears just after 12 minutes, and the second, slightly larger but still modest, occurs around 21 minutes. Unlike chromatograms showing multiple pronounced peaks, this plot is largely featureless, highlighting the near absence of detectable compounds in the sample. The clean, almost baseline-level trace indicates that the diluent mixture of methanol and water contains minimal impurities or interfering substances under the conditions of this analysis.
Figure 2.GC-MS chromatogram (TIC) for diluent (blank) containing methanol and water (95:5 v/v).
Replicates N=5
EG
PG
DEG
GLY
TEG
Mean Area Counts
(31.7 ± 0.3) × 10⁶
(50.6± 0.7) × 107
(82.4± 1.9) × 107
(65.6± 1.9) × 107
(61.7± 2.3) × 107
% RSD
1.02
1.43
2.34
2.97
3.85
Table 6.Repeatability data obtained for reference standard mixture containing mixture of glycol standards of ethylene glycol (EG), propylene glycol (PG), diethylene glycol (DEG), triethylene glycol (TEG), and glycerin (Gly) as depicted in Figure 1
Original Sample Analysis
The method was applied for analyzing two different commercially available cough syrup samples (A & B) solutions (see Figure 3). Table 7 shows the results for the analysis of glycols present in the two commercially available cough syrup samples (A & B).
A GC-MS chromatogram plotted with retention time in minutes on the x-axis, ranging from 12 to 27, and intensity in counts on the y-axis, reaching up to about 2.4 times ten to the power of eight. A single chromatographic trace runs across the plot, starting near the baseline and producing a series of peaks. The first noticeable peak, labeled “2,” appears at around 14 minutes and is relatively narrow, corresponding to propylene glycol. The trace then remains close to the baseline until around 21 minutes, where a sharp, prominent peak labeled “4” rises abruptly, indicating the presence of glycerin. Following this, the trace shows multiple small adjacent rises and falls, forming a cluster of minor overlapping peaks just after the glycerin peak, before settling back to the baseline by around 24 minutes. The overall chromatogram reveals that this cough syrup sample primarily contains propylene glycol and glycerin, with the labeled peaks distinctly standing out against the otherwise low background signal.
A GC-MS chromatogram with retention time in minutes displayed on the x-axis from 12 to 27, and intensity in counts on the y-axis rising up to approximately 2.4 times ten to the power of eight. A single smooth trace runs across the graph, starting near the baseline and producing several distinct peaks. The first notable peak, labeled “2,” appears around 14 minutes and corresponds to propylene glycol, showing as a sharp and narrow rise. Further along, a smaller but clear peak labeled “3” emerges just before 20 minutes, indicating the presence of diethylene glycol. A prominent, tall, and narrow peak labeled “4,” representing glycerin, appears sharply at around 21 minutes, standing out as the highest point on the chromatogram. Shortly after, the trace shows a series of smaller, uneven humps, indicating minor unresolved compounds or matrix components. Toward 24 minutes, a modest peak labeled “5” rises distinctly, corresponding to triethylene glycol. The chromatogram line remains relatively stable between these peaks and returns to near baseline levels beyond 25 minutes.
Figure 3. GC-MS chromatograms of commercially available cough syrups (left) sample A and (right) sample B. (2), diethylene glycol (3) glycerin (4), and triethylene glycol (5). 
Peak No.
Name
Glycols in sample A
(mg/g)
Glycols in sample B
(mg/g)
1
Ethylene glycol
Not detected
Not detected
2
Propylene glycol
3.53
5.31
3
Diethylene glycol
Not detected
3.72
4
Glycerin
5.88
15.68
5
Triethylene glycol
Not detected
6.33
Table 7.Cconcentration of glycols present in two commercially available cough syrups samples represented in mg/g of sample.
Conclusion
This study presents a GC-MS/MS method for quantifying ethylene glycol, propylene glycol, diethylene glycol, triethylene glycol, and glycerin in formulations utilizing an SPB®-624 capillary column. The % RSD for system repeatability with a standard solution (SS) of the glycol mixture was well below 10% (1.02 to 3.85%). Derived from the external calibration data (R≥ 0.996), the GC methods were in the range of 2.46 µg/mL to 4.41 µg/mL. Two commercially available cough syrups were spiked with glycols and displayed recovery rates ranging from 77.2% to 103.6%. The investigated unspiked samples showed, besides a glycerin background for one and three other glycols respectively.
Related Products
GC Columns & Accessories
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Reagents & Solvents
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Certified Reference Materials (CRMs)
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
References
1.
Forkner MW, Robson JH, Snellings WM, Martin AE, Murphy FH, Parsons TE, Albin BA, Burgess LM, Proulx G. 2022. Glycols. Kirk‐Othmer Encyclopedia of Chemical Technology.1-37. https://doi.org/10.1002/0471238961.0520082506151811.a01.pub3
2.
Rebsdat S, Mayer D. 2000. Ethylene Glycol. Ullmann’s Encyclopedia of Industrial Chemistry. https://doi.org/10.1002/14356007.a10_101
3.
Propylene Glycol. [Internet]. Agency for Toxic Substances and Disease Registry. Available from: https://wwwn.cdc.gov/TSP/substances/ToxSubstance.aspx?toxid=240
4.
Lim TY, Poole RL, Pageler NM. 2014. Propylene Glycol Toxicity in Children. J Pediatr Pharmacol Ther.. 19(4):277-282. https://doi.org/10.5863/1551-6776-19.4.277
5.
Schep LJ, Slaughter RJ, Temple WA, Beasley DMG. 2009. Diethylene glycol poisoning. Clinical Toxicology. 47(6):525-535. https://doi.org/10.1080/15563650903086444
6.
Fikri E, Firmansyah YW. A Case Report of Contamination and Toxicity of Ethylene Glycol and Diethylene Glycol on Drugs in Indonesia. Environment and Ecology Research. 11(2):378-384. https://doi.org/10.13189/eer.2023.110211
7.
MohanaSundaram A, Padhi BK, Mohanty A, Shrestha S, Sah R. 2023. The silent epidemic of substandard and falsified medicines in low- and middle-income countries: heed lessons from the tragic deaths of children in Indonesia. 109(3):523-525. https://doi.org/10.1097/js9.0000000000000059
8.
Saied AA, Metwally AA, Dhama K. 2023. Gambian children’s deaths due to contaminated cough syrups are a mutual responsibility. International journal of surgery. 109(2):115-116. https://doi.org/10.1097/js9.0000000000000112
9.
U.S. FDA Guidance on Testing High-Risk Drug Components for Diethylene Glycol and Ethylene Glycol . [Internet].[updated 23 Apr 2025]. Available from: https://qualitymatters.usp.org/ensuring-product-safety-us-fda-guidance-testing-high-risk-drug-components
10.
USP-NF Propylene Glycol Monograph: USP47-NF42. Pharmacopeial Forum: Volume No. PF 33(2). [Internet]. Available from: https://online.uspnf.com/uspnf/document/1_GUID-689CC49B-D676-4822-9F97-1404FD00CF7C_4_en-US?source=Search%20Results&highlight=Propylene%20Glycol
11.
ICH Harmonised Guideline ICH Q2 (R2) – Validation of Analytical Procedures.. [Internet]. Available from: https://database.ich.org/sites/default/files/ICH_Q2%28R2%29_Guideline_2023_1130.pdf