ISOGRO® Amplify Recombinant Protein Expression
  • Decrease lag time by as much as 60%.
  • Maximize OD and recombinant protein expression.
  • Supplement with as little as 10% ISOGRO and improve the production of difficult to express proteins in E. coli.
Cardiac troponin cTnC(1-89) in pLysS
Figure 1.Cardiac troponin cTnC(1-89) in pLysS. Cells grown at 37 °C in shaker flasks. Red curve is ISOGRO supplemented minimal media and black curve is minimal media alone.
SDS-PAGE cTnC(1-89) cell lysates
Figure 2.SDS-PAGE cTnC(1-89) cell lysates. M-low molecular weight marker; 0= uninduced; 1=induced minimal media; 2=induced minimal media + ISOGRO; L=Lysozyme. Data provided by Dr. Paul R. Rosevear The Department of Molecular Genetics, Biochemistry and Microbiology, University of Cincinnati Medical Center, Cincinnati, OH.
Procedure for 10% ISOGRO Supplementation of M9 Minimal Media:
  1. Inoculate E. coli BL21(DE3) pLysS/(cTnC 1-89) in Luria Bertani Broth (LB) containing Carbenicillin (Carb) and Chloramphenicol (Chl).
  2. Grow at 37 °C with shaking at 250 rpm until slightly turbid.
  3. Transfer 1mL to 50 mL of LB/Carb/Chl and grow at 37°C. Harvested cells at OD600 nm ~ 0.9 by centrifugation.
  4. Harvest cells at OD600 nm ~ 0.9 by centrifugation.
  5. Resuspend starter culture in 50 mL of sterile filtered minimal media (pH 7.0) supplemented with 1 g/L of ISOGRO®-13C,15N powder growth medium and containing the following:
  1. Inoculate 50 mL starter culture into 1 L of minimal media supplemented with 10% ISOGRO.
  2. Grow at 37 °C with shaking at 250 rpm.
  3. Induce at OD600 nm ~ 0.9 by the addition of isopropyl b-D-1-thiogalactopyranoside (IPTG) to a final concentration of 0.1 mM.
  4. Harvest cells eight hours post-induction.
  5. Analyze protein production by SDS-PAGE.
Diminish lag time by > 60% and the number of hours required to attain high cell density by > 45% with the inclusion of 10% ISOGRO in M9.