OAMLS compounds are conveniently provided at 5µg per well, enough for multiple injections. These are high purity (>95%) compounds supplied in an economical, ready to use format. Compounds are arranged in a 96-well polypropylene plate. Metabolite descriptors provided with product include: Name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Substance ID.