Product Name
SpheriCal™ Neat Peptide High, suitable for MALDI-MS
Product Number
PFS22
Product Brand
SIAL
CAS Number
Molecular Weight
Test | Specification |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Powder or Solid |
| SUITABILITY TEST MALDI-MS | Corresponds |
| APPLICATION TEST | Dendritic Calibrants PFS3A : PFS3B : PFS3C : PFS3D = 1:1:1:1 |
| EXPIRY PERIOD | 36 MONTHS |