Skip to Content
Merck
Nutrient Agar GranuCult®, ISO 6579, ISO 10273, ISO 19250, ISO 21528, APHA, suitable for microbiology

Nutrient Agar

Nutrient Agar GranuCult®, ISO 6579, ISO 10273, ISO 19250, ISO 21528, APHA, suitable for microbiology

Products

Product Category
Brand
Physical Form
Quality Segment
Application
Sterility
Technique