Skip to Content
Merck

CH4182077450

ChemBridge Corporation

3-[(4-chlorophenyl)thio]-1-propanamine

Synonym(s):

3-[(4-chlorophenyl)sulfanyl]propylamine

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
201.72
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C9H12ClNS/c10-8-2-4-9(5-3-8)12-7-1-6-11/h2-5H,1,6-7,11H2

Inchi Key

VVSRYXXUZUUHDJ-UHFFFAOYSA-N

Storage temp.

RT

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

liquid

MSDS

https://www.hit2lead.com/sds/4032953_sds.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service