Merck
All Photos(1)

58310C

SAFC

Sodium hydroxide solution

0.5 M, sterile; 0.1 μm filtered, ready-to-use-bag, liquid

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:

vapor pressure

3 mmHg ( 37 °C)

Quality Level

sterility

sterile; 0.1 μm filtered

form

liquid

concentration

0.5 M

impurities

≤1 EU/mL Endotoxin

pH

13.0-13.8 (25 °C±0.5)

conductivity

89.4-109.3 mS/cm at 25 °C±0.5

storage temp.

15-25°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

  • Manufactured in an ISO 9001:2015 certified facility
  • A 12-month shelf life from the date of production (with a target of 24 months)
  • Manufactured using Emprove® raw materials (complying with Ph. Eur. and most with USP)
  • Ready-to-use bags with standard MPC/MPX connectors
  • Liquid Solutions MyWay products are intended for bioprocessing and further manufacturing uses

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service