VN
EN
Applications
Products
Services
Documents
Support
Home
Search Results
Silver nitrate solution
(1)
Silver nitrate solution
Empirical Formula (Hill Notation):
AgNO
3
CAS Number:
7761-88-8
Molecular Weight:
169.87
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Showing 1-1 of 1
Page 1 of 1
Page 1 of 1
Compare
Product Number
Product Description
Pricing
85193
Silver nitrate solution,
2.5 % (w/v) AgNO
3
in H
2
O
Expand
View Pricing
Page 1 of 1
Page 1 of 1