VN
EN
Applications
Products
Services
Support
Home
Search Results
Validation and uncertainty estimation of analytical method for determination of benzene in beverages.
Validation and uncertainty estimation of analytical method for determination of benzene in beverages.
Eurasian Journal of Analytical Chemistry (2011)
Styarini D, et al.