Merck
  • Mechanistic Study on the Photogeneration of Hydrogen by Decamethylruthenocene.
Mechanistic Study on the Photogeneration of Hydrogen by Decamethylruthenocene.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-07-10)
Lucie Rivier, Pekka Peljo, Sunny Maye, Manuel A Méndez, Heron Vrubel, Laurent A C Vannay, Clémence Corminboeuf, Micheál D Scanlon, Hubert H Girault
ABSTRACT
Detailed studies on hydrogen evolution by decamethylruthenocene ([Cp*2 RuII ]) highlighted that metallocenes are capable of photoreducing hydrogen without the need for an additional sensitizer. Electrochemical, gas chromatographic, and spectroscopic (UV/Vis, 1 H and 13 C NMR) measurements corroborated by DFT calculations indicated that the production of hydrogen occurs by a two-step process. First, decamethylruthenocene hydride [Cp*2 RuIV (H)]+ is formed in the presence of an organic acid. Subsequently, [Cp*2 RuIV (H)]+ is reversibly reduced in a heterolytic reaction with one-photon excitation leading to a first release of hydrogen. Thereafter, the resultant decamethylruthenocenium ion [Cp*2 RuIII ]+ is further reduced with a second release of hydrogen by deprotonation of a methyl group of [Cp*2 RuIII ]+ . Experimental and computational data show spontaneous conversion of [Cp*2 RuII ] to [Cp*2 RuIV (H)]+ in the presence of protons. Calculations highlight that the first reduction is endergonic (ΔG0 =108 kJ mol-1 ) and needs an input of energy by light for the reaction to occur. The hydricity of the methyl protons of [Cp*2 RuII ] was also considered.
Sigma-Aldrich
Ferrocenium hexafluorophosphate, 97%
Sigma-Aldrich
Lithium tetrakis(pentafluorophenyl)borate ethyl etherate