Merck
  • Transcription Factor T-bet Attenuates the Development of Elastase-induced Emphysema in Mice.
Transcription Factor T-bet Attenuates the Development of Elastase-induced Emphysema in Mice.
American journal of respiratory cell and molecular biology (2019-04-10)
Shigen Hayashi, Yosuke Matsuno, Yoshiya Tsunoda, Hirofumi Sakurai, Takumi Kiwamoto, Yuko Morishima, Yukio Ishii, Keigyou Yoh, Satoru Takahashi, Nobuyuki Hizawa
ABSTRACT
Chronic obstructive pulmonary disease (COPD) is a progressive lung disease characterized by peripheral airways inflammation and emphysema. Emerging evidence indicates a contribution of both innate and adaptive immune cells to the development of COPD. Transcription factor T-bet modulates the function of immune cells and therefore might be involved in the pathogenesis of COPD. To elucidate the role for T-bet in elastase-induced emphysema, pathological phenotypes were compared between wild-type and T-bet -/- mice. T-bet -/- mice demonstrated enhanced emphysema development on histological analyses, with higher values of mean linear intercept and dynamic compliance relative to wild-type mice. The number of neutrophils in BAL fluids, lung IL-6 and IL-17 expression, and the proportion of CD4+ T cells positive for IL-17 or retinoic acid receptor-related orphan receptor-γt were higher in T-bet -/- mice than in wild-type mice. Although T-bet downregulates cytokine expression in bone marrow-derived macrophages and MH-S cells, a murine alveolar cell line, depending on the surrounding environment, IL-6 expression in alveolar macrophages isolated from elastase-treated mice was not dependent on T-bet. Coculture of bone marrow-derived macrophages and CD4+ T cells revealed that T-bet regulation of IL-17 expression was dependent on CD4+ T cells. Neutralizing antibodies against IL-6R or IL-17 ameliorated the development of emphysema in T-bet -/- mice. In conclusion, we demonstrate that T-bet ameliorates elastase-induced emphysema formation by modulating the host immune response in the lungs.