Merck
  • Primary amino acid profiles of Greek white wines and their use in classification according to variety, origin and vintage
Primary amino acid profiles of Greek white wines and their use in classification according to variety, origin and vintage
Food Chemistry (2003)
Soufleros EH, et al.