Merck
  • High level expression and crystallization of recombinant human cathepsin S.
High level expression and crystallization of recombinant human cathepsin S.
Protein science : a publication of the Protein Society (1996-04-01)
D Brömme, M E McGrath
ABSTRACT
We have expressed active human cathepsin S to 60 mg/L in Sf9 cells using a baculovirus system. Production of milligram quantities has facilitated crystallographic studies to determine the structure of this enzyme, which has unique properties among lysosomal cysteine proteinases. Recombinant, irreversibly inhibited cathepsin S was crystallized from ammonium phosphate at 17 degrees C. The crystals diffract to at least 2.3 A, and belong to the orthorhombic crystal system with a primitive lattice. Approximate cell dimensions are: a = 37.7 A, b = 73.9 A, and c = 106.7 A. There is most likely one molecule per asymmetric unit.
Sigma-Aldrich
Cathepsin S Active human, recombinant, expressed in FreeStyle 293-F cells, ≥90% (SDS-PAGE)
Pricing and availability is not currently available.