Merck
  • Defective responses to oxidative stress in protein l-isoaspartyl repair-deficient Caenorhabditis elegans.
Defective responses to oxidative stress in protein l-isoaspartyl repair-deficient Caenorhabditis elegans.
Mechanisms of ageing and development (2009-08-18)
Shilpi Khare, Tara Gomez, Carole L Linster, Steven G Clarke
ABSTRACT
We have shown that Caenorhabditis elegans lacking the PCM-1 protein repair l-isoaspartyl methyltransferase are more sensitive to oxidative stress than wild-type nematodes. Exposure to the redox-cycling quinone juglone upon exit from dauer diapause results in defective egg-laying (Egl phenotype) in the pcm-1 mutants only. Treatment with paraquat, a redox-cycling dipyridyl, causes a more severe developmental delay at the second larval stage in pcm-1 mutants than in wild-type nematodes. Finally, exposure to homocysteine and homocysteine thiolactone, molecules that can induce oxidative stress via distinct mechanisms, results in a more pronounced delay in development at the first larval stage in pcm-1 mutants than in wild-type animals. Homocysteine treatment also induced the Egl phenotype in mutant but not wild-type nematodes. All of the effects of these agents were reversed upon addition of vitamin C, indicating that the developmental delay and egg-laying defects result from oxidative stress. Furthermore, we have demonstrated that a mutation in the gene encoding the insulin-like receptor DAF-2 suppresses the Egl phenotype in pcm-1 mutants treated with juglone. Our results support a role of PCM-1 in the cellular responses mediated by the DAF-2 insulin-like signaling pathway in C. elegans for optimal protection against oxidative stress.
Sigma-Aldrich
Methyl viologen dichloride hydrate, 98%