Merck
  • Reasoned opinion on the toxicological properties and maximum residue levels (MRLs) for the benzimidazole substances carbendazim and thiophanate-methyl.
Reasoned opinion on the toxicological properties and maximum residue levels (MRLs) for the benzimidazole substances carbendazim and thiophanate-methyl.
EFSA journal. European Food Safety Authority (2021-09-02)
Giulia Bellisai, Giovanni Bernasconi, Alba Brancato, Luis Carrasco Cabrera, Lucien Ferreira, German Giner, Luna Greco, Samira Jarrah, Aija Kazocina, Renata Leuschner, Jose Oriol Magrans, Ileana Miron, Stefanie Nave, Ragnor Pedersen, Hermine Reich, Silvia Ruocco, Miguel Santos, Alessia Pia Scarlato, Anne Theobald, Benedicte Vagenende, Alessia Verani
ABSTRACT
In compliance with Article 43 of Regulation (EC) No 396/2005, EFSA received from the European Commission a mandate to provide its reasoned opinion on the toxicological properties and maximum residue levels (MRLs) for the benzimidazole substances carbendazim and thiophanate-methyl. Specifically, EFSA was asked to assess whether thiophanate-methyl or carbendazim have clastogenic potential and, in case clastogenic potential can be excluded, to derive toxicological reference values necessary for consumer risk assessment and assessment of maximum residue levels (MRLs). Although these active substances are no longer authorised within the European Union, MRLs were established by the Codex Alimentarius Commission (codex maximum residue limits; CXLs), and import tolerances are in place. Based on the assessment of the available data, toxicological reference values and MRL proposals were derived and a consumer risk assessment was carried out. Some information required by the regulatory framework was found to be missing and a possible acute risk to consumers was identified. Hence, the consumer risk assessment is considered indicative only, all MRL proposals derived by EFSA still require further consideration by risk managers and measures for reduction of the consumer exposure should also be considered.
Product Description
Supelco
Carbendazim-(ring-4,5,6,7-d4), PESTANAL®, analytical standard