Merck
  • Synthesis and application of a new cleavable linker for "click"-based affinity chromatography.
Synthesis and application of a new cleavable linker for "click"-based affinity chromatography.
Organic & biomolecular chemistry (2009-12-22)
Felicetta Landi, Conny M Johansson, Dominic J Campopiano, Alison N Hulme
ABSTRACT
A new chemically-cleavable linker has been synthesised for the affinity-independent elution of biomolecules by classical affinity chromatography. This azo-based linker is shown to couple efficiently with "click" derivatised ligands such as biotin propargyl amide through a copper(I)-catalysed Huisgen 1,3-dipolar cycloaddition reaction. Binding to Affi-Gel matrices displaying ligands coupled to the new linker is both efficient and selective. The captured material may be readily released from the resin upon treatment with sodium dithionite. These mild elution conditions have allowed for the efficient isolation of the affinity partner from complex protein mixtures such as those found in fetal bovine serum.
Sigma-Aldrich
tert-Butyl 2-(4-{[4-(3-azidopropoxy)phenyl]azo}benzamido)ethylcarbamate, 95%
Sigma-Aldrich
4-(4′-Hydroxyphenylazo)benzoic acid, 97%