Merck
1-Methylguanine and 7-methylguanine increase cell agglutinability.
Cell and tissue research (1985-01-01)
C Hanski, S J Kerr
ABSTRACT
1-Methylguanine and 7-methylguanine, both metabolic products of tRNA degradation, are known to induce transformation of Chinese hamster fibroblasts in culture. The effects of these compounds on the cell membrane have been studied by the method of Concanavalin A-mediated hemadsorption. 1-Methylguanine or 7-methylguanine induced a 50% increase of Con A-mediated hemadsorption within 20 hours of exposure of the cells to the agent at a concentration of 10(-5) M. This alteration was reversed within 13 days when the cells were grown in the control medium. Prolonged treatment with 1-methylguanine or 7-methylguanine resulted in changes which were only slowly reversed during growth of the cells in the control medium. The effect of the methylated purines on the cell membrane could be completely inhibited by simultaneous addition of dibutyryl-cAMP at a concentration of 10(-5) M. The possible mechanism of cell membrane alteration by methylated purines and its relevance to transformation in vitro are discussed.