Merck
  • Synthesis and exploration of novel curcumin analogues as anti-malarial agents.
Synthesis and exploration of novel curcumin analogues as anti-malarial agents.
Bioorganic & medicinal chemistry (2008-01-16)
Satyendra Mishra, Krishanpal Karmodiya, Namita Surolia, Avadhesha Surolia
ABSTRACT
Curcumin, a major yellow pigment and active component of turmeric, has been shown to possess anti-inflammatory and anti-cancer activities. Recent studies have indicated that curcumin inhibits chloroquine-sensitive (CQ-S) and chloroquine-resistant (CQ-R) Plasmodium falciparum growth in culture with an IC(50) of approximately 3.25 microM (MIC=13.2 microM) and IC(50) 4.21 microM (MIC=14.4 microM), respectively. In order to expand their potential as anti-malarials a series of novel curcumin derivatives were synthesized and evaluated for their ability to inhibit P. falciparum growth in culture. Several curcumin analogues examined show more effective inhibition of P. falciparum growth than curcumin. The most potent curcumin compounds 3, 6, and 11 were inhibitory for CQ-S P. falciparum at IC(50) of 0.48, 0.87, 0.92 microM and CQ-R P. falciparum at IC(50) of 0.45 microM, 0.89, 0.75 microM, respectively. Pyrazole analogue of curcumin (3) exhibited sevenfold higher anti-malarial potency against CQ-S and ninefold higher anti-malarial potency against CQ-R. Curcumin analogues described here represent a novel class of highly selective P. falciparum inhibitors and promising candidates for the design of novel anti-malarial agents.
Sigma-Aldrich
2,4-Dichlorophenylhydrazine hydrochloride, 98%
Sigma-Aldrich
2-Fluorophenylhydrazine hydrochloride, 97%