VN
EN
Applications
Products
Services
Support
Home
Search Results
The most stable conformer of benzyl alcohol.
The most stable conformer of benzyl alcohol.
Chemical Physics Letters (2008)
Miller BJ, et al.