- Spatiotemporal regulation of the Pak1 kinase.
Spatiotemporal regulation of the Pak1 kinase.
Biochemical Society transactions (2005-07-27)
M C Parrini, M Matsuda, J de Gunzburg
PMID16042564
ABSTRACT
Pak1 (p21-activated kinase 1) is a key regulator of the actin cytoskeleton, adhesion and cell motility. Such biological roles require a tight spatial and kinetic control of its localization and activity. We summarize here the current knowledge on Pak1 dynamics in vivo. Inactive dimeric Pak1 is mainly cytosolic. Localized interaction with the activators Cdc42-GTP and Rac1-GTP stimulates the kinase at the sites of cellular protrusions. Moreover, Pak1 is dynamically engaged into multiprotein complexes forming adhesions to the extracellular matrix. Cutting edge microscopy technologies on living cells are finally shedding light on the intricate spatiotemporal mechanisms regulating Pak1.
Sigma-Aldrich
PAK1/CDC42, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
PAK1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution