Merck
Pathways of root uptake and membrane transport of Cd
Environmental toxicology and chemistry (2016-10-30)
Lian-Zhen Li, Chen Tu, Long-Hua Wu, Willie J G M Peijnenburg, Stephen Ebbs, Yong-Ming Luo
ABSTRACT
Uptake and membrane transport of cadmium (Cd) in roots of the hyperaccumulator Sedum plumbizincicola X.H. Guo et S.B. Zhou ex L.H. Wu was characterized by assessing the impact of various inhibitors and ion channel blockers on Cd accumulation as well as the real-time net Cd
Product Description
Supelco
Cadmium ionophore I, Selectophore
Pricing and availability is not currently available.