Merck
  • B7-H4 overexpression is essential for early hepatocellular carcinoma progression and recurrence.
B7-H4 overexpression is essential for early hepatocellular carcinoma progression and recurrence.
Oncotarget (2017-11-09)
Fu-Biao Kang, Ling Wang, Dian-Xing Sun, Hai-Jun Li, Dong Li, Yan Wang, Ji-Wen Kang
ABSTRACT
B7-H4, another member of costimulatory molecule, has been shown to be overexpressed in multiple types of tumors, including hepatocellular carcinoma (HCC). However, the specific biological role of B7-H4 in HCC still needs to be further explored. In this study, we observed that B7-H4 was highly overexpressed in HCC tissues and cells, and its overexpression strongly correlated with patient's TNM stage, overall survival and early recurrence. Downregulation of B7-H4 significantly suppressed cell growth, invasion, and stemness of HCC by inducing apoptosis in the