Skip to Content
Merck

GF86309343

Copper

foil, thickness 0.075 mm, length 2 m, coil width 10 mm

Synonym(s):

CU000531

Sign In to View Organizational & Contract Pricing.

Select a Size

1 EA

R 5,685.96

R 5,685.96

List PriceR 5,802.00

Please contact Customer Service for Availability

Request a Bulk Order

About This Item

Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141711
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/Cu

SMILES string

[Cu]

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

form

foil

manufacturer/tradename

Goodfellow 863-093-43

resistivity

1.673 μΩ-cm, 20°C

L

2 m

coil width

10 mm

thickness

0.075 mm

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF89236750GF55298380GF63352795
manufacturer/tradename

Goodfellow 863-093-43

manufacturer/tradename

Goodfellow 892-367-50

manufacturer/tradename

Goodfellow 552-983-80

manufacturer/tradename

Goodfellow 633-527-95

form

foil

form

foil

form

foil

form

foil

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

bp

2567 °C (lit.)

bp

2567 °C (lit.)

bp

2567 °C (lit.)

bp

2567 °C (lit.)

resistivity

1.673 μΩ-cm, 20°C

resistivity

1.673 μΩ-cm, 20°C

resistivity

1.673 μΩ-cm, 20°C

resistivity

1.673 μΩ-cm, 20°C

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service