Skip to Content
Merck

MDCISTAGE-SL

Millicell® DCI XY Slide holder

Millicell® DCI Digital Cell Imager

Synonym(s):

Mechanical stage, Slide holder, XY mechanical stage

Sign In to View Organizational & Contract Pricing.

Select a Size

MDCISTAGE-SL

R 7,039.34

R 7,039.34

List PriceR 7,183.00

Please contact Customer Service for Availability


About This Item

UNSPSC Code:
41122601
NACRES:
NB.13

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

General description

MDCISTAGE_SL is an XY-mechanical stage that can be mounted on Millicell® DCI Digital Cell Imager (MDCI10000). It can be used to hold slides such as the hemocytometer slides for counting cells or histology slides for imaging histology sections.

Application

Cell culture, Cell counting, Histology

Features and Benefits

Dimensions: L x W x H = 12.7 x 12.7 x 5 inches

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cell counting protocol using a hemocytometer for monitoring cell viability. Discover other cell counting tools such as the Scepter™ 3.0 Cell Counter.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service