Merck
  • Loading effects of silver oxides upon generation of reactive oxygen species in semiconductor photocatalysis.
Loading effects of silver oxides upon generation of reactive oxygen species in semiconductor photocatalysis.
Physical chemistry chemical physics : PCCP (2008-05-14)
Shigeru Kohtani, Kazuhiro Yoshida, Toshiyasu Maekawa, Akihide Iwase, Akihiko Kudo, Hideto Miyabe, Ryoichi Nakagaki
ABSTRACT
Superoxide anion radical (O(2)(-*)) and OH radical generations in suspensions of Ag metal-, Ag(2)O-, or AgO-loaded TiO(2) and BiVO(4) photocatalysts in alkaline conditions (pH 12.0) were examined by means of a luminol chemiluminescence (CL) technique and a spin-trapping fluorescence one in which terephthalic acid reacts with an OH radical to afford the highly fluorescent 2-hydroxyterephthalic acid (TAOH), respectively. The observed luminol CL intensity was remarkably enhanced by the AgO loading on TiO(2) as well as BiVO(4). This can be explained by enhancement of O(2)(-*) production on the AgO-loaded photocatalysts caused by the synergetic effects on the thermocatalytic activity upon the AgO surface and the efficient electron-hole separation at the photocatalyst/AgO interface. On the other hand, loading effects of AgO on the TAOH formation were not so significant compared to those on the CL observation, though the TAOH formation rates for the TiO(2) samples were much larger than those for the BiVO(4) ones by about three orders of magnitude. The properties of O(2)(-*) and OH radical generations on these photocatalysts are discussed on the basis of the luminol CL kinetics and approximate band edge positions of TiO(2), BiVO(4), and silver oxides.
Sigma-Aldrich
2-Hydroxyterephthalic acid, 97%
Pricing and availability is not currently available.