Analytical Standards
TraceCERT® CRMs for quantitative NMR (qNMR)
We offer a set of NIST SRM traceable certified reference materials for use as internal standards in quantitative NMR studies. The offering includes qNMR standards for 1H, 31P and 19F NMR experiments.  
  • Manufactured according to ISO/IEC 17025 and ISO Guide 34
  • Certified Content by quantitative NMR (qNMR)
  • Superior level of accuracy, calculated uncertainties and lot-specific values
  • Traceability to NIST SRM
  • Comprehensive documentation delivered with the product
    (certification according to ISO Guide 31)
Quantitative NMR - Technical Details and TraceCERT® Certified Reference Materials (PDF, 1.5 MB)
Find product tables and spectra for all available qNMR CRMs below:
The tables give an overview of spectral shifts, solubility and relaxation times in different solvents of 16 1H-qNMR standards, 3 31P-qNMR standards and 3 19F-qNMR standards. They cover the entire spectral range, allowing the quantification of almost any organic molecule by qNMR. Solubility tests were done at room-temperature using commercially available NMR solvents. Tests were performed starting from 1 mg/ml up to 250 mg/ml (mg CRM/ml solvent). T1 relaxation times were recorded for the CRM only (c ≈ 5 to 20 mg/ml at 25°C), but may vary in the mixture. Therefore it is recommended to check the T1 delay prior to the qNMR experiment. (*with DCl or NaOD)
Standards for 1H quantitative NMR, TraceCERT®
PN Substance
D2O
CDCl3
DMSO-d6
CD3OD
CD3CN
δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml)
01380
Ethylene carbonate
4.5
5.5
>250
4.5
7.0
>250
4.5
2.7
>250
4.5
5.3
>250
4.5
2.0
>250
03826
Calcium formate
8.4
3.4
>250
-
-
-
-
-
-
-
-
-
-
-
-
06185
Benzoic acid
-
-
-
8.1
7.7
7.5
3.7
4.0
3.4
150
8.0
7.6
7.5
3.3
3.7
3.0
>250
8.0
7.6
7.5
4.3
4.4
3.9
>250
8.0
7.7
7.5
4.5
2.5
2.6
80
06856
Duroquinone
-
-
-
2.0
3.3
>250
2.0
3.1
30
2.0
4.0
10
2.0
5.4
20
07038
Dimethyl
terephthalate
-
-
-
8.1
4.0
3.6
1.8
160
8.1
3.9
2.9
1.1
20
8.1
4.0
4.4
2.4
4
8.1
3.9
4.9
2.6
20
14659
Potassium hydrogen phthalate
7.5
2.5
>250*
-
-
-
-
-
-
7.9
2.5
5
-
-
-
40384
1,2,4,5-Tetrachloro-  3-nitro-benzene
-
-
-
7.8
10.7
>250
8.5
12.6
>250
8.1
6.4
30
8.0
9.6
10
41867
Dimethyl sulfone
3.0
2.9
>250
3.0
2.7
80
3.0
2.4
>250
3.0
3.3
40
2.9
2.6
>250
42582
Ethyl 4(dimethyl-amino)benzoate
-
-
-
7.9
6.7
4.3
3.1
1.4
4.1
2.4
2.8
2.3
2.6
>250
7.8
6.7
4.2
3.0
1.3
2.7
1.4
1.8
1.5
2.1
>250
7.9
6.7
4.3
3.0
1.4
4.3
2.4
2.9
2.8
2.8
>250
7.9
6.7
4.3
3.0
1.3
5.6
3.7
4.1
3.6
3.6
50
50409
Thymol
-
-
-
7.1
6.8
6.6
3.2
2.3
1.3
3.8
4.5
4.8
4.3
3.1
1.9
>250
7.0
6.7
6.5
3.1
2.2
1.1
2.0
2.2
2.6
2.3
2.0
0.9
>250
7.0
6.6
6.5
3.2
2.2
1.2
3.7
4.4
5.8
4.0
2.8
1.8
>250
7.1
6.7
6.6
3.2
2.2
1.2
4.9
5.7
5.7
5.2
3.5
2.4
>250
55177
Benzyl benzoate
-
-
-
5.4
8.1
4.3
13.6
>250
5.4
8.0
2.2
8.8
>250
5.4
8.1
2.0
3.9
>250
5.4
8.1
1.0
2.4
>250
74599
1,3,5-Trimethoxy- benzene
-
-
-
6.1
3.8
4.7
2.2
>250
6.1
3.7
3.2
1.4
>250
6.1
3.8
4.8
2.7
>250
6.7
4.3
3.4
2.6
>250
74658
1,2,4,5-Tetramethyl-benzene
-
-
-
7.0
2.2
6.1
4.0
>250
6.9
2.1
4.7
2.6
10
6.9
2.2
5.9
4.3
10
6.9
2.2
7.7
5.0
50
89151
Dimethylmalonic acid
1.3
1.0
100
-
-
-
1.3
0.7
250
1.4
1.0
250
1.4
2.0
30
92816
Maleic acid
6.3
6.1
>250
-
-
-
6.3
3.0
>250
6.3
3.9
20
6.4
6.2
20
93074
Pentachlorobenzene
-
-
-
7.6
5.8
>250
8.2
13.8
15
7.9
8.2
9
7.8
12.2
15
94681
Methyl 3,5-dinitrobenzoate
-
-
-
9.3
9.2
4.1
8.0
6.1
2.6
>250
9.1
8.9
4.0
8.1
8.1
1.6
100
-
-
-
9.1
9.0
4.0
9.0
8.2
3.5
>250
*with DCl or NaOD
Standards for 31P quantitative NMR
PN Substance
D2O
CDCl3
DMSO-d6
CD3OD
CD3CN
δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml)
05498 Triphenyl phosphate - - - -17.7 2.7 >250 -17.3 1.2 >100 -17.5 3.1 >10 -17 4.3 >250
92214 Potassium phosphate monobasic 0.08 8.0 >250 - - - - - - - - - - - -
96708 Phosphonoacetic acid 15.7 4.6 >250 - - - 14.9 1.5 >250 17.7 2.9 >250 - - -
Standards for 19F quantitative NMR
PN Substance
D2O
CDCl3
DMSO-d6
CD3OD
CD3CN
δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml) δ
(ppm)
T1 (s) s (mg/ml)
07563 4,4’-Difluorobenzophenone
- - - -105.8 2.4 >250 -106.5 1.4 150 -108.1 2.8 30 -108.3 2.3 140
53396 2,4-Dichlorobenzotrifluoride
- - - -62.5 2.3 >250 -61.2 1.2 >250 -65.4 3.3 >250 -63 2.9 >250
80730 2-Chloro-4-fluorotoluene
- - - -115.8 4.4 >250 -115.3 3.3 >250 -117.7 4.8 >250 -117.3 4.7 >250