Discovery Zr PBD HPLC Columns
Discovery logo
Ordering Information
Discovery Zr-PBD HPLC columns
Discovery Zr-PBD consists of spherical, porous zirconia particles with a durable coating of polybutadiene. It operates via reversed-phase and ion-exchange modes. Discovery Zr-PBD complements the selectivity offering of the other zirconia-, silica-, and polymer-based Discovery phases, and allows the use of the full range of mobile phase pH from pH 1 to 13.
Particle:   Zirconia (ZrO2)
Bonded Phase:   Polybutadiene
Particle Shape:   Spherical
Particle Size:   3 and 5µm
Pore Size:   300Å
Surface Area:   30m2/g
pH Range:   1 to 13
Temperature Range:   up to 100°C in standard hardware (up to 150°C in custom all-steel hardware)
Unique retention and selectivity for method development at pH and temperature extremes
Good for bases, amines
Similar to ODS-silica
pH stable from 1-13
Thermally stable up to 100°C or higher
Here's the
A significant difference between Discovery Zr-PBD and C18-silica is that it can be used with basic pH mobile phases and elevated temperatures where basic analytes have better peak shape and higher efficiency. This is demonstrated in the separation of basic antihistamine compounds in the figure below.
figure g
Another example of the utility of Discovery Zr-PBD for basic compounds is shown in the separation of tricyclic antidepressants in the figure below.
figure h
Ordering Information
Phase ID (mm) Length (cm) Cat. No.
3µm Discovery Zr-PBD Columns
  2.1 5 65713-U
  2.1 7.5 65714-U
  2.1 15 65715-U
  4.6 5 65716-U
  4.6 7.5 65717-U
  4.6 15 65718-U
5µm Discovery Zr-PBD Columns
  2.1 5 65719-U
  2.1 15 65720-U
  4.6 5 65722-U
  4.6 15 65723-U
  4.6 25 65724-U
Description 2.1mm ID 4.0mm ID
1cm Supelguard Cartridges with Discovery Zr-PBD Packings
  3µm Particle Size
  2 Pack
65812-U 65814-U
  Kit triangle
65811-U 65813-U
  5µm Particle Size
  2 Pack
65816-U 65818-U
  Kit triangle
65815-U 65817-U
For 4.0mm ID or 4.6mm ID analytical columns
Kit includes a holder and one replaceable cartridge.
For additional information, click the pdf icons below.
view the pdf document Bulletin 932 - Discovery Zr: Method Development Guidelines (48 KB)
view the pdf documentCare and Use Guide for Discovery Zr-PBD (19KB)
How to Contact Supelco
Phone: 800-247-6628
  or 814-359-3441
fax: 800-325-5052
email: techservice@sial.com
Outside the USA: click here
Discovery, and Supelguard are trademarks of Sigma-Aldrich Co.