Milliflex Oasis® Filtration System for Bioburden and Pharmaceutical Water Testing
Unmatched comfort in microbial testing
Milliflex Oasis® system for pharmaceutical water and bioburden testingMembrane filtration is the method of choice for water and bioburden testing. We have taken time to closely observe your complete laboratory workflow to identify improvement areas and developed, hand-in-hand with users, a solution that simplifies and secures your microbial testing process.
Vacuum pump and filtration funnels
The Milliflex Oasis® vacuum pump is designed for biosafety cabinets and laminar flow hoods, with a small footprint, low weight and easy to decontaminate surfaces. The funnels standardize handling steps and reduce filtration time thanks to new drainage design. Our touch-free membrane transfer increases result reliability, independent from the operator.
With our new Milliflex Oasis® system, we raised comfort in bioburden testing to new levels:
Productivity
• Fast hardware set-up and 50%
  faster filtration time
• Hardware scalability: attach up to
  three pumps and filter 6 samples
  with a single push of a button
• Reduced decontamination time
  and maintenance, no need for
  autoclaving
Regulatory Compliance
• Compliance with USP, EP and JP
  recommendations
• Labels with 2D Code on the
  packaging: lot number and
  expiration date can be checked
  and recorded as mentioned in the
  USP <1117>
• Detachable label on
  primary/secondary package for
  easy transfer to log books
Result Reliability
• Operator independent
• Touch-free membrane transfer
• Perfect contact between the
  membrane and the media plate
• A check valve situated in the
  pump head prevents
  contamination from backflow
Milliflex® culture media
Ready-to-use solid media culture cassettes for use with the Milliflex Oasis® system for bioburden testingOur media plates are designed for perfect contact between membrane and culture media. The unique plate color coding offers the con¬trol of critical handling steps all along the workflow. The Milliflex Rack & Stack®  feature also includes a lockable lid for safe transportation and incubation. Together, these innova¬tions help to reduce the risk of false results.
Learn more.
Rapid detection methods
Rapid detection method for microbial testingThe Milliflex® Quantum system can reduce your time-to-result by up to one third compared to traditional methods, enabling faster release or corrective actions. It works perfectly with the Milliflex Oasis® system.
Learn more.
Milliflex Oasis® services
Our experienced scientists and engineers provide comprehensive services to ensure regulatory compliance as well as optimal testing workflows and equipment performance with our validation, training and maintenance services.
Learn more.
Materials
Vacuum pump
Description Quantity Catalogue Nos.
Milliflex Oasis® filtration pump, one system with two filtration heads and no power supply 1 MMSYSTMM1
Milliflex Oasis® filtration pump without filtration head and no power supply 1 MMSYSTVAC
Milliflex Oasis® filtration head 1 MMHEADMM1
Milliflex Oasis® filtration head gaskets replacement kit, including gaskets removal tool 5 MMGASKTMM
Milliflex Oasis® filtration head cover set 1 MMHEADCV1
Milliflex Oasis® drain tubing 1 MMDRNTUBE
Milliflex Oasis® internal flow sanitization kit, including one vacuum gauge 1 MMSANKIT1
Milliflex Oasis® internal flow sanitization gasket kit 5 MMGASKTSK
Milliflex Oasis® consumables for internal flow sanitization kit 24 MMSANSYFU
Milliflex Oasis® vacuum gauge for pump performance testing 1 MMGAUGEMM
Milliflex Oasis® electrical cable for connecting two pumps 1 MMCABLEMM
Milliflex Oasis® T connectors for tubings for connecting two pumps 3 MMTCNNECT
Milliflex Oasis® guides for electrical cable and tubing 20 MMTUGUIDE
Milliflex Oasis® membrane support removal tool 1 MMSUPREMV
Stickers and filtration head rings for customizing the Milliflex Oasis® pump 4 MMCUSTKIT
Milliflex® Quantum membrane transfer tool for Milliflex Oasis® system 1 MMHEADQU1
Milliflex® Quantum membrane transfer tool for Milliflex Oasis® gaskets replacement kit 5 MMGASKTQU
Milliflex® Quantum membrane removal tool for all Milliflex® and Milliflex Oasis® devices and media types 1 REMRACKMM
Customer validation protocol format A4 1 MM2PA4VP1
Customer validation protocol format letter 1 MM2PLTVP1
Filtration funnels
Description Quantity Catalogue Nos.
Milliflex Oasis® 100 mL funnel, 0.45 µm white gridded, mixed cellulose esters (MCE) membrane 24 MMHAWG124
Milliflex Oasis® 250 mL funnel, 0.45 µm white gridded, mixed cellulose esters (MCE) membrane 24 MMHAWG224
Milliflex Oasis® 100 mL funnel, 0.45 µm black gridded, mixed cellulose esters (MCE) membrane 24 MMHABG124
Milliflex Oasis® 100 mL funnel, 0.22 µm white gridded, mixed cellulose esters (MCE) membrane 24 MMGSWG124
Milliflex Oasis® 100 mL funnel, 0.45 µm white plain PVDF Durapore® membrane 24 MMHVWP124
Milliflex Oasis® 250 mL funnel, 0.45 µm white plain, PVDF Durapore® membrane 24 MMHVWP224
Milliflex Oasis® rapid 100 mL funnel, 0.45 µm white plain, PVDF Durapore® membrane 24 MMHVMFX24
Milliflex Oasis® funnel without membrane, growth promotion accessory 24 MMRECVERY